Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 201970144-201970618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TIMM17A
Entrez Gene ID
10440
Gene Name
translocase of inner mitochondrial membrane 17 homolog A (yeast)
Gene Aliases
TIM17, TIM17A
UniGene
Hs.20716
Gene Chromosome Location
Chr.1: 201955491-201970661 on Build GRCh38
Transcripts
NM_006335.2

SNP Detail

SNP ID Chromosome Location
rs958760312 Chr.1: 201970397-201970397 on Build GRCh38
rs978721140 Chr.1: 201970371-201970371 on Build GRCh38
rs774324388 Chr.1: 201970468-201970468 on Build GRCh38
rs114598968 Chr.1: 201970297-201970297 on Build GRCh38
rs776119956 Chr.1: 201970372-201970372 on Build GRCh38
rs770277753 Chr.1: 201970364-201970364 on Build GRCh38
rs1018738914 Chr.1: 201970547-201970547 on Build GRCh38
rs552825392 Chr.1: 201970370-201970370 on Build GRCh38
rs113526422 Chr.1: 201970351-201970351 on Build GRCh38
rs886140412 Chr.1: 201970234-201970234 on Build GRCh38
rs1047400024 Chr.1: 201970222-201970222 on Build GRCh38
rs1022481172 Chr.1: 201970441-201970441 on Build GRCh38
rs186057725 Chr.1: 201970442-201970442 on Build GRCh38
rs747241691 Chr.1: 201970540-201970541 on Build GRCh38
rs984667583 Chr.1: 201970264-201970264 on Build GRCh38
rs76379110 Chr.1: 201970274-201970274 on Build GRCh38
rs114953472 Chr.1: 201970423-201970423 on Build GRCh38
rs927396270 Chr.1: 201970374-201970374 on Build GRCh38
rs762077690 Chr.1: 201970540-201970540 on Build GRCh38
rs965698688 Chr.1: 201970556-201970556 on Build GRCh38
rs990671734 Chr.1: 201970398-201970398 on Build GRCh38
rs1005872152 Chr.1: 201970240-201970240 on Build GRCh38
rs1128660 Chr.1: 201970225-201970225 on Build GRCh38
rs919213428 Chr.1: 201970192-201970192 on Build GRCh38
rs114298627 Chr.1: 201970332-201970332 on Build GRCh38
rs61821491 Chr.1: 201970216-201970216 on Build GRCh38
rs1007318269 Chr.1: 201970534-201970534 on Build GRCh38
rs146125138 Chr.1: 201970366-201970366 on Build GRCh38
rs954991140 Chr.1: 201970282-201970282 on Build GRCh38
rs941689136 Chr.1: 201970452-201970453 on Build GRCh38
rs1030874912 Chr.1: 201970280-201970280 on Build GRCh38
rs544518155 Chr.1: 201970491-201970491 on Build GRCh38
rs996293522 Chr.1: 201970532-201970532 on Build GRCh38
rs917709716 Chr.1: 201970408-201970408 on Build GRCh38
rs948178539 Chr.1: 201970294-201970294 on Build GRCh38
rs572278043 Chr.1: 201970382-201970382 on Build GRCh38
rs1020489292 Chr.1: 201970302-201970302 on Build GRCh38
rs763380433 Chr.1: 201970393-201970393 on Build GRCh38
rs555619143 Chr.1: 201970176-201970176 on Build GRCh38
rs900222987 Chr.1: 201970273-201970273 on Build GRCh38
rs530781856 Chr.1: 201970465-201970465 on Build GRCh38
rs906538234 Chr.1: 201970353-201970353 on Build GRCh38
rs757923136 Chr.1: 201970514-201970514 on Build GRCh38
rs965600164 Chr.1: 201970333-201970333 on Build GRCh38
rs1128672 Chr.1: 201970290-201970290 on Build GRCh38
rs907780160 Chr.1: 201970451-201970451 on Build GRCh38
rs968716168 Chr.1: 201970327-201970327 on Build GRCh38

View Full Product Details