Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 202128347-202128877 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR37L1
Entrez Gene ID
9283
Gene Name
G protein-coupled receptor 37 like 1
Gene Aliases
ET(B)R-LP-2, ETBR-LP-2
UniGene
Hs.132049
Gene Chromosome Location
Chr.1: 202118669-202133588 on Build GRCh38
Transcripts
NM_004767.3 XM_011510158.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2212855 c.1352C>T p.S451L Chr.1: 202128462-202128462 on Build GRCh38
6527999 c.1336T>C p.S446P Chr.1: 202128446-202128446 on Build GRCh38
3710507 c.1337C>G p.S446C Chr.1: 202128447-202128447 on Build GRCh38
5643916 c.1358A>G p.N453S Chr.1: 202128468-202128468 on Build GRCh38
3710506 c.1257C>G p.I419M Chr.1: 202128367-202128367 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202038749 Chr.1: 202128376-202128376 on Build GRCh38
rs749733623 Chr.1: 202128397-202128399 on Build GRCh38
rs377535954 Chr.1: 202128370-202128370 on Build GRCh38
rs760421842 Chr.1: 202128468-202128468 on Build GRCh38
rs138003036 Chr.1: 202128380-202128380 on Build GRCh38
rs116810937 Chr.1: 202128520-202128520 on Build GRCh38
rs139797932 Chr.1: 202128452-202128452 on Build GRCh38
rs748607219 Chr.1: 202128438-202128438 on Build GRCh38
rs923371109 Chr.1: 202128579-202128579 on Build GRCh38
rs773098917 Chr.1: 202128463-202128463 on Build GRCh38
rs552435625 Chr.1: 202128490-202128490 on Build GRCh38
rs561235038 Chr.1: 202128836-202128836 on Build GRCh38
rs781386235 Chr.1: 202128540-202128540 on Build GRCh38
rs751408912 Chr.1: 202128414-202128414 on Build GRCh38
rs148475636 Chr.1: 202128485-202128485 on Build GRCh38
rs1028522598 Chr.1: 202128624-202128624 on Build GRCh38
rs767258187 Chr.1: 202128424-202128424 on Build GRCh38
rs764750408 Chr.1: 202128482-202128482 on Build GRCh38
rs142438985 Chr.1: 202128558-202128558 on Build GRCh38
rs770780756 Chr.1: 202128829-202128829 on Build GRCh38
rs776163678 Chr.1: 202128573-202128573 on Build GRCh38
rs769776658 Chr.1: 202128548-202128548 on Build GRCh38
rs778049527 Chr.1: 202128448-202128448 on Build GRCh38
rs538995092 Chr.1: 202128454-202128454 on Build GRCh38
rs535953735 Chr.1: 202128598-202128598 on Build GRCh38
rs777237313 Chr.1: 202128716-202128719 on Build GRCh38
rs965708636 Chr.1: 202128780-202128780 on Build GRCh38
rs760511827 Chr.1: 202128569-202128569 on Build GRCh38
rs775791874 Chr.1: 202128590-202128590 on Build GRCh38
rs773187185 Chr.1: 202128567-202128567 on Build GRCh38
rs750113131 Chr.1: 202128427-202128427 on Build GRCh38
rs1025858379 Chr.1: 202128823-202128823 on Build GRCh38
rs61734417 Chr.1: 202128431-202128431 on Build GRCh38
rs779287214 Chr.1: 202128433-202128433 on Build GRCh38
rs761506972 Chr.1: 202128423-202128423 on Build GRCh38
rs772082137 Chr.1: 202128564-202128564 on Build GRCh38
rs375036161 Chr.1: 202128481-202128481 on Build GRCh38
rs747666563 Chr.1: 202128411-202128411 on Build GRCh38
rs767423544 Chr.1: 202128462-202128462 on Build GRCh38
rs1021338290 Chr.1: 202128720-202128720 on Build GRCh38
rs938851443 Chr.1: 202128645-202128645 on Build GRCh38
rs117132426 Chr.1: 202128599-202128599 on Build GRCh38
rs976608632 Chr.1: 202128820-202128820 on Build GRCh38
rs758707358 Chr.1: 202128439-202128439 on Build GRCh38
rs757602604 Chr.1: 202128608-202128608 on Build GRCh38
rs764688242 Chr.1: 202128594-202128594 on Build GRCh38
rs1018176577 Chr.1: 202128772-202128772 on Build GRCh38
rs772452942 Chr.1: 202128576-202128576 on Build GRCh38
rs535629807 Chr.1: 202128845-202128845 on Build GRCh38
rs200746512 Chr.1: 202128430-202128430 on Build GRCh38
rs573637497 Chr.1: 202128591-202128591 on Build GRCh38
rs572294919 Chr.1: 202128706-202128706 on Build GRCh38
rs753467741 Chr.1: 202128473-202128473 on Build GRCh38
rs766195168 Chr.1: 202128571-202128571 on Build GRCh38
rs762972110 Chr.1: 202128383-202128383 on Build GRCh38
rs534860277 Chr.1: 202128530-202128530 on Build GRCh38
rs367653940 Chr.1: 202128502-202128502 on Build GRCh38
rs771388636 Chr.1: 202128396-202128397 on Build GRCh38
rs763917378 Chr.1: 202128392-202128392 on Build GRCh38
rs555834757 Chr.1: 202128633-202128633 on Build GRCh38
rs778472284 Chr.1: 202128733-202128733 on Build GRCh38
rs558859323 Chr.1: 202128662-202128662 on Build GRCh38
rs748062878 Chr.1: 202128559-202128559 on Build GRCh38
rs1006534896 Chr.1: 202128771-202128771 on Build GRCh38
rs541180255 Chr.1: 202128732-202128732 on Build GRCh38
rs981222490 Chr.1: 202128815-202128815 on Build GRCh38
rs114760760 Chr.1: 202128850-202128850 on Build GRCh38
rs966740335 Chr.1: 202128724-202128724 on Build GRCh38
rs949776460 Chr.1: 202128809-202128810 on Build GRCh38
rs957313780 Chr.1: 202128761-202128761 on Build GRCh38
rs753151926 Chr.1: 202128495-202128495 on Build GRCh38
rs759831814 Chr.1: 202128540-202128540 on Build GRCh38
rs766104125 Chr.1: 202128471-202128471 on Build GRCh38
rs747264401 Chr.1: 202128449-202128449 on Build GRCh38
rs904173078 Chr.1: 202128755-202128755 on Build GRCh38
rs755756498 Chr.1: 202128428-202128428 on Build GRCh38
rs745906677 Chr.1: 202128456-202128456 on Build GRCh38
rs374456955 Chr.1: 202128783-202128783 on Build GRCh38
rs372455063 Chr.1: 202128600-202128600 on Build GRCh38
rs891545130 Chr.1: 202128687-202128687 on Build GRCh38
rs761751995 Chr.1: 202128461-202128461 on Build GRCh38
rs774140568 Chr.1: 202128457-202128457 on Build GRCh38
rs747354319 Chr.1: 202128527-202128527 on Build GRCh38
rs746015314 Chr.1: 202128544-202128544 on Build GRCh38
rs1044630673 Chr.1: 202128613-202128613 on Build GRCh38
rs996890220 Chr.1: 202128445-202128449 on Build GRCh38
rs773413931 Chr.1: 202128375-202128375 on Build GRCh38
rs1057492478 Chr.1: 202128669-202128669 on Build GRCh38
rs377248033 Chr.1: 202128603-202128603 on Build GRCh38
rs1008331607 Chr.1: 202128673-202128673 on Build GRCh38

View Full Product Details