Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 205659321-205659826 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC45A3
Entrez Gene ID
85414
Gene Name
solute carrier family 45 member 3
Gene Aliases
IPCA-2, IPCA-6, IPCA-8, IPCA6, PCANAP2, PCANAP6, PCANAP8, PRST
UniGene
Hs.278695
Gene Chromosome Location
Chr.1: 205657851-205680502 on Build GRCh38
Transcripts
NM_033102.2 XM_005245556.3 XM_005245557.4 XM_017002638.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4568706 c.1283T>A p.M428K Chr.1: 205659613-205659613 on Build GRCh38
1748108 c.1288A>T p.S430C Chr.1: 205659608-205659608 on Build GRCh38
4727441 c.1435G>A p.E479K Chr.1: 205659461-205659461 on Build GRCh38
3789390 c.1471G>T p.D491Y Chr.1: 205659425-205659425 on Build GRCh38
209635 c.1501C>T p.L501L Chr.1: 205659395-205659395 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142259819 Chr.1: 205659440-205659440 on Build GRCh38
rs769630273 Chr.1: 205659474-205659474 on Build GRCh38
rs976934436 Chr.1: 205659729-205659729 on Build GRCh38
rs145219219 Chr.1: 205659563-205659563 on Build GRCh38
rs369811484 Chr.1: 205659653-205659653 on Build GRCh38
rs750499199 Chr.1: 205659437-205659437 on Build GRCh38
rs759712471 Chr.1: 205659526-205659527 on Build GRCh38
rs769418885 Chr.1: 205659561-205659561 on Build GRCh38
rs145712905 Chr.1: 205659652-205659652 on Build GRCh38
rs758969117 Chr.1: 205659460-205659460 on Build GRCh38
rs961513990 Chr.1: 205659704-205659704 on Build GRCh38
rs776091785 Chr.1: 205659411-205659411 on Build GRCh38
rs745477887 Chr.1: 205659469-205659469 on Build GRCh38
rs775238251 Chr.1: 205659485-205659485 on Build GRCh38
rs749083178 Chr.1: 205659568-205659568 on Build GRCh38
rs755552178 Chr.1: 205659687-205659687 on Build GRCh38
rs751972382 Chr.1: 205659537-205659537 on Build GRCh38
rs915164912 Chr.1: 205659769-205659769 on Build GRCh38
rs770605686 Chr.1: 205659407-205659407 on Build GRCh38
rs922375806 Chr.1: 205659750-205659750 on Build GRCh38
rs761631942 Chr.1: 205659613-205659613 on Build GRCh38
rs932346135 Chr.1: 205659752-205659752 on Build GRCh38
rs766319015 Chr.1: 205659516-205659516 on Build GRCh38
rs892288859 Chr.1: 205659523-205659523 on Build GRCh38
rs115342538 Chr.1: 205659767-205659767 on Build GRCh38
rs1035760449 Chr.1: 205659575-205659575 on Build GRCh38
rs778349230 Chr.1: 205659532-205659532 on Build GRCh38
rs113738580 Chr.1: 205659591-205659591 on Build GRCh38
rs1023644413 Chr.1: 205659592-205659592 on Build GRCh38
rs748705004 Chr.1: 205659449-205659449 on Build GRCh38
rs752984647 Chr.1: 205659695-205659695 on Build GRCh38
rs760168590 Chr.1: 205659461-205659461 on Build GRCh38
rs772747448 Chr.1: 205659348-205659348 on Build GRCh38
rs776379420 Chr.1: 205659465-205659465 on Build GRCh38
rs538901068 Chr.1: 205659647-205659647 on Build GRCh38
rs137949511 Chr.1: 205659488-205659488 on Build GRCh38
rs376079634 Chr.1: 205659419-205659419 on Build GRCh38
rs35201492 Chr.1: 205659513-205659513 on Build GRCh38
rs749944845 Chr.1: 205659683-205659683 on Build GRCh38
rs368927767 Chr.1: 205659444-205659444 on Build GRCh38
rs765764027 Chr.1: 205659521-205659521 on Build GRCh38
rs779709885 Chr.1: 205659691-205659691 on Build GRCh38
rs200659267 Chr.1: 205659445-205659445 on Build GRCh38
rs774366329 Chr.1: 205659600-205659600 on Build GRCh38
rs200496056 Chr.1: 205659423-205659423 on Build GRCh38
rs768293551 Chr.1: 205659590-205659590 on Build GRCh38
rs780750978 Chr.1: 205659678-205659678 on Build GRCh38
rs377393987 Chr.1: 205659758-205659758 on Build GRCh38
rs757677233 Chr.1: 205659666-205659666 on Build GRCh38
rs919455083 Chr.1: 205659639-205659639 on Build GRCh38
rs780661012 Chr.1: 205659548-205659548 on Build GRCh38
rs756086521 Chr.1: 205659439-205659439 on Build GRCh38
rs764318789 Chr.1: 205659649-205659649 on Build GRCh38
rs754160862 Chr.1: 205659519-205659519 on Build GRCh38
rs758633571 Chr.1: 205659522-205659522 on Build GRCh38
rs147838006 Chr.1: 205659551-205659551 on Build GRCh38
rs771756992 Chr.1: 205659343-205659343 on Build GRCh38
rs745563098 Chr.1: 205659550-205659550 on Build GRCh38
rs765881137 Chr.1: 205659484-205659484 on Build GRCh38
rs774087128 Chr.1: 205659512-205659512 on Build GRCh38
rs752901803 Chr.1: 205659642-205659642 on Build GRCh38
rs960355244 Chr.1: 205659579-205659579 on Build GRCh38
rs765570956 Chr.1: 205659633-205659633 on Build GRCh38
rs980824896 Chr.1: 205659367-205659367 on Build GRCh38
rs140263007 Chr.1: 205659520-205659520 on Build GRCh38
rs148291270 Chr.1: 205659422-205659422 on Build GRCh38
rs770282409 Chr.1: 205659463-205659463 on Build GRCh38
rs149147695 Chr.1: 205659420-205659420 on Build GRCh38
rs547421942 Chr.1: 205659462-205659462 on Build GRCh38
rs534006274 Chr.1: 205659441-205659441 on Build GRCh38
rs201668481 Chr.1: 205659489-205659489 on Build GRCh38
rs375360087 Chr.1: 205659621-205659621 on Build GRCh38
rs774281642 Chr.1: 205659525-205659525 on Build GRCh38
rs572206763 Chr.1: 205659784-205659784 on Build GRCh38
rs746707250 Chr.1: 205659397-205659397 on Build GRCh38
rs756816234 Chr.1: 205659546-205659546 on Build GRCh38
rs1042259498 Chr.1: 205659800-205659800 on Build GRCh38
rs762202564 Chr.1: 205659387-205659387 on Build GRCh38
rs867488615 Chr.1: 205659573-205659573 on Build GRCh38
rs767557057 Chr.1: 205659436-205659436 on Build GRCh38
rs767604466 Chr.1: 205659609-205659609 on Build GRCh38

View Full Product Details