Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206770828-206771330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL10
Entrez Gene ID
3586
Gene Name
interleukin 10
Gene Aliases
CSIF, GVHDS, IL-10, IL10A, TGIF
UniGene
Hs.193717
Gene Chromosome Location
Chr.1: 206767603-206772494 on Build GRCh38
Transcripts
NM_000572.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3482495 c.258G>A p.M86I Chr.1: 206771027-206771027 on Build GRCh38
903005 c.239G>A p.C80Y Chr.1: 206771046-206771046 on Build GRCh38
3789406 c.297G>A p.E99E Chr.1: 206770988-206770988 on Build GRCh38
6944881 c.277G>A p.E93K Chr.1: 206771008-206771008 on Build GRCh38
6948695 c.304G>A p.D102N Chr.1: 206770981-206770981 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs998971154 Chr.1: 206771222-206771222 on Build GRCh38
rs370871933 Chr.1: 206770871-206770871 on Build GRCh38
rs904512012 Chr.1: 206771130-206771130 on Build GRCh38
rs952642115 Chr.1: 206770916-206770916 on Build GRCh38
rs371608920 Chr.1: 206770920-206770920 on Build GRCh38
rs776655495 Chr.1: 206770925-206770925 on Build GRCh38
rs1014838285 Chr.1: 206771182-206771182 on Build GRCh38
rs1043665980 Chr.1: 206771181-206771181 on Build GRCh38
rs757522932 Chr.1: 206770870-206770870 on Build GRCh38
rs1554286 Chr.1: 206770888-206770888 on Build GRCh38
rs149143243 Chr.1: 206770969-206770969 on Build GRCh38
rs780612967 Chr.1: 206771011-206771011 on Build GRCh38
rs1016634050 Chr.1: 206771231-206771231 on Build GRCh38
rs753924360 Chr.1: 206771073-206771073 on Build GRCh38
rs991441780 Chr.1: 206770913-206770913 on Build GRCh38
rs759430658 Chr.1: 206770926-206770926 on Build GRCh38
rs549828182 Chr.1: 206771066-206771066 on Build GRCh38
rs756542399 Chr.1: 206770883-206770883 on Build GRCh38
rs755403362 Chr.1: 206770910-206770910 on Build GRCh38
rs767727378 Chr.1: 206770949-206770949 on Build GRCh38
rs775288303 Chr.1: 206771033-206771033 on Build GRCh38
rs777582308 Chr.1: 206770984-206770984 on Build GRCh38
rs757756856 Chr.1: 206771192-206771192 on Build GRCh38
rs1518111 Chr.1: 206771300-206771300 on Build GRCh38
rs2352792 Chr.1: 206770867-206770867 on Build GRCh38
rs762018888 Chr.1: 206770944-206770944 on Build GRCh38
rs750441744 Chr.1: 206770958-206770958 on Build GRCh38
rs769682299 Chr.1: 206771030-206771030 on Build GRCh38
rs571608526 Chr.1: 206771180-206771180 on Build GRCh38
rs758435689 Chr.1: 206771308-206771308 on Build GRCh38
rs374619208 Chr.1: 206770911-206770911 on Build GRCh38
rs770531048 Chr.1: 206771008-206771008 on Build GRCh38
rs750564052 Chr.1: 206770874-206770874 on Build GRCh38
rs758058722 Chr.1: 206770983-206770983 on Build GRCh38
rs369055073 Chr.1: 206771048-206771048 on Build GRCh38
rs932332598 Chr.1: 206771071-206771071 on Build GRCh38
rs538037506 Chr.1: 206771117-206771117 on Build GRCh38
rs998902524 Chr.1: 206770852-206770852 on Build GRCh38
rs756460032 Chr.1: 206770996-206770996 on Build GRCh38
rs371748044 Chr.1: 206770891-206770891 on Build GRCh38
rs569399482 Chr.1: 206771095-206771095 on Build GRCh38
rs867226419 Chr.1: 206770856-206770856 on Build GRCh38
rs1043052843 Chr.1: 206771104-206771104 on Build GRCh38
rs913710321 Chr.1: 206771303-206771303 on Build GRCh38
rs937619899 Chr.1: 206771142-206771142 on Build GRCh38
rs772231902 Chr.1: 206771041-206771041 on Build GRCh38
rs547437506 Chr.1: 206770961-206770961 on Build GRCh38
rs755490123 Chr.1: 206770965-206770965 on Build GRCh38
rs777530778 Chr.1: 206771309-206771309 on Build GRCh38
rs150423829 Chr.1: 206770942-206770942 on Build GRCh38
rs914866462 Chr.1: 206771155-206771155 on Build GRCh38
rs534191384 Chr.1: 206771259-206771259 on Build GRCh38
rs560908141 Chr.1: 206770964-206770964 on Build GRCh38
rs148703263 Chr.1: 206771272-206771272 on Build GRCh38
rs200235675 Chr.1: 206770894-206770894 on Build GRCh38
rs746471077 Chr.1: 206771000-206771000 on Build GRCh38
rs753066499 Chr.1: 206770884-206770885 on Build GRCh38
rs760083929 Chr.1: 206771091-206771091 on Build GRCh38
rs886869543 Chr.1: 206771208-206771208 on Build GRCh38
rs946103550 Chr.1: 206771087-206771087 on Build GRCh38
rs1035697567 Chr.1: 206770860-206770860 on Build GRCh38
rs139352858 Chr.1: 206771057-206771057 on Build GRCh38
rs1014037243 Chr.1: 206770859-206770859 on Build GRCh38
rs745801059 Chr.1: 206771014-206771014 on Build GRCh38
rs771912629 Chr.1: 206770912-206770912 on Build GRCh38
rs377175850 Chr.1: 206771277-206771277 on Build GRCh38
rs753179739 Chr.1: 206770978-206770978 on Build GRCh38
rs752079099 Chr.1: 206771154-206771154 on Build GRCh38
rs760929586 Chr.1: 206771064-206771064 on Build GRCh38
rs762750138 Chr.1: 206771036-206771036 on Build GRCh38
rs376002450 Chr.1: 206770975-206770975 on Build GRCh38
rs746523858 Chr.1: 206770915-206770915 on Build GRCh38
rs767321028 Chr.1: 206770881-206770882 on Build GRCh38
rs5743626 Chr.1: 206770940-206770940 on Build GRCh38
rs987614203 Chr.1: 206770976-206770976 on Build GRCh38
rs765131042 Chr.1: 206771096-206771096 on Build GRCh38
rs551516761 Chr.1: 206771160-206771160 on Build GRCh38
rs370491306 Chr.1: 206771282-206771282 on Build GRCh38
rs372883095 Chr.1: 206770971-206770971 on Build GRCh38
rs201365412 Chr.1: 206770914-206770914 on Build GRCh38
rs1004062279 Chr.1: 206771101-206771101 on Build GRCh38

View Full Product Details