Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206866331-206866828 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL20
Entrez Gene ID
50604
Gene Name
interleukin 20
Gene Aliases
IL-20, IL10D, ZCYTO10
UniGene
Hs.272373
Gene Chromosome Location
Chr.1: 206865247-206869223 on Build GRCh38
Transcripts
NM_018724.3 XM_005273152.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3720124 c.204T>A p.T68T Chr.1: 206866343-206866343 on Build GRCh38
4027557 c.325A>T p.I109F Chr.1: 206866583-206866583 on Build GRCh38
364937 c.327C>G p.I109M Chr.1: 206866585-206866585 on Build GRCh38
7135840 c.231G>T p.A77A Chr.1: 206866489-206866489 on Build GRCh38
7135841 c.235C>A p.R79R Chr.1: 206866493-206866493 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs190712250 Chr.1: 206866696-206866696 on Build GRCh38
rs757554164 Chr.1: 206866657-206866657 on Build GRCh38
rs1026311013 Chr.1: 206866599-206866599 on Build GRCh38
rs373331152 Chr.1: 206866577-206866577 on Build GRCh38
rs368827189 Chr.1: 206866447-206866447 on Build GRCh38
rs913549785 Chr.1: 206866743-206866743 on Build GRCh38
rs572480727 Chr.1: 206866801-206866801 on Build GRCh38
rs552681587 Chr.1: 206866789-206866789 on Build GRCh38
rs185979169 Chr.1: 206866474-206866474 on Build GRCh38
rs373822104 Chr.1: 206866663-206866663 on Build GRCh38
rs202145940 Chr.1: 206866612-206866612 on Build GRCh38
rs765725485 Chr.1: 206866401-206866401 on Build GRCh38
rs548155536 Chr.1: 206866582-206866582 on Build GRCh38
rs1046117796 Chr.1: 206866780-206866780 on Build GRCh38
rs374740863 Chr.1: 206866443-206866443 on Build GRCh38
rs111506841 Chr.1: 206866478-206866478 on Build GRCh38
rs934912481 Chr.1: 206866716-206866716 on Build GRCh38
rs757168986 Chr.1: 206866444-206866444 on Build GRCh38
rs558858635 Chr.1: 206866509-206866509 on Build GRCh38
rs1002003744 Chr.1: 206866788-206866788 on Build GRCh38
rs369436616 Chr.1: 206866629-206866629 on Build GRCh38
rs758525930 Chr.1: 206866558-206866558 on Build GRCh38
rs750304865 Chr.1: 206866524-206866524 on Build GRCh38
rs904946980 Chr.1: 206866786-206866786 on Build GRCh38
rs150575323 Chr.1: 206866628-206866628 on Build GRCh38
rs753724528 Chr.1: 206866557-206866557 on Build GRCh38
rs754290740 Chr.1: 206866560-206866561 on Build GRCh38
rs949072237 Chr.1: 206866771-206866771 on Build GRCh38
rs369016895 Chr.1: 206866488-206866488 on Build GRCh38
rs754762138 Chr.1: 206866630-206866630 on Build GRCh38
rs763441745 Chr.1: 206866414-206866414 on Build GRCh38
rs760017039 Chr.1: 206866394-206866394 on Build GRCh38
rs751771925 Chr.1: 206866652-206866652 on Build GRCh38
rs370272158 Chr.1: 206866406-206866406 on Build GRCh38
rs141716593 Chr.1: 206866493-206866493 on Build GRCh38
rs372073353 Chr.1: 206866459-206866459 on Build GRCh38
rs764962041 Chr.1: 206866633-206866633 on Build GRCh38
rs964880344 Chr.1: 206866689-206866689 on Build GRCh38
rs374849164 Chr.1: 206866482-206866482 on Build GRCh38
rs138566326 Chr.1: 206866365-206866365 on Build GRCh38
rs772404778 Chr.1: 206866460-206866460 on Build GRCh38
rs941288522 Chr.1: 206866694-206866694 on Build GRCh38
rs993452015 Chr.1: 206866581-206866581 on Build GRCh38
rs751143076 Chr.1: 206866441-206866441 on Build GRCh38
rs773346133 Chr.1: 206866364-206866364 on Build GRCh38
rs766084077 Chr.1: 206866530-206866530 on Build GRCh38
rs752240076 Chr.1: 206866402-206866402 on Build GRCh38
rs904999556 Chr.1: 206866525-206866525 on Build GRCh38
rs768935657 Chr.1: 206866595-206866595 on Build GRCh38
rs375942033 Chr.1: 206866545-206866545 on Build GRCh38
rs377545697 Chr.1: 206866664-206866664 on Build GRCh38
rs762677214 Chr.1: 206866411-206866411 on Build GRCh38
rs781151357 Chr.1: 206866445-206866445 on Build GRCh38
rs746304533 Chr.1: 206866527-206866527 on Build GRCh38
rs780648406 Chr.1: 206866424-206866424 on Build GRCh38
rs539132265 Chr.1: 206866782-206866782 on Build GRCh38
rs769010425 Chr.1: 206866688-206866688 on Build GRCh38
rs781413639 Chr.1: 206866661-206866661 on Build GRCh38
rs926188441 Chr.1: 206866705-206866705 on Build GRCh38
rs377665029 Chr.1: 206866602-206866602 on Build GRCh38
rs201490344 Chr.1: 206866568-206866568 on Build GRCh38
rs908440085 Chr.1: 206866632-206866632 on Build GRCh38
rs371312456 Chr.1: 206866508-206866508 on Build GRCh38
rs771532281 Chr.1: 206866472-206866472 on Build GRCh38
rs777701594 Chr.1: 206866462-206866462 on Build GRCh38
rs1038034466 Chr.1: 206866714-206866714 on Build GRCh38
rs773706986 Chr.1: 206866494-206866494 on Build GRCh38
rs374625441 Chr.1: 206866731-206866731 on Build GRCh38
rs777112163 Chr.1: 206866384-206866384 on Build GRCh38
rs780574455 Chr.1: 206866683-206866683 on Build GRCh38
rs781431973 Chr.1: 206866573-206866573 on Build GRCh38
rs899568619 Chr.1: 206866470-206866470 on Build GRCh38
rs779913461 Chr.1: 206866457-206866457 on Build GRCh38
rs756551969 Chr.1: 206866675-206866675 on Build GRCh38
rs747133267 Chr.1: 206866471-206866471 on Build GRCh38
rs570355390 Chr.1: 206866717-206866717 on Build GRCh38
rs35856950 Chr.1: 206866578-206866578 on Build GRCh38
rs771989159 Chr.1: 206866362-206866362 on Build GRCh38
rs112285489 Chr.1: 206866489-206866489 on Build GRCh38
rs778095574 Chr.1: 206866560-206866560 on Build GRCh38
rs749784858 Chr.1: 206866687-206866687 on Build GRCh38
rs749705601 Chr.1: 206866587-206866587 on Build GRCh38
rs755857210 Chr.1: 206866448-206866448 on Build GRCh38
rs536804184 Chr.1: 206866624-206866624 on Build GRCh38
rs933389869 Chr.1: 206866392-206866392 on Build GRCh38
rs766948661 Chr.1: 206866615-206866615 on Build GRCh38

View Full Product Details