Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206962126-206962626 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCAMR
Entrez Gene ID
83953
Gene Name
Fc fragment of IgA and IgM receptor
Gene Aliases
CD351, FCA/MR, FKSG87
UniGene
Hs.145519
Gene Chromosome Location
Chr.1: 206957955-206970657 on Build GRCh38
Transcripts
NM_001122979.2 NM_001170631.1 NM_032029.4 XM_011510051.2 XM_011510052.2 XM_017002442.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6123980 c.376C>T p.Q126* Chr.1: 206962489-206962489 on Build GRCh38
3934434 c.333C>A p.G111G Chr.1: 206962532-206962532 on Build GRCh38
5641726 c.431G>T p.R144L Chr.1: 206962434-206962434 on Build GRCh38
1338343 c.523G>A p.G175S Chr.1: 206962270-206962270 on Build GRCh38
3864298 c.580G>A p.G194S Chr.1: 206962213-206962213 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771872646 Chr.1: 206962361-206962361 on Build GRCh38
rs368893206 Chr.1: 206962428-206962428 on Build GRCh38
rs371809424 Chr.1: 206962412-206962412 on Build GRCh38
rs774787300 Chr.1: 206962249-206962249 on Build GRCh38
rs896927538 Chr.1: 206962472-206962472 on Build GRCh38
rs761038004 Chr.1: 206962365-206962365 on Build GRCh38
rs535083117 Chr.1: 206962470-206962470 on Build GRCh38
rs769484305 Chr.1: 206962233-206962234 on Build GRCh38
rs924090996 Chr.1: 206962380-206962380 on Build GRCh38
rs939132718 Chr.1: 206962346-206962346 on Build GRCh38
rs927722413 Chr.1: 206962333-206962333 on Build GRCh38
rs947694511 Chr.1: 206962284-206962284 on Build GRCh38
rs763140040 Chr.1: 206962229-206962229 on Build GRCh38
rs202042926 Chr.1: 206962280-206962280 on Build GRCh38
rs1043897339 Chr.1: 206962161-206962161 on Build GRCh38
rs200521290 Chr.1: 206962467-206962467 on Build GRCh38
rs753533479 Chr.1: 206962367-206962367 on Build GRCh38
rs267598335 Chr.1: 206962415-206962415 on Build GRCh38
rs1048619697 Chr.1: 206962585-206962585 on Build GRCh38
rs755087173 Chr.1: 206962475-206962475 on Build GRCh38
rs979417718 Chr.1: 206962530-206962530 on Build GRCh38
rs777795004 Chr.1: 206962557-206962557 on Build GRCh38
rs751387228 Chr.1: 206962386-206962386 on Build GRCh38
rs199638609 Chr.1: 206962196-206962196 on Build GRCh38
rs375632066 Chr.1: 206962542-206962542 on Build GRCh38
rs750272728 Chr.1: 206962473-206962473 on Build GRCh38
rs750819666 Chr.1: 206962271-206962271 on Build GRCh38
rs139272162 Chr.1: 206962328-206962328 on Build GRCh38
rs749374377 Chr.1: 206962351-206962351 on Build GRCh38
rs368195235 Chr.1: 206962354-206962354 on Build GRCh38
rs747469340 Chr.1: 206962506-206962506 on Build GRCh38
rs779549688 Chr.1: 206962599-206962599 on Build GRCh38
rs768251382 Chr.1: 206962466-206962466 on Build GRCh38
rs763933914 Chr.1: 206962458-206962458 on Build GRCh38
rs546302574 Chr.1: 206962435-206962435 on Build GRCh38
rs868159707 Chr.1: 206962218-206962218 on Build GRCh38
rs767564605 Chr.1: 206962538-206962538 on Build GRCh38
rs752628649 Chr.1: 206962319-206962319 on Build GRCh38
rs765999152 Chr.1: 206962366-206962366 on Build GRCh38
rs762339034 Chr.1: 206962533-206962533 on Build GRCh38
rs753000721 Chr.1: 206962549-206962549 on Build GRCh38
rs764950067 Chr.1: 206962444-206962444 on Build GRCh38
rs778235453 Chr.1: 206962501-206962501 on Build GRCh38
rs200786919 Chr.1: 206962360-206962360 on Build GRCh38
rs972627590 Chr.1: 206962304-206962304 on Build GRCh38
rs548207600 Chr.1: 206962515-206962515 on Build GRCh38
rs377248549 Chr.1: 206962451-206962451 on Build GRCh38
rs746216712 Chr.1: 206962575-206962575 on Build GRCh38
rs367804054 Chr.1: 206962285-206962285 on Build GRCh38
rs774936386 Chr.1: 206962321-206962322 on Build GRCh38
rs370685674 Chr.1: 206962509-206962509 on Build GRCh38
rs781457039 Chr.1: 206962205-206962205 on Build GRCh38
rs748160527 Chr.1: 206962484-206962484 on Build GRCh38
rs372119748 Chr.1: 206962295-206962295 on Build GRCh38
rs147389157 Chr.1: 206962483-206962483 on Build GRCh38
rs370955692 Chr.1: 206962151-206962151 on Build GRCh38
rs561968221 Chr.1: 206962371-206962371 on Build GRCh38
rs780391610 Chr.1: 206962401-206962401 on Build GRCh38
rs370990821 Chr.1: 206962186-206962186 on Build GRCh38
rs559768863 Chr.1: 206962270-206962270 on Build GRCh38
rs370635048 Chr.1: 206962251-206962251 on Build GRCh38
rs183689490 Chr.1: 206962153-206962153 on Build GRCh38
rs74148895 Chr.1: 206962465-206962465 on Build GRCh38
rs764606736 Chr.1: 206962370-206962370 on Build GRCh38
rs377626977 Chr.1: 206962173-206962173 on Build GRCh38
rs762583154 Chr.1: 206962454-206962454 on Build GRCh38
rs202227399 Chr.1: 206962286-206962286 on Build GRCh38
rs769273194 Chr.1: 206962236-206962236 on Build GRCh38
rs754536090 Chr.1: 206962296-206962296 on Build GRCh38
rs199611731 Chr.1: 206962325-206962325 on Build GRCh38
rs768896930 Chr.1: 206962353-206962353 on Build GRCh38
rs747975636 Chr.1: 206962200-206962200 on Build GRCh38
rs185655202 Chr.1: 206962541-206962541 on Build GRCh38
rs1572808 Chr.1: 206962201-206962201 on Build GRCh38
rs751659664 Chr.1: 206962535-206962535 on Build GRCh38
rs758204600 Chr.1: 206962372-206962372 on Build GRCh38
rs770173232 Chr.1: 206962213-206962213 on Build GRCh38
rs763708941 Chr.1: 206962534-206962534 on Build GRCh38
rs758574667 Chr.1: 206962554-206962554 on Build GRCh38
rs770234106 Chr.1: 206962597-206962597 on Build GRCh38
rs748261902 Chr.1: 206962425-206962425 on Build GRCh38
rs769319619 Chr.1: 206962523-206962523 on Build GRCh38
rs763406539 Chr.1: 206962246-206962246 on Build GRCh38
rs779501115 Chr.1: 206962162-206962162 on Build GRCh38
rs182257994 Chr.1: 206962427-206962427 on Build GRCh38
rs752885588 Chr.1: 206962601-206962601 on Build GRCh38
rs766748652 Chr.1: 206962294-206962298 on Build GRCh38
rs773534484 Chr.1: 206962235-206962235 on Build GRCh38
rs778882688 Chr.1: 206962317-206962317 on Build GRCh38
rs201548850 Chr.1: 206962407-206962407 on Build GRCh38
rs751645943 Chr.1: 206962561-206962561 on Build GRCh38
rs747783775 Chr.1: 206962191-206962191 on Build GRCh38
rs55633368 Chr.1: 206962391-206962391 on Build GRCh38
rs1044727988 Chr.1: 206962297-206962297 on Build GRCh38
rs776188186 Chr.1: 206962176-206962177 on Build GRCh38
rs767703423 Chr.1: 206962509-206962510 on Build GRCh38
rs746030957 Chr.1: 206962209-206962209 on Build GRCh38
rs377157283 Chr.1: 206962217-206962217 on Build GRCh38
rs758567403 Chr.1: 206962487-206962487 on Build GRCh38
rs758385523 Chr.1: 206962321-206962321 on Build GRCh38
rs568650021 Chr.1: 206962511-206962511 on Build GRCh38
rs577402005 Chr.1: 206962570-206962570 on Build GRCh38
rs188207561 Chr.1: 206962378-206962378 on Build GRCh38
rs750182760 Chr.1: 206962539-206962539 on Build GRCh38
rs759028700 Chr.1: 206962368-206962368 on Build GRCh38
rs759469591 Chr.1: 206962325-206962325 on Build GRCh38
rs377470491 Chr.1: 206962420-206962420 on Build GRCh38
rs374305028 Chr.1: 206962434-206962434 on Build GRCh38
rs780481679 Chr.1: 206962330-206962330 on Build GRCh38

View Full Product Details