Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 212064796-212065340 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTL
Entrez Gene ID
51514
Gene Name
denticleless E3 ubiquitin protein ligase homolog
Gene Aliases
CDT2, DCAF2, L2DTL, RAMP
UniGene
Hs.656473
Gene Chromosome Location
Chr.1: 212035553-212105006 on Build GRCh38
Transcripts
NM_001286229.1 NM_001286230.1 NM_016448.3 XM_011509614.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
349955 c.612G>A p.Q204Q Chr.1: 212065002-212065002 on Build GRCh38
4934711 c.527-2A>G p.? Chr.1: 212064915-212064915 on Build GRCh38
5845384 c.610C>T p.Q204* Chr.1: 212065000-212065000 on Build GRCh38
7089718 c.590C>A p.P197H Chr.1: 212064980-212064980 on Build GRCh38
4027917 c.561A>G p.G187G Chr.1: 212064951-212064951 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766139696 Chr.1: 212064873-212064873 on Build GRCh38
rs747196342 Chr.1: 212065058-212065059 on Build GRCh38
rs368018761 Chr.1: 212064877-212064877 on Build GRCh38
rs572886766 Chr.1: 212065225-212065227 on Build GRCh38
rs1004137609 Chr.1: 212065221-212065221 on Build GRCh38
rs888370150 Chr.1: 212065218-212065218 on Build GRCh38
rs1037647030 Chr.1: 212065259-212065259 on Build GRCh38
rs1044477142 Chr.1: 212065053-212065053 on Build GRCh38
rs948433287 Chr.1: 212064857-212064857 on Build GRCh38
rs1015402908 Chr.1: 212065247-212065247 on Build GRCh38
rs1019351651 Chr.1: 212064859-212064859 on Build GRCh38
rs767625292 Chr.1: 212064867-212064867 on Build GRCh38
rs765070650 Chr.1: 212064882-212064882 on Build GRCh38
rs139190048 Chr.1: 212064871-212064871 on Build GRCh38
rs367916197 Chr.1: 212065315-212065315 on Build GRCh38
rs1028106455 Chr.1: 212065201-212065203 on Build GRCh38
rs777451533 Chr.1: 212064921-212064921 on Build GRCh38
rs773383298 Chr.1: 212064870-212064870 on Build GRCh38
rs1022695985 Chr.1: 212065267-212065267 on Build GRCh38
rs769005657 Chr.1: 212065061-212065079 on Build GRCh38
rs374382665 Chr.1: 212065277-212065277 on Build GRCh38
rs776967527 Chr.1: 212065079-212065079 on Build GRCh38
rs780303898 Chr.1: 212065054-212065055 on Build GRCh38
rs749885698 Chr.1: 212064834-212064834 on Build GRCh38
rs756978216 Chr.1: 212064937-212064937 on Build GRCh38
rs765997063 Chr.1: 212064980-212064980 on Build GRCh38
rs748685719 Chr.1: 212065052-212065052 on Build GRCh38
rs536860245 Chr.1: 212065305-212065305 on Build GRCh38
rs1039111629 Chr.1: 212065141-212065141 on Build GRCh38
rs140634631 Chr.1: 212065004-212065004 on Build GRCh38
rs900606511 Chr.1: 212065159-212065159 on Build GRCh38
rs757946526 Chr.1: 212064908-212064908 on Build GRCh38
rs769115066 Chr.1: 212065043-212065043 on Build GRCh38
rs948702569 Chr.1: 212065038-212065038 on Build GRCh38
rs555705046 Chr.1: 212065304-212065304 on Build GRCh38
rs978862214 Chr.1: 212064864-212064864 on Build GRCh38
rs373660724 Chr.1: 212065071-212065071 on Build GRCh38
rs11119838 Chr.1: 212064891-212064891 on Build GRCh38
rs970244310 Chr.1: 212065302-212065302 on Build GRCh38
rs773454183 Chr.1: 212065152-212065154 on Build GRCh38
rs994836846 Chr.1: 212065167-212065167 on Build GRCh38
rs569400270 Chr.1: 212065287-212065287 on Build GRCh38
rs897315540 Chr.1: 212065275-212065275 on Build GRCh38
rs780968796 Chr.1: 212064959-212064959 on Build GRCh38
rs367567616 Chr.1: 212065072-212065072 on Build GRCh38
rs571595893 Chr.1: 212065123-212065123 on Build GRCh38
rs371926545 Chr.1: 212064911-212064911 on Build GRCh38
rs746406987 Chr.1: 212064991-212064993 on Build GRCh38
rs958234122 Chr.1: 212064977-212064977 on Build GRCh38
rs772090987 Chr.1: 212065060-212065060 on Build GRCh38
rs181904808 Chr.1: 212065257-212065257 on Build GRCh38
rs189644922 Chr.1: 212065019-212065019 on Build GRCh38
rs773292910 Chr.1: 212065068-212065068 on Build GRCh38
rs753579263 Chr.1: 212064875-212064875 on Build GRCh38
rs923882764 Chr.1: 212064976-212064976 on Build GRCh38
rs532796988 Chr.1: 212065224-212065224 on Build GRCh38
rs746606042 Chr.1: 212064934-212064934 on Build GRCh38
rs201151313 Chr.1: 212064904-212064904 on Build GRCh38
rs938867650 Chr.1: 212065258-212065258 on Build GRCh38
rs758531289 Chr.1: 212065053-212065056 on Build GRCh38

View Full Product Details