Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 213129400-213129931 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS6KC1
Entrez Gene ID
26750
Gene Name
ribosomal protein S6 kinase C1
Gene Aliases
RPK118, RSKL1, S6K-delta-1, S6PKh1, humS6PKh1
UniGene
Hs.591416
Gene Chromosome Location
Chr.1: 213051233-213378753 on Build GRCh38
Transcripts
NM_001136138.2 NM_001287218.1 NM_001287219.1 NM_001287220.1 NM_001287221.1 NM_012424.4 XM_005273095.1 XM_005273096.1 XM_006711272.1 XM_017001024.1 XM_017001025.1 XM_017001026.1 XM_017001027.1 XM_017001028.1 XM_017001029.1 XM_017001030.1 XM_017001031.1 XM_017001032.1 XM_017001033.1 XM_017001034.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6124523 c.655G>T p.E219* Chr.1: 213129709-213129709 on Build GRCh38
1744297 c.669T>A p.R223R Chr.1: 213129723-213129723 on Build GRCh38
6769611 c.771C>T p.D257D Chr.1: 213129825-213129825 on Build GRCh38
4027962 c.596C>T p.S199L Chr.1: 213129650-213129650 on Build GRCh38
4722764 c.659G>A p.R220Q Chr.1: 213129713-213129713 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746933743 Chr.1: 213129596-213129596 on Build GRCh38
rs762294619 Chr.1: 213129485-213129485 on Build GRCh38
rs550052799 Chr.1: 213129425-213129425 on Build GRCh38
rs538313329 Chr.1: 213129424-213129424 on Build GRCh38
rs1010225886 Chr.1: 213129811-213129811 on Build GRCh38
rs751357136 Chr.1: 213129680-213129680 on Build GRCh38
rs762735521 Chr.1: 213129636-213129636 on Build GRCh38
rs756984597 Chr.1: 213129740-213129740 on Build GRCh38
rs781341505 Chr.1: 213129905-213129905 on Build GRCh38
rs534090468 Chr.1: 213129858-213129858 on Build GRCh38
rs758673123 Chr.1: 213129673-213129673 on Build GRCh38
rs756134497 Chr.1: 213129752-213129752 on Build GRCh38
rs191838083 Chr.1: 213129671-213129671 on Build GRCh38
rs553554592 Chr.1: 213129571-213129571 on Build GRCh38
rs769160109 Chr.1: 213129702-213129702 on Build GRCh38
rs779655205 Chr.1: 213129761-213129761 on Build GRCh38
rs769346061 Chr.1: 213129764-213129764 on Build GRCh38
rs754504568 Chr.1: 213129880-213129880 on Build GRCh38
rs878931355 Chr.1: 213129674-213129674 on Build GRCh38
rs966314060 Chr.1: 213129576-213129576 on Build GRCh38
rs747741490 Chr.1: 213129885-213129885 on Build GRCh38
rs755186439 Chr.1: 213129541-213129541 on Build GRCh38
rs771195924 Chr.1: 213129780-213129780 on Build GRCh38
rs768372423 Chr.1: 213129765-213129765 on Build GRCh38
rs372834454 Chr.1: 213129637-213129637 on Build GRCh38
rs759512467 Chr.1: 213129660-213129660 on Build GRCh38
rs748813450 Chr.1: 213129762-213129762 on Build GRCh38
rs779023286 Chr.1: 213129549-213129549 on Build GRCh38
rs778400175 Chr.1: 213129489-213129493 on Build GRCh38
rs377208928 Chr.1: 213129644-213129644 on Build GRCh38
rs144929190 Chr.1: 213129826-213129826 on Build GRCh38
rs776142805 Chr.1: 213129624-213129624 on Build GRCh38
rs755598192 Chr.1: 213129721-213129721 on Build GRCh38
rs1029632670 Chr.1: 213129873-213129873 on Build GRCh38
rs571982253 Chr.1: 213129606-213129606 on Build GRCh38
rs753873638 Chr.1: 213129862-213129862 on Build GRCh38
rs768047259 Chr.1: 213129488-213129488 on Build GRCh38
rs753879162 Chr.1: 213129535-213129535 on Build GRCh38
rs779255224 Chr.1: 213129900-213129901 on Build GRCh38
rs747874894 Chr.1: 213129776-213129776 on Build GRCh38
rs778631756 Chr.1: 213129881-213129881 on Build GRCh38
rs766825388 Chr.1: 213129650-213129650 on Build GRCh38
rs757453154 Chr.1: 213129814-213129816 on Build GRCh38
rs767203661 Chr.1: 213129836-213129836 on Build GRCh38
rs751322713 Chr.1: 213129735-213129735 on Build GRCh38
rs756751428 Chr.1: 213129506-213129506 on Build GRCh38
rs891877033 Chr.1: 213129778-213129778 on Build GRCh38
rs761187700 Chr.1: 213129646-213129646 on Build GRCh38
rs187696373 Chr.1: 213129474-213129474 on Build GRCh38
rs770783438 Chr.1: 213129620-213129620 on Build GRCh38
rs758078436 Chr.1: 213129895-213129895 on Build GRCh38
rs762648443 Chr.1: 213129902-213129902 on Build GRCh38
rs1054441923 Chr.1: 213129473-213129473 on Build GRCh38
rs376958657 Chr.1: 213129749-213129749 on Build GRCh38
rs764804553 Chr.1: 213129478-213129478 on Build GRCh38
rs199560994 Chr.1: 213129710-213129710 on Build GRCh38
rs771374489 Chr.1: 213129715-213129715 on Build GRCh38
rs775263280 Chr.1: 213129633-213129633 on Build GRCh38
rs765284722 Chr.1: 213129722-213129722 on Build GRCh38
rs777108160 Chr.1: 213129787-213129787 on Build GRCh38
rs139986699 Chr.1: 213129907-213129907 on Build GRCh38
rs781402869 Chr.1: 213129766-213129766 on Build GRCh38
rs758511305 Chr.1: 213129457-213129457 on Build GRCh38
rs369704036 Chr.1: 213129713-213129713 on Build GRCh38
rs77602608 Chr.1: 213129682-213129682 on Build GRCh38
rs372361416 Chr.1: 213129733-213129733 on Build GRCh38
rs200064577 Chr.1: 213129573-213129573 on Build GRCh38
rs958349038 Chr.1: 213129652-213129652 on Build GRCh38
rs748429567 Chr.1: 213129460-213129460 on Build GRCh38
rs375756109 Chr.1: 213129608-213129608 on Build GRCh38
rs756050938 Chr.1: 213129844-213129844 on Build GRCh38
rs367564641 Chr.1: 213129590-213129590 on Build GRCh38
rs745870658 Chr.1: 213129695-213129695 on Build GRCh38
rs753962178 Chr.1: 213129702-213129704 on Build GRCh38
rs139296657 Chr.1: 213129676-213129676 on Build GRCh38
rs757182331 Chr.1: 213129684-213129684 on Build GRCh38
rs779777949 Chr.1: 213129709-213129709 on Build GRCh38
rs150727119 Chr.1: 213129822-213129822 on Build GRCh38
rs766470816 Chr.1: 213129517-213129517 on Build GRCh38
rs770063633 Chr.1: 213129830-213129830 on Build GRCh38
rs773772139 Chr.1: 213129641-213129641 on Build GRCh38
rs763091911 Chr.1: 213129732-213129732 on Build GRCh38
rs768437981 Chr.1: 213129712-213129712 on Build GRCh38
rs750324637 Chr.1: 213129750-213129750 on Build GRCh38
rs774739990 Chr.1: 213129481-213129481 on Build GRCh38
rs147945661 Chr.1: 213129835-213129835 on Build GRCh38
rs746298886 Chr.1: 213129792-213129792 on Build GRCh38
rs753903508 Chr.1: 213129659-213129659 on Build GRCh38
rs888847297 Chr.1: 213129434-213129434 on Build GRCh38
rs745404182 Chr.1: 213129632-213129632 on Build GRCh38
rs535231866 Chr.1: 213129560-213129560 on Build GRCh38
rs573792172 Chr.1: 213129825-213129825 on Build GRCh38
rs781017308 Chr.1: 213129687-213129687 on Build GRCh38
rs750246745 Chr.1: 213129839-213129839 on Build GRCh38
rs141240814 Chr.1: 213129664-213129664 on Build GRCh38
rs750930682 Chr.1: 213129490-213129490 on Build GRCh38

View Full Product Details