Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 226601878-226602384 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STUM
Entrez Gene ID
375057
Gene Name
stum, mechanosensory transduction mediator homolog
Gene Aliases
C1orf95
UniGene
Hs.116827
Gene Chromosome Location
Chr.1: 226548238-226609214 on Build GRCh38
Transcripts
NM_001003665.3 XM_011544183.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
400424 c.406G>T p.G136C Chr.1: 226602020-226602020 on Build GRCh38
6009830 c.392-7C>T p.? Chr.1: 226601999-226601999 on Build GRCh38
6009831 c.387C>T p.S129S Chr.1: 226601999-226601999 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs745928326 Chr.1: 226601987-226601987 on Build GRCh38
rs771588128 Chr.1: 226602048-226602048 on Build GRCh38
rs551817147 Chr.1: 226602110-226602110 on Build GRCh38
rs747934058 Chr.1: 226602086-226602087 on Build GRCh38
rs780064711 Chr.1: 226602068-226602068 on Build GRCh38
rs910999745 Chr.1: 226602121-226602121 on Build GRCh38
rs1012755131 Chr.1: 226602292-226602292 on Build GRCh38
rs1014371773 Chr.1: 226602187-226602187 on Build GRCh38
rs772821879 Chr.1: 226602088-226602088 on Build GRCh38
rs201088618 Chr.1: 226601931-226601931 on Build GRCh38
rs199904485 Chr.1: 226602073-226602073 on Build GRCh38
rs41314284 Chr.1: 226602300-226602300 on Build GRCh38
rs999432237 Chr.1: 226602170-226602170 on Build GRCh38
rs1043362247 Chr.1: 226602107-226602107 on Build GRCh38
rs777757555 Chr.1: 226602064-226602064 on Build GRCh38
rs150754455 Chr.1: 226602239-226602239 on Build GRCh38
rs768297603 Chr.1: 226602359-226602359 on Build GRCh38
rs921671040 Chr.1: 226602301-226602301 on Build GRCh38
rs747648874 Chr.1: 226601913-226601913 on Build GRCh38
rs970742954 Chr.1: 226602325-226602325 on Build GRCh38
rs966253074 Chr.1: 226602298-226602298 on Build GRCh38
rs373649817 Chr.1: 226602046-226602046 on Build GRCh38
rs201235908 Chr.1: 226602319-226602319 on Build GRCh38
rs143970893 Chr.1: 226602069-226602069 on Build GRCh38
rs753174771 Chr.1: 226601945-226601945 on Build GRCh38
rs370712642 Chr.1: 226602054-226602054 on Build GRCh38
rs758304626 Chr.1: 226602053-226602053 on Build GRCh38
rs190988627 Chr.1: 226602052-226602052 on Build GRCh38
rs746836082 Chr.1: 226602071-226602071 on Build GRCh38
rs779831980 Chr.1: 226601960-226601960 on Build GRCh38
rs188971820 Chr.1: 226602058-226602058 on Build GRCh38
rs376926159 Chr.1: 226602077-226602077 on Build GRCh38
rs936545444 Chr.1: 226602092-226602092 on Build GRCh38
rs369956246 Chr.1: 226601939-226601939 on Build GRCh38
rs566180265 Chr.1: 226602235-226602235 on Build GRCh38
rs756238249 Chr.1: 226602067-226602067 on Build GRCh38
rs778075307 Chr.1: 226601948-226601948 on Build GRCh38
rs757412570 Chr.1: 226601970-226601970 on Build GRCh38
rs374374742 Chr.1: 226602159-226602159 on Build GRCh38
rs754776037 Chr.1: 226602067-226602067 on Build GRCh38
rs548602045 Chr.1: 226602284-226602284 on Build GRCh38
rs547951444 Chr.1: 226602196-226602196 on Build GRCh38
rs886678405 Chr.1: 226602257-226602257 on Build GRCh38
rs532543143 Chr.1: 226602193-226602193 on Build GRCh38
rs750691640 Chr.1: 226601964-226601967 on Build GRCh38
rs922263182 Chr.1: 226602183-226602183 on Build GRCh38
rs1053268913 Chr.1: 226602175-226602177 on Build GRCh38
rs1048041536 Chr.1: 226602194-226602194 on Build GRCh38
rs904743877 Chr.1: 226602120-226602120 on Build GRCh38
rs770303141 Chr.1: 226602090-226602090 on Build GRCh38
rs1030907196 Chr.1: 226602366-226602366 on Build GRCh38
rs12755695 Chr.1: 226602237-226602237 on Build GRCh38
rs771782149 Chr.1: 226602086-226602086 on Build GRCh38
rs772770695 Chr.1: 226602049-226602049 on Build GRCh38
rs553158 Chr.1: 226602047-226602047 on Build GRCh38
rs749158343 Chr.1: 226602037-226602037 on Build GRCh38
rs751687830 Chr.1: 226601967-226601967 on Build GRCh38
rs200120506 Chr.1: 226601982-226601982 on Build GRCh38
rs942455552 Chr.1: 226602149-226602149 on Build GRCh38
rs765153524 Chr.1: 226601908-226601908 on Build GRCh38
rs1006412244 Chr.1: 226602259-226602259 on Build GRCh38
rs41303099 Chr.1: 226602268-226602268 on Build GRCh38
rs779822024 Chr.1: 226602016-226602016 on Build GRCh38
rs760197842 Chr.1: 226602089-226602089 on Build GRCh38
rs758666178 Chr.1: 226601947-226601947 on Build GRCh38
rs1027530181 Chr.1: 226602198-226602198 on Build GRCh38
rs576568963 Chr.1: 226602365-226602365 on Build GRCh38
rs781560504 Chr.1: 226602060-226602060 on Build GRCh38
rs182763913 Chr.1: 226602057-226602057 on Build GRCh38
rs536945965 Chr.1: 226602260-226602260 on Build GRCh38
rs563856260 Chr.1: 226602162-226602162 on Build GRCh38
rs895901917 Chr.1: 226602178-226602178 on Build GRCh38
rs370391051 Chr.1: 226601999-226601999 on Build GRCh38
rs781073108 Chr.1: 226602068-226602068 on Build GRCh38
rs373516968 Chr.1: 226601907-226601907 on Build GRCh38

View Full Product Details