Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 226603643-226604137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STUM
Entrez Gene ID
375057
Gene Name
stum, mechanosensory transduction mediator homolog
Gene Aliases
C1orf95
UniGene
Hs.116827
Gene Chromosome Location
Chr.1: 226548238-226609214 on Build GRCh38
Transcripts
NM_001003665.3 XM_011544183.2

SNP Detail

SNP ID Chromosome Location
rs908065567 Chr.1: 226603685-226603685 on Build GRCh38
rs749013891 Chr.1: 226604103-226604103 on Build GRCh38
rs201698412 Chr.1: 226603806-226603806 on Build GRCh38
rs953420454 Chr.1: 226604100-226604100 on Build GRCh38
rs114210919 Chr.1: 226603701-226603701 on Build GRCh38
rs921342430 Chr.1: 226603812-226603812 on Build GRCh38
rs986855701 Chr.1: 226604060-226604060 on Build GRCh38
rs764851586 Chr.1: 226603808-226603808 on Build GRCh38
rs556801207 Chr.1: 226603669-226603669 on Build GRCh38
rs1044151904 Chr.1: 226603876-226603876 on Build GRCh38
rs761019594 Chr.1: 226603737-226603737 on Build GRCh38
rs952273205 Chr.1: 226603668-226603668 on Build GRCh38
rs1009823964 Chr.1: 226603986-226603986 on Build GRCh38
rs184479414 Chr.1: 226604083-226604083 on Build GRCh38
rs777549153 Chr.1: 226603869-226603869 on Build GRCh38
rs939968230 Chr.1: 226603833-226603833 on Build GRCh38
rs923345239 Chr.1: 226603726-226603726 on Build GRCh38
rs557636885 Chr.1: 226603988-226603988 on Build GRCh38
rs1022966938 Chr.1: 226603987-226603987 on Build GRCh38
rs971068787 Chr.1: 226603746-226603746 on Build GRCh38
rs1012678037 Chr.1: 226603683-226603683 on Build GRCh38
rs1001842306 Chr.1: 226603888-226603888 on Build GRCh38
rs188144898 Chr.1: 226603777-226603777 on Build GRCh38
rs975974456 Chr.1: 226603991-226603991 on Build GRCh38
rs572463754 Chr.1: 226604040-226604040 on Build GRCh38
rs906941380 Chr.1: 226603880-226603880 on Build GRCh38
rs975563371 Chr.1: 226603809-226603809 on Build GRCh38
rs41307716 Chr.1: 226603821-226603821 on Build GRCh38
rs986146300 Chr.1: 226604110-226604110 on Build GRCh38
rs1002552386 Chr.1: 226603890-226603890 on Build GRCh38
rs955670955 Chr.1: 226603976-226603976 on Build GRCh38
rs988768572 Chr.1: 226603779-226603779 on Build GRCh38
rs1034567934 Chr.1: 226603667-226603667 on Build GRCh38
rs1028435935 Chr.1: 226604043-226604043 on Build GRCh38
rs139807412 Chr.1: 226603873-226603873 on Build GRCh38
rs570281538 Chr.1: 226603807-226603807 on Build GRCh38
rs1051076823 Chr.1: 226603819-226603819 on Build GRCh38
rs751062337 Chr.1: 226604104-226604104 on Build GRCh38
rs976612755 Chr.1: 226603719-226603719 on Build GRCh38
rs915420523 Chr.1: 226603800-226603800 on Build GRCh38
rs888101084 Chr.1: 226603895-226603895 on Build GRCh38
rs934009388 Chr.1: 226603813-226603813 on Build GRCh38
rs1002480236 Chr.1: 226603753-226603753 on Build GRCh38
rs763690341 Chr.1: 226604072-226604072 on Build GRCh38
rs1035256522 Chr.1: 226603775-226603775 on Build GRCh38
rs931980831 Chr.1: 226603727-226603727 on Build GRCh38
rs955460549 Chr.1: 226604048-226604048 on Build GRCh38
rs925211215 Chr.1: 226603778-226603778 on Build GRCh38
rs1031310421 Chr.1: 226603970-226603970 on Build GRCh38
rs878150 Chr.1: 226603868-226603868 on Build GRCh38
rs183843401 Chr.1: 226603771-226603771 on Build GRCh38

View Full Product Details