Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 235976538-235977037 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NID1
Entrez Gene ID
4811
Gene Name
nidogen 1
Gene Aliases
NID
UniGene
Hs.356624
Gene Chromosome Location
Chr.1: 235975830-236065181 on Build GRCh38
Transcripts
NM_002508.2 XM_011544195.2

SNP Detail

SNP ID Chromosome Location
rs893816256 Chr.1: 235976845-235976845 on Build GRCh38
rs913666565 Chr.1: 235977013-235977013 on Build GRCh38
rs1020909607 Chr.1: 235976875-235976875 on Build GRCh38
rs982792857 Chr.1: 235976619-235976619 on Build GRCh38
rs189841386 Chr.1: 235976942-235976942 on Build GRCh38
rs531639394 Chr.1: 235976593-235976593 on Build GRCh38
rs73113265 Chr.1: 235976726-235976726 on Build GRCh38
rs575734406 Chr.1: 235976630-235976630 on Build GRCh38
rs926467264 Chr.1: 235976929-235976929 on Build GRCh38
rs545536699 Chr.1: 235976956-235976956 on Build GRCh38
rs970590106 Chr.1: 235976628-235976628 on Build GRCh38
rs943424604 Chr.1: 235976995-235976995 on Build GRCh38
rs1053879351 Chr.1: 235977001-235977001 on Build GRCh38
rs959016346 Chr.1: 235976688-235976688 on Build GRCh38
rs769063228 Chr.1: 235976756-235976756 on Build GRCh38
rs1007496498 Chr.1: 235976810-235976810 on Build GRCh38
rs1051788568 Chr.1: 235976782-235976782 on Build GRCh38
rs889228438 Chr.1: 235976793-235976793 on Build GRCh38
rs980617962 Chr.1: 235976927-235976927 on Build GRCh38
rs147768599 Chr.1: 235976843-235976843 on Build GRCh38
rs546179965 Chr.1: 235976651-235976651 on Build GRCh38
rs959089309 Chr.1: 235976926-235976926 on Build GRCh38
rs951422429 Chr.1: 235976618-235976618 on Build GRCh38
rs187562369 Chr.1: 235976876-235976876 on Build GRCh38
rs958217909 Chr.1: 235976606-235976606 on Build GRCh38
rs867857882 Chr.1: 235976566-235976566 on Build GRCh38
rs564120189 Chr.1: 235976957-235976957 on Build GRCh38
rs1009860372 Chr.1: 235976874-235976874 on Build GRCh38
rs933477810 Chr.1: 235976971-235976971 on Build GRCh38
rs951493482 Chr.1: 235976840-235976840 on Build GRCh38
rs760542319 Chr.1: 235976992-235976992 on Build GRCh38
rs767080819 Chr.1: 235976950-235976950 on Build GRCh38
rs773754175 Chr.1: 235976823-235976823 on Build GRCh38
rs189489022 Chr.1: 235976762-235976762 on Build GRCh38
rs772191851 Chr.1: 235976773-235976773 on Build GRCh38
rs1048311 Chr.1: 235976594-235976594 on Build GRCh38
rs891415489 Chr.1: 235976789-235976789 on Build GRCh38
rs983885716 Chr.1: 235976939-235976939 on Build GRCh38
rs1033413297 Chr.1: 235976914-235976914 on Build GRCh38
rs910738272 Chr.1: 235976991-235976991 on Build GRCh38
rs572220567 Chr.1: 235976943-235976943 on Build GRCh38
rs987621738 Chr.1: 235976984-235976984 on Build GRCh38
rs561947873 Chr.1: 235976794-235976794 on Build GRCh38
rs576311695 Chr.1: 235976765-235976765 on Build GRCh38
rs373208225 Chr.1: 235976974-235976974 on Build GRCh38
rs567933307 Chr.1: 235976607-235976607 on Build GRCh38
rs574303310 Chr.1: 235976851-235976851 on Build GRCh38
rs1003408955 Chr.1: 235976907-235976907 on Build GRCh38
rs909570454 Chr.1: 235976739-235976739 on Build GRCh38
rs1048253 Chr.1: 235976966-235976966 on Build GRCh38
rs558178049 Chr.1: 235976764-235976764 on Build GRCh38
rs535245121 Chr.1: 235976623-235976623 on Build GRCh38
rs181926770 Chr.1: 235976821-235976821 on Build GRCh38
rs942245991 Chr.1: 235976749-235976749 on Build GRCh38

View Full Product Details