Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 235979730-235980224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NID1
Entrez Gene ID
4811
Gene Name
nidogen 1
Gene Aliases
NID
UniGene
Hs.356624
Gene Chromosome Location
Chr.1: 235975830-236065181 on Build GRCh38
Transcripts
NM_002508.2 XM_011544195.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6125276 c.3478G>C p.G1160R Chr.1: 235979853-235979853 on Build GRCh38
5511664 c.3509+4C>T p.? Chr.1: 235979818-235979818 on Build GRCh38
226494 c.3421C>T p.P1141S Chr.1: 235979910-235979910 on Build GRCh38
6726094 c.3470C>T p.T1157M Chr.1: 235979861-235979861 on Build GRCh38
2000387 c.3454T>A p.Y1152N Chr.1: 235979877-235979877 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs952961904 Chr.1: 235980195-235980195 on Build GRCh38
rs869260300 Chr.1: 235980011-235980012 on Build GRCh38
rs765927256 Chr.1: 235979780-235979780 on Build GRCh38
rs745984247 Chr.1: 235979845-235979845 on Build GRCh38
rs149472196 Chr.1: 235979937-235979937 on Build GRCh38
rs376548281 Chr.1: 235979797-235979797 on Build GRCh38
rs776388939 Chr.1: 235979976-235979976 on Build GRCh38
rs997126554 Chr.1: 235979788-235979788 on Build GRCh38
rs750175837 Chr.1: 235979930-235979937 on Build GRCh38
rs764040797 Chr.1: 235979968-235979968 on Build GRCh38
rs763095893 Chr.1: 235979985-235979985 on Build GRCh38
rs866229548 Chr.1: 235979867-235979867 on Build GRCh38
rs932701811 Chr.1: 235980021-235980021 on Build GRCh38
rs1027749639 Chr.1: 235980179-235980179 on Build GRCh38
rs764290474 Chr.1: 235979805-235979805 on Build GRCh38
rs909147761 Chr.1: 235979939-235979939 on Build GRCh38
rs748880759 Chr.1: 235979861-235979861 on Build GRCh38
rs761634980 Chr.1: 235979981-235979981 on Build GRCh38
rs899827994 Chr.1: 235980019-235980019 on Build GRCh38
rs754003889 Chr.1: 235979817-235979817 on Build GRCh38
rs751115263 Chr.1: 235979784-235979784 on Build GRCh38
rs201692355 Chr.1: 235979813-235979813 on Build GRCh38
rs35565926 Chr.1: 235979889-235979889 on Build GRCh38
rs983001036 Chr.1: 235980196-235980196 on Build GRCh38
rs571178697 Chr.1: 235980058-235980058 on Build GRCh38
rs777144924 Chr.1: 235979860-235979860 on Build GRCh38
rs774123796 Chr.1: 235979956-235979956 on Build GRCh38
rs3831989 Chr.1: 235980003-235980004 on Build GRCh38
rs773945964 Chr.1: 235979886-235979886 on Build GRCh38
rs762535779 Chr.1: 235979779-235979779 on Build GRCh38
rs1040993013 Chr.1: 235979759-235979759 on Build GRCh38
rs574847647 Chr.1: 235980165-235980165 on Build GRCh38
rs763811570 Chr.1: 235979899-235979899 on Build GRCh38
rs757872662 Chr.1: 235980109-235980109 on Build GRCh38
rs747328744 Chr.1: 235979853-235979853 on Build GRCh38
rs944090942 Chr.1: 235979936-235979936 on Build GRCh38
rs1043352875 Chr.1: 235980025-235980025 on Build GRCh38
rs747489214 Chr.1: 235979908-235979908 on Build GRCh38
rs760546775 Chr.1: 235979897-235979897 on Build GRCh38
rs764430687 Chr.1: 235979841-235979842 on Build GRCh38
rs895655902 Chr.1: 235980079-235980079 on Build GRCh38
rs1022755873 Chr.1: 235980140-235980140 on Build GRCh38
rs397764310 Chr.1: 235980010-235980011 on Build GRCh38
rs369791550 Chr.1: 235979959-235979959 on Build GRCh38
rs376562147 Chr.1: 235979926-235979926 on Build GRCh38
rs775558151 Chr.1: 235979895-235979895 on Build GRCh38
rs758306083 Chr.1: 235979963-235979966 on Build GRCh38
rs758809303 Chr.1: 235979848-235979848 on Build GRCh38
rs16833032 Chr.1: 235979844-235979844 on Build GRCh38
rs904117303 Chr.1: 235980075-235980075 on Build GRCh38
rs753021292 Chr.1: 235979933-235979933 on Build GRCh38
rs556845801 Chr.1: 235980136-235980136 on Build GRCh38
rs369664139 Chr.1: 235979901-235979901 on Build GRCh38
rs759348935 Chr.1: 235979787-235979787 on Build GRCh38
rs117142664 Chr.1: 235979850-235979850 on Build GRCh38
rs189056938 Chr.1: 235980071-235980071 on Build GRCh38
rs145280265 Chr.1: 235979932-235979932 on Build GRCh38
rs144419312 Chr.1: 235980145-235980145 on Build GRCh38
rs755544534 Chr.1: 235979917-235979917 on Build GRCh38
rs114355234 Chr.1: 235979800-235979800 on Build GRCh38
rs1041112063 Chr.1: 235979992-235979992 on Build GRCh38
rs770735060 Chr.1: 235979869-235979869 on Build GRCh38
rs938436463 Chr.1: 235980015-235980015 on Build GRCh38
rs774869080 Chr.1: 235979966-235979966 on Build GRCh38
rs1052608288 Chr.1: 235980126-235980126 on Build GRCh38
rs756402815 Chr.1: 235979982-235979982 on Build GRCh38
rs778250771 Chr.1: 235980059-235980059 on Build GRCh38
rs767542741 Chr.1: 235979794-235979794 on Build GRCh38
rs200587626 Chr.1: 235979824-235979824 on Build GRCh38
rs200339931 Chr.1: 235980012-235980012 on Build GRCh38
rs528175020 Chr.1: 235980014-235980014 on Build GRCh38
rs780410514 Chr.1: 235979907-235979907 on Build GRCh38
rs971882676 Chr.1: 235979990-235979990 on Build GRCh38
rs371653356 Chr.1: 235979778-235979778 on Build GRCh38
rs760623411 Chr.1: 235979967-235979967 on Build GRCh38
rs569218445 Chr.1: 235980197-235980197 on Build GRCh38
rs115093040 Chr.1: 235979854-235979854 on Build GRCh38
rs1006660994 Chr.1: 235980187-235980187 on Build GRCh38
rs373203237 Chr.1: 235979905-235979905 on Build GRCh38
rs1014012051 Chr.1: 235980116-235980116 on Build GRCh38
rs908675778 Chr.1: 235980200-235980200 on Build GRCh38
rs577463598 Chr.1: 235980002-235980003 on Build GRCh38
rs546641377 Chr.1: 235980030-235980030 on Build GRCh38
rs769698854 Chr.1: 235980047-235980047 on Build GRCh38
rs769297488 Chr.1: 235979776-235979776 on Build GRCh38
rs375292088 Chr.1: 235979900-235979900 on Build GRCh38
rs999756627 Chr.1: 235980117-235980117 on Build GRCh38

View Full Product Details