Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 237884691-237885204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100130331
Entrez Gene ID
100130331
Gene Name
POTE ankyrin domain family, member F pseudogene
Gene Aliases
-
UniGene
Hs.661736
Gene Chromosome Location
Chr.1: 237862175-237928321 on Build GRCh38
Transcripts
NR_027247.2 NM_021186.4

Gene Symbol
ZP4
Entrez Gene ID
57829
Gene Name
zona pellucida glycoprotein 4
Gene Aliases
ZBP, ZP1, ZPB, Zp-4
UniGene
Hs.136241
Gene Chromosome Location
Chr.1: 237877864-237890922 on Build GRCh38
Transcripts
NR_027247.2 NM_021186.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6061818 c.1309C>A p.P437T Chr.1: 237885167-237885167 on Build GRCh38
6061817 c.1284A>T p.T428T Chr.1: 237885192-237885192 on Build GRCh38
1340516 c.1345C>T p.P449S Chr.1: 237884814-237884814 on Build GRCh38
3485900 c.1362C>T p.S454S Chr.1: 237884797-237884797 on Build GRCh38
7399840 c.1305G>T p.R435S Chr.1: 237885171-237885171 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1016511679 Chr.1: 237885177-237885177 on Build GRCh38
rs749791394 Chr.1: 237884848-237884848 on Build GRCh38
rs987672524 Chr.1: 237885049-237885049 on Build GRCh38
rs377446136 Chr.1: 237885062-237885062 on Build GRCh38
rs568918206 Chr.1: 237885111-237885111 on Build GRCh38
rs751843901 Chr.1: 237884824-237884824 on Build GRCh38
rs199631612 Chr.1: 237884830-237884830 on Build GRCh38
rs373960078 Chr.1: 237884778-237884778 on Build GRCh38
rs767641159 Chr.1: 237885141-237885141 on Build GRCh38
rs755105340 Chr.1: 237884735-237884735 on Build GRCh38
rs779513702 Chr.1: 237885159-237885159 on Build GRCh38
rs760401172 Chr.1: 237884768-237884768 on Build GRCh38
rs755160197 Chr.1: 237884825-237884825 on Build GRCh38
rs771493289 Chr.1: 237884860-237884860 on Build GRCh38
rs922177842 Chr.1: 237884789-237884789 on Build GRCh38
rs767523527 Chr.1: 237885023-237885023 on Build GRCh38
rs778761115 Chr.1: 237885172-237885173 on Build GRCh38
rs202090191 Chr.1: 237885156-237885156 on Build GRCh38
rs757440065 Chr.1: 237885169-237885169 on Build GRCh38
rs753696272 Chr.1: 237885137-237885140 on Build GRCh38
rs946537876 Chr.1: 237884878-237884878 on Build GRCh38
rs780721219 Chr.1: 237885172-237885172 on Build GRCh38
rs1007674719 Chr.1: 237885173-237885173 on Build GRCh38
rs181228543 Chr.1: 237885084-237885084 on Build GRCh38
rs758172052 Chr.1: 237884722-237884722 on Build GRCh38
rs542908414 Chr.1: 237884904-237884904 on Build GRCh38
rs769057238 Chr.1: 237884856-237884856 on Build GRCh38
rs781241984 Chr.1: 237884742-237884742 on Build GRCh38
rs759124039 Chr.1: 237885000-237885000 on Build GRCh38
rs541608710 Chr.1: 237884814-237884814 on Build GRCh38
rs769290469 Chr.1: 237884897-237884897 on Build GRCh38
rs953599704 Chr.1: 237885039-237885039 on Build GRCh38
rs748725827 Chr.1: 237884748-237884748 on Build GRCh38
rs997716205 Chr.1: 237884933-237884933 on Build GRCh38
rs936692979 Chr.1: 237885114-237885114 on Build GRCh38
rs757169352 Chr.1: 237885168-237885169 on Build GRCh38
rs563725129 Chr.1: 237884749-237884749 on Build GRCh38
rs889396172 Chr.1: 237885013-237885013 on Build GRCh38
rs915705523 Chr.1: 237885089-237885089 on Build GRCh38
rs777907570 Chr.1: 237885154-237885154 on Build GRCh38
rs746266633 Chr.1: 237885165-237885165 on Build GRCh38
rs1015892515 Chr.1: 237885035-237885035 on Build GRCh38
rs963050211 Chr.1: 237885050-237885050 on Build GRCh38
rs201485611 Chr.1: 237885153-237885153 on Build GRCh38
rs746710083 Chr.1: 237884867-237884867 on Build GRCh38
rs1029555717 Chr.1: 237885031-237885031 on Build GRCh38
rs759274000 Chr.1: 237885137-237885137 on Build GRCh38
rs545550765 Chr.1: 237884956-237884956 on Build GRCh38
rs914971321 Chr.1: 237885080-237885080 on Build GRCh38
rs779391875 Chr.1: 237884866-237884866 on Build GRCh38
rs2298100 Chr.1: 237885025-237885025 on Build GRCh38
rs143904065 Chr.1: 237884801-237884801 on Build GRCh38
rs762748345 Chr.1: 237884817-237884817 on Build GRCh38
rs749623600 Chr.1: 237884755-237884755 on Build GRCh38
rs762869598 Chr.1: 237884899-237884899 on Build GRCh38
rs953775523 Chr.1: 237884795-237884795 on Build GRCh38
rs762506331 Chr.1: 237885052-237885052 on Build GRCh38
rs747174689 Chr.1: 237884734-237884734 on Build GRCh38
rs116738146 Chr.1: 237884927-237884927 on Build GRCh38
rs766148332 Chr.1: 237884822-237884822 on Build GRCh38
rs776308144 Chr.1: 237884892-237884892 on Build GRCh38
rs532075292 Chr.1: 237885059-237885059 on Build GRCh38
rs1004889650 Chr.1: 237884994-237884994 on Build GRCh38
rs202241361 Chr.1: 237884769-237884769 on Build GRCh38
rs978639023 Chr.1: 237885104-237885104 on Build GRCh38
rs775981829 Chr.1: 237884771-237884771 on Build GRCh38
rs778160622 Chr.1: 237884838-237884838 on Build GRCh38
rs773079353 Chr.1: 237884898-237884898 on Build GRCh38
rs768445867 Chr.1: 237884889-237884889 on Build GRCh38
rs762732432 Chr.1: 237884920-237884920 on Build GRCh38
rs956337189 Chr.1: 237885058-237885058 on Build GRCh38
rs895812061 Chr.1: 237884992-237884992 on Build GRCh38
rs35651972 Chr.1: 237885166-237885166 on Build GRCh38
rs752910899 Chr.1: 237885143-237885143 on Build GRCh38
rs546203250 Chr.1: 237884986-237884986 on Build GRCh38
rs1052414279 Chr.1: 237885119-237885119 on Build GRCh38
rs115277322 Chr.1: 237884894-237884894 on Build GRCh38
rs994991635 Chr.1: 237885029-237885029 on Build GRCh38
rs948666141 Chr.1: 237885090-237885090 on Build GRCh38
rs771366793 Chr.1: 237884757-237884757 on Build GRCh38
rs529902072 Chr.1: 237885124-237885124 on Build GRCh38
rs781377450 Chr.1: 237884829-237884829 on Build GRCh38
rs1015051235 Chr.1: 237884739-237884739 on Build GRCh38
rs780258033 Chr.1: 237884733-237884733 on Build GRCh38
rs765117673 Chr.1: 237884783-237884783 on Build GRCh38
rs766199912 Chr.1: 237885115-237885115 on Build GRCh38
rs188722689 Chr.1: 237884957-237884957 on Build GRCh38
rs777513126 Chr.1: 237884819-237884819 on Build GRCh38

View Full Product Details