Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 245927840-245928346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMYD3
Entrez Gene ID
64754
Gene Name
SET and MYND domain containing 3
Gene Aliases
KMT3E, ZMYND1, ZNFN3A1, bA74P14.1
UniGene
Hs.567571
Gene Chromosome Location
Chr.1: 245749340-246507342 on Build GRCh38
Transcripts
NM_001167740.1 NM_022743.2 XM_011544253.1 XM_011544254.2 XM_011544257.1 XM_011544258.2 XM_011544259.2 XM_011544260.1 XM_011544261.2 XM_017002094.1 XM_017002095.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1724304 c.600-5delT p.? Chr.1: 245928038-245928038 on Build GRCh38
4728612 c.494G>A p.R165Q Chr.1: 245927962-245927962 on Build GRCh38
1734034 c.638C>T p.S213L Chr.1: 245927995-245927995 on Build GRCh38
1602044 c.687C>A p.I229I Chr.1: 245927946-245927946 on Build GRCh38
2051952 c.462G>A p.S154S Chr.1: 245927994-245927994 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371218690 Chr.1: 245927938-245927938 on Build GRCh38
rs769898823 Chr.1: 245928029-245928029 on Build GRCh38
rs779988023 Chr.1: 245928050-245928050 on Build GRCh38
rs140568346 Chr.1: 245927891-245927891 on Build GRCh38
rs201266204 Chr.1: 245928000-245928000 on Build GRCh38
rs755914433 Chr.1: 245927962-245927962 on Build GRCh38
rs144971092 Chr.1: 245928214-245928214 on Build GRCh38
rs916691160 Chr.1: 245927902-245927902 on Build GRCh38
rs763055220 Chr.1: 245928046-245928046 on Build GRCh38
rs753112522 Chr.1: 245928046-245928047 on Build GRCh38
rs1056564524 Chr.1: 245928139-245928139 on Build GRCh38
rs769217941 Chr.1: 245927933-245927933 on Build GRCh38
rs1050586090 Chr.1: 245928226-245928226 on Build GRCh38
rs778618992 Chr.1: 245927908-245927908 on Build GRCh38
rs386641635 Chr.1: 245928155-245928157 on Build GRCh38
rs147009044 Chr.1: 245927945-245927945 on Build GRCh38
rs963028514 Chr.1: 245928278-245928278 on Build GRCh38
rs11544848 Chr.1: 245927978-245927978 on Build GRCh38
rs572069758 Chr.1: 245928237-245928237 on Build GRCh38
rs192767045 Chr.1: 245928098-245928098 on Build GRCh38
rs530459126 Chr.1: 245927876-245927876 on Build GRCh38
rs776092769 Chr.1: 245928155-245928156 on Build GRCh38
rs753788836 Chr.1: 245928080-245928080 on Build GRCh38
rs760455663 Chr.1: 245928065-245928065 on Build GRCh38
rs546712948 Chr.1: 245928222-245928222 on Build GRCh38
rs1030664084 Chr.1: 245928322-245928322 on Build GRCh38
rs1021923865 Chr.1: 245928066-245928066 on Build GRCh38
rs369541466 Chr.1: 245928001-245928001 on Build GRCh38
rs41308150 Chr.1: 245927974-245927974 on Build GRCh38
rs201150925 Chr.1: 245928155-245928157 on Build GRCh38
rs1037231159 Chr.1: 245928125-245928125 on Build GRCh38
rs770403010 Chr.1: 245927907-245927907 on Build GRCh38
rs903791813 Chr.1: 245928201-245928201 on Build GRCh38
rs756756709 Chr.1: 245927970-245927970 on Build GRCh38
rs777292029 Chr.1: 245927934-245927934 on Build GRCh38
rs781639648 Chr.1: 245928038-245928038 on Build GRCh38
rs374960548 Chr.1: 245928063-245928063 on Build GRCh38
rs777721683 Chr.1: 245927901-245927901 on Build GRCh38
rs777190405 Chr.1: 245928019-245928019 on Build GRCh38
rs41308148 Chr.1: 245928048-245928048 on Build GRCh38
rs182731807 Chr.1: 245928110-245928110 on Build GRCh38
rs745405067 Chr.1: 245927994-245927994 on Build GRCh38
rs1043721955 Chr.1: 245928186-245928186 on Build GRCh38
rs2275318 Chr.1: 245928213-245928213 on Build GRCh38
rs375219309 Chr.1: 245927911-245927911 on Build GRCh38
rs769635424 Chr.1: 245928256-245928256 on Build GRCh38
rs566281194 Chr.1: 245927954-245927954 on Build GRCh38
rs745543077 Chr.1: 245927909-245927909 on Build GRCh38
rs564530785 Chr.1: 245927867-245927867 on Build GRCh38
rs557341272 Chr.1: 245928114-245928114 on Build GRCh38
rs753782195 Chr.1: 245927966-245927966 on Build GRCh38
rs751233491 Chr.1: 245928035-245928036 on Build GRCh38
rs942456141 Chr.1: 245928234-245928234 on Build GRCh38
rs999383419 Chr.1: 245928038-245928038 on Build GRCh38
rs748643391 Chr.1: 245928022-245928022 on Build GRCh38
rs2275317 Chr.1: 245928163-245928163 on Build GRCh38
rs149059384 Chr.1: 245928254-245928254 on Build GRCh38
rs6669768 Chr.1: 245928155-245928155 on Build GRCh38
rs2275315 Chr.1: 245928156-245928156 on Build GRCh38
rs377154353 Chr.1: 245928178-245928178 on Build GRCh38
rs941591524 Chr.1: 245928117-245928117 on Build GRCh38
rs761150059 Chr.1: 245927916-245927916 on Build GRCh38
rs775947540 Chr.1: 245927914-245927914 on Build GRCh38
rs765301713 Chr.1: 245927946-245927946 on Build GRCh38
rs755376291 Chr.1: 245928037-245928038 on Build GRCh38
rs748770973 Chr.1: 245927905-245927905 on Build GRCh38
rs752451818 Chr.1: 245927958-245927958 on Build GRCh38
rs781008447 Chr.1: 245927882-245927882 on Build GRCh38
rs756647101 Chr.1: 245928058-245928059 on Build GRCh38
rs879597324 Chr.1: 245927871-245927871 on Build GRCh38
rs551309933 Chr.1: 245927866-245927866 on Build GRCh38
rs769203532 Chr.1: 245928008-245928008 on Build GRCh38
rs529441035 Chr.1: 245927889-245927889 on Build GRCh38
rs543109958 Chr.1: 245928130-245928130 on Build GRCh38
rs973393179 Chr.1: 245928286-245928286 on Build GRCh38
rs903476512 Chr.1: 245928079-245928079 on Build GRCh38
rs530240837 Chr.1: 245928157-245928158 on Build GRCh38
rs984447113 Chr.1: 245928257-245928257 on Build GRCh38
rs558521752 Chr.1: 245928052-245928052 on Build GRCh38
rs2275316 Chr.1: 245928157-245928157 on Build GRCh38
rs1016332221 Chr.1: 245928268-245928268 on Build GRCh38
rs758239475 Chr.1: 245928067-245928067 on Build GRCh38
rs574008962 Chr.1: 245928128-245928128 on Build GRCh38
rs766035692 Chr.1: 245927954-245927954 on Build GRCh38
rs371775320 Chr.1: 245928033-245928033 on Build GRCh38
rs939521906 Chr.1: 245928304-245928304 on Build GRCh38
rs6669866 Chr.1: 245928229-245928229 on Build GRCh38
rs879268111 Chr.1: 245928155-245928157 on Build GRCh38
rs2275314 Chr.1: 245928134-245928134 on Build GRCh38
rs74523188 Chr.1: 245928030-245928030 on Build GRCh38
rs552023426 Chr.1: 245928321-245928321 on Build GRCh38
rs1012266518 Chr.1: 245928173-245928173 on Build GRCh38
rs757093070 Chr.1: 245928082-245928082 on Build GRCh38
rs758081490 Chr.1: 245927995-245927995 on Build GRCh38
rs543829825 Chr.1: 245928199-245928199 on Build GRCh38
rs549272016 Chr.1: 245927953-245927953 on Build GRCh38
rs763912023 Chr.1: 245928077-245928077 on Build GRCh38
rs777672375 Chr.1: 245927963-245927963 on Build GRCh38
rs918807163 Chr.1: 245928303-245928303 on Build GRCh38
rs1031232211 Chr.1: 245928040-245928040 on Build GRCh38

View Full Product Details