Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 247589075-247589582 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR2G2
Entrez Gene ID
81470
Gene Name
olfactory receptor family 2 subfamily G member 2
Gene Aliases
OR1-32
UniGene
Hs.690208
Gene Chromosome Location
Chr.1: 247588360-247589313 on Build GRCh38
Transcripts
NM_001001915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6348317 c.796G>T p.A266S Chr.1: 247589155-247589155 on Build GRCh38
3486725 c.947T>A p.I316N Chr.1: 247589306-247589306 on Build GRCh38
4709770 c.801G>A p.K267K Chr.1: 247589160-247589160 on Build GRCh38
7089805 c.725G>A p.G242E Chr.1: 247589084-247589084 on Build GRCh38
4030571 c.834T>C p.S278S Chr.1: 247589193-247589193 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs952634676 Chr.1: 247589324-247589324 on Build GRCh38
rs767092529 Chr.1: 247589292-247589292 on Build GRCh38
rs971319070 Chr.1: 247589388-247589388 on Build GRCh38
rs371732298 Chr.1: 247589120-247589120 on Build GRCh38
rs35280339 Chr.1: 247589349-247589349 on Build GRCh38
rs150772119 Chr.1: 247589417-247589417 on Build GRCh38
rs978252582 Chr.1: 247589493-247589493 on Build GRCh38
rs757903767 Chr.1: 247589325-247589326 on Build GRCh38
rs758506476 Chr.1: 247589329-247589329 on Build GRCh38
rs180926135 Chr.1: 247589431-247589431 on Build GRCh38
rs772134729 Chr.1: 247589221-247589221 on Build GRCh38
rs1044262575 Chr.1: 247589508-247589508 on Build GRCh38
rs756487359 Chr.1: 247589109-247589109 on Build GRCh38
rs944504116 Chr.1: 247589510-247589510 on Build GRCh38
rs59002362 Chr.1: 247589420-247589420 on Build GRCh38
rs766309780 Chr.1: 247589175-247589175 on Build GRCh38
rs112232857 Chr.1: 247589325-247589325 on Build GRCh38
rs749769720 Chr.1: 247589111-247589111 on Build GRCh38
rs934692536 Chr.1: 247589422-247589422 on Build GRCh38
rs376766612 Chr.1: 247589107-247589107 on Build GRCh38
rs745809995 Chr.1: 247589520-247589520 on Build GRCh38
rs780891464 Chr.1: 247589106-247589106 on Build GRCh38
rs565147584 Chr.1: 247589366-247589366 on Build GRCh38
rs200950288 Chr.1: 247589238-247589238 on Build GRCh38
rs770852923 Chr.1: 247589348-247589349 on Build GRCh38
rs761612257 Chr.1: 247589135-247589135 on Build GRCh38
rs770689849 Chr.1: 247589334-247589334 on Build GRCh38
rs767288630 Chr.1: 247589184-247589184 on Build GRCh38
rs770609388 Chr.1: 247589098-247589098 on Build GRCh38
rs77560957 Chr.1: 247589405-247589405 on Build GRCh38
rs778203734 Chr.1: 247589337-247589338 on Build GRCh38
rs759070345 Chr.1: 247589181-247589181 on Build GRCh38
rs750876826 Chr.1: 247589328-247589328 on Build GRCh38
rs150355891 Chr.1: 247589200-247589200 on Build GRCh38
rs752147426 Chr.1: 247589142-247589142 on Build GRCh38
rs762751266 Chr.1: 247589165-247589165 on Build GRCh38
rs567939635 Chr.1: 247589486-247589486 on Build GRCh38
rs1042999056 Chr.1: 247589418-247589418 on Build GRCh38
rs752329312 Chr.1: 247589193-247589193 on Build GRCh38
rs74152638 Chr.1: 247589216-247589216 on Build GRCh38
rs914931347 Chr.1: 247589436-247589436 on Build GRCh38
rs745766002 Chr.1: 247589353-247589353 on Build GRCh38
rs755258702 Chr.1: 247589331-247589331 on Build GRCh38
rs895911770 Chr.1: 247589451-247589451 on Build GRCh38
rs149547123 Chr.1: 247589215-247589215 on Build GRCh38
rs747192851 Chr.1: 247589228-247589228 on Build GRCh38
rs769513033 Chr.1: 247589136-247589136 on Build GRCh38
rs756355531 Chr.1: 247589415-247589415 on Build GRCh38
rs199975925 Chr.1: 247589247-247589247 on Build GRCh38
rs769599869 Chr.1: 247589233-247589233 on Build GRCh38
rs1015232920 Chr.1: 247589548-247589548 on Build GRCh38
rs865888004 Chr.1: 247589133-247589133 on Build GRCh38
rs749925600 Chr.1: 247589319-247589319 on Build GRCh38
rs764793959 Chr.1: 247589110-247589110 on Build GRCh38
rs763386328 Chr.1: 247589263-247589263 on Build GRCh38
rs754236597 Chr.1: 247589210-247589210 on Build GRCh38
rs774183673 Chr.1: 247589352-247589352 on Build GRCh38
rs189686159 Chr.1: 247589402-247589402 on Build GRCh38
rs749048017 Chr.1: 247589332-247589332 on Build GRCh38
rs888339913 Chr.1: 247589529-247589529 on Build GRCh38
rs1005473407 Chr.1: 247589530-247589530 on Build GRCh38
rs774585851 Chr.1: 247589349-247589350 on Build GRCh38
rs752415547 Chr.1: 247589293-247589293 on Build GRCh38
rs1038987609 Chr.1: 247589323-247589323 on Build GRCh38
rs747070219 Chr.1: 247589330-247589330 on Build GRCh38
rs760262201 Chr.1: 247589362-247589362 on Build GRCh38
rs910829928 Chr.1: 247589149-247589149 on Build GRCh38
rs528210855 Chr.1: 247589448-247589448 on Build GRCh38
rs1015726215 Chr.1: 247589377-247589377 on Build GRCh38
rs1038914039 Chr.1: 247589557-247589557 on Build GRCh38
rs550612779 Chr.1: 247589509-247589509 on Build GRCh38
rs1003729313 Chr.1: 247589403-247589403 on Build GRCh38
rs574198356 Chr.1: 247589327-247589327 on Build GRCh38
rs754227626 Chr.1: 247589311-247589311 on Build GRCh38
rs770830134 Chr.1: 247589249-247589249 on Build GRCh38
rs780262118 Chr.1: 247589224-247589224 on Build GRCh38
rs771019766 Chr.1: 247589462-247589462 on Build GRCh38
rs758970434 Chr.1: 247589291-247589291 on Build GRCh38
rs751096514 Chr.1: 247589180-247589180 on Build GRCh38
rs775678329 Chr.1: 247589132-247589132 on Build GRCh38
rs888535069 Chr.1: 247589346-247589346 on Build GRCh38
rs1013204332 Chr.1: 247589252-247589252 on Build GRCh38
rs267598474 Chr.1: 247589295-247589295 on Build GRCh38
rs771061644 Chr.1: 247589119-247589119 on Build GRCh38
rs765129946 Chr.1: 247589318-247589319 on Build GRCh38
rs748540501 Chr.1: 247589327-247589328 on Build GRCh38
rs1027020712 Chr.1: 247589342-247589342 on Build GRCh38
rs757180401 Chr.1: 247589272-247589274 on Build GRCh38
rs781697532 Chr.1: 247589365-247589365 on Build GRCh38
rs749374035 Chr.1: 247589435-247589435 on Build GRCh38
rs562228291 Chr.1: 247589326-247589326 on Build GRCh38
rs775184371 Chr.1: 247589359-247589359 on Build GRCh38
rs756575209 Chr.1: 247589196-247589196 on Build GRCh38
rs772109020 Chr.1: 247589358-247589358 on Build GRCh38
rs186342971 Chr.1: 247589496-247589496 on Build GRCh38
rs138403077 Chr.1: 247589163-247589163 on Build GRCh38
rs745933717 Chr.1: 247589123-247589123 on Build GRCh38
rs961075095 Chr.1: 247589340-247589340 on Build GRCh38
rs774284853 Chr.1: 247589279-247589279 on Build GRCh38
rs773030936 Chr.1: 247589237-247589237 on Build GRCh38
rs546271683 Chr.1: 247589483-247589483 on Build GRCh38
rs573326534 Chr.1: 247589317-247589317 on Build GRCh38
rs954884823 Chr.1: 247589372-247589372 on Build GRCh38
rs778912851 Chr.1: 247589122-247589122 on Build GRCh38

View Full Product Details