Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 247815377-247815885 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR14A16
Entrez Gene ID
284532
Gene Name
olfactory receptor family 14 subfamily A member 16
Gene Aliases
OR5AT1
UniGene
Hs.626620
Gene Chromosome Location
Chr.1: 247814800-247815729 on Build GRCh38
Transcripts
NM_001001966.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
146851 c.169G>A p.V57M Chr.1: 247815561-247815561 on Build GRCh38
6037290 c.168C>T p.P56P Chr.1: 247815562-247815562 on Build GRCh38
3400593 c.350C>T p.S117F Chr.1: 247815380-247815380 on Build GRCh38
7381232 c.162C>A p.H54Q Chr.1: 247815568-247815568 on Build GRCh38
6056783 c.41G>A p.G14E Chr.1: 247815689-247815689 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs893606356 Chr.1: 247815564-247815564 on Build GRCh38
rs767461899 Chr.1: 247815732-247815732 on Build GRCh38
rs200956472 Chr.1: 247815583-247815583 on Build GRCh38
rs370400033 Chr.1: 247815727-247815727 on Build GRCh38
rs756426074 Chr.1: 247815696-247815696 on Build GRCh38
rs890359031 Chr.1: 247815853-247815853 on Build GRCh38
rs896900910 Chr.1: 247815543-247815543 on Build GRCh38
rs546776676 Chr.1: 247815604-247815604 on Build GRCh38
rs773654431 Chr.1: 247815653-247815653 on Build GRCh38
rs761472828 Chr.1: 247815651-247815651 on Build GRCh38
rs749504272 Chr.1: 247815709-247815709 on Build GRCh38
rs200122075 Chr.1: 247815712-247815712 on Build GRCh38
rs763778071 Chr.1: 247815809-247815809 on Build GRCh38
rs746353304 Chr.1: 247815429-247815429 on Build GRCh38
rs759520297 Chr.1: 247815548-247815548 on Build GRCh38
rs149557201 Chr.1: 247815457-247815457 on Build GRCh38
rs781601484 Chr.1: 247815542-247815542 on Build GRCh38
rs771871126 Chr.1: 247815623-247815623 on Build GRCh38
rs199954467 Chr.1: 247815512-247815512 on Build GRCh38
rs369360768 Chr.1: 247815757-247815759 on Build GRCh38
rs752072873 Chr.1: 247815593-247815593 on Build GRCh38
rs762650845 Chr.1: 247815776-247815776 on Build GRCh38
rs774475321 Chr.1: 247815467-247815467 on Build GRCh38
rs1005732876 Chr.1: 247815510-247815510 on Build GRCh38
rs865945451 Chr.1: 247815427-247815427 on Build GRCh38
rs191948291 Chr.1: 247815782-247815782 on Build GRCh38
rs758970004 Chr.1: 247815589-247815589 on Build GRCh38
rs200090728 Chr.1: 247815451-247815451 on Build GRCh38
rs1036703557 Chr.1: 247815519-247815519 on Build GRCh38
rs547883422 Chr.1: 247815669-247815669 on Build GRCh38
rs79307181 Chr.1: 247815640-247815640 on Build GRCh38
rs530186109 Chr.1: 247815561-247815561 on Build GRCh38
rs779896731 Chr.1: 247815721-247815721 on Build GRCh38
rs980022218 Chr.1: 247815747-247815747 on Build GRCh38
rs746955953 Chr.1: 247815730-247815730 on Build GRCh38
rs11204514 Chr.1: 247815759-247815759 on Build GRCh38
rs367893985 Chr.1: 247815491-247815491 on Build GRCh38
rs774283113 Chr.1: 247815546-247815546 on Build GRCh38
rs937875009 Chr.1: 247815527-247815527 on Build GRCh38
rs768706376 Chr.1: 247815461-247815461 on Build GRCh38
rs760284306 Chr.1: 247815542-247815542 on Build GRCh38
rs753547702 Chr.1: 247815577-247815577 on Build GRCh38
rs201642586 Chr.1: 247815535-247815535 on Build GRCh38
rs143397542 Chr.1: 247815556-247815556 on Build GRCh38
rs772785246 Chr.1: 247815764-247815764 on Build GRCh38
rs768183236 Chr.1: 247815775-247815776 on Build GRCh38
rs148164063 Chr.1: 247815652-247815652 on Build GRCh38
rs759377699 Chr.1: 247815469-247815469 on Build GRCh38
rs773160402 Chr.1: 247815463-247815463 on Build GRCh38
rs577712203 Chr.1: 247815801-247815802 on Build GRCh38
rs763697318 Chr.1: 247815493-247815493 on Build GRCh38
rs777898392 Chr.1: 247815697-247815697 on Build GRCh38
rs867530636 Chr.1: 247815525-247815525 on Build GRCh38
rs765917147 Chr.1: 247815511-247815512 on Build GRCh38
rs758768765 Chr.1: 247815508-247815508 on Build GRCh38
rs770022217 Chr.1: 247815562-247815562 on Build GRCh38
rs866940782 Chr.1: 247815458-247815458 on Build GRCh38
rs561271503 Chr.1: 247815538-247815538 on Build GRCh38
rs755536505 Chr.1: 247815688-247815688 on Build GRCh38
rs61740923 Chr.1: 247815466-247815466 on Build GRCh38
rs763572953 Chr.1: 247815573-247815573 on Build GRCh38
rs374382731 Chr.1: 247815846-247815846 on Build GRCh38
rs772255559 Chr.1: 247815444-247815444 on Build GRCh38
rs762792975 Chr.1: 247815465-247815465 on Build GRCh38
rs372061599 Chr.1: 247815565-247815565 on Build GRCh38
rs752060227 Chr.1: 247815674-247815674 on Build GRCh38
rs140410550 Chr.1: 247815595-247815595 on Build GRCh38
rs893647840 Chr.1: 247815840-247815840 on Build GRCh38
rs146819402 Chr.1: 247815506-247815506 on Build GRCh38
rs769312981 Chr.1: 247815762-247815762 on Build GRCh38
rs781405230 Chr.1: 247815599-247815599 on Build GRCh38
rs267598483 Chr.1: 247815662-247815662 on Build GRCh38
rs778780443 Chr.1: 247815614-247815614 on Build GRCh38
rs776145969 Chr.1: 247815748-247815748 on Build GRCh38
rs10888249 Chr.1: 247815694-247815694 on Build GRCh38
rs767709551 Chr.1: 247815692-247815692 on Build GRCh38
rs369712674 Chr.1: 247815600-247815600 on Build GRCh38
rs757667932 Chr.1: 247815505-247815505 on Build GRCh38
rs775341631 Chr.1: 247815624-247815624 on Build GRCh38
rs774862927 Chr.1: 247815544-247815544 on Build GRCh38
rs774836330 Chr.1: 247815538-247815539 on Build GRCh38
rs56958172 Chr.1: 247815622-247815622 on Build GRCh38
rs745757180 Chr.1: 247815443-247815444 on Build GRCh38
rs779437415 Chr.1: 247815408-247815408 on Build GRCh38
rs568853673 Chr.1: 247815844-247815844 on Build GRCh38
rs267598484 Chr.1: 247815722-247815722 on Build GRCh38
rs529649606 Chr.1: 247815668-247815668 on Build GRCh38
rs1056981019 Chr.1: 247815484-247815484 on Build GRCh38
rs1008257836 Chr.1: 247815855-247815855 on Build GRCh38
rs138666893 Chr.1: 247815847-247815847 on Build GRCh38
rs767121430 Chr.1: 247815472-247815472 on Build GRCh38
rs755947181 Chr.1: 247815806-247815806 on Build GRCh38
rs904944361 Chr.1: 247815521-247815521 on Build GRCh38
rs752078813 Chr.1: 247815477-247815477 on Build GRCh38
rs139638287 Chr.1: 247815711-247815711 on Build GRCh38
rs564801640 Chr.1: 247815503-247815503 on Build GRCh38
rs910539927 Chr.1: 247815415-247815415 on Build GRCh38
rs1053438226 Chr.1: 247815827-247815827 on Build GRCh38
rs983412632 Chr.1: 247815486-247815486 on Build GRCh38
rs780198575 Chr.1: 247815511-247815511 on Build GRCh38
rs956106792 Chr.1: 247815679-247815679 on Build GRCh38
rs751009197 Chr.1: 247815507-247815507 on Build GRCh38
rs768951334 Chr.1: 247815532-247815532 on Build GRCh38
rs975568490 Chr.1: 247815450-247815450 on Build GRCh38
rs926398770 Chr.1: 247815801-247815801 on Build GRCh38
rs750697533 Chr.1: 247815584-247815584 on Build GRCh38
rs933727352 Chr.1: 247815829-247815829 on Build GRCh38
rs200580825 Chr.1: 247815490-247815490 on Build GRCh38
rs150693558 Chr.1: 247815655-247815655 on Build GRCh38
rs780382048 Chr.1: 247815592-247815592 on Build GRCh38

View Full Product Details