Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 247841634-247842135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR11L1
Entrez Gene ID
391189
Gene Name
olfactory receptor family 11 subfamily L member 1
Gene Aliases
-
UniGene
Hs.553781
Gene Chromosome Location
Chr.1: 247840928-247841896 on Build GRCh38
Transcripts
NM_001001959.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5685107 c.105C>A p.Y35* Chr.1: 247841792-247841792 on Build GRCh38
2232523 c.263G>T p.W88L Chr.1: 247841634-247841634 on Build GRCh38
7080626 c.20C>G p.S7C Chr.1: 247841877-247841877 on Build GRCh38
2232526 c.153G>C p.Q51H Chr.1: 247841744-247841744 on Build GRCh38
5695499 c.151C>A p.Q51K Chr.1: 247841746-247841746 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs964239332 Chr.1: 247841907-247841907 on Build GRCh38
rs373451280 Chr.1: 247841771-247841771 on Build GRCh38
rs767690956 Chr.1: 247841716-247841716 on Build GRCh38
rs763388529 Chr.1: 247841668-247841668 on Build GRCh38
rs753984060 Chr.1: 247841804-247841804 on Build GRCh38
rs760838160 Chr.1: 247841793-247841793 on Build GRCh38
rs766949526 Chr.1: 247841912-247841914 on Build GRCh38
rs771403367 Chr.1: 247841768-247841768 on Build GRCh38
rs754848682 Chr.1: 247841819-247841819 on Build GRCh38
rs201418677 Chr.1: 247841855-247841855 on Build GRCh38
rs751499973 Chr.1: 247841818-247841818 on Build GRCh38
rs760133636 Chr.1: 247841857-247841857 on Build GRCh38
rs934824898 Chr.1: 247841995-247841995 on Build GRCh38
rs7522811 Chr.1: 247841911-247841911 on Build GRCh38
rs75330382 Chr.1: 247841966-247841966 on Build GRCh38
rs367729132 Chr.1: 247841944-247841944 on Build GRCh38
rs376918027 Chr.1: 247841737-247841737 on Build GRCh38
rs573258207 Chr.1: 247842055-247842055 on Build GRCh38
rs536628249 Chr.1: 247841824-247841824 on Build GRCh38
rs201205415 Chr.1: 247841860-247841860 on Build GRCh38
rs756293820 Chr.1: 247841725-247841725 on Build GRCh38
rs777534354 Chr.1: 247841840-247841840 on Build GRCh38
rs201802457 Chr.1: 247841736-247841736 on Build GRCh38
rs752323036 Chr.1: 247841868-247841868 on Build GRCh38
rs369061630 Chr.1: 247841843-247841843 on Build GRCh38
rs766949268 Chr.1: 247841676-247841676 on Build GRCh38
rs200135626 Chr.1: 247841813-247841813 on Build GRCh38
rs763454821 Chr.1: 247841766-247841766 on Build GRCh38
rs779818114 Chr.1: 247841809-247841809 on Build GRCh38
rs546233207 Chr.1: 247842035-247842035 on Build GRCh38
rs576222079 Chr.1: 247841807-247841807 on Build GRCh38
rs202099060 Chr.1: 247841887-247841887 on Build GRCh38
rs1043251526 Chr.1: 247841678-247841678 on Build GRCh38
rs374737953 Chr.1: 247841834-247841834 on Build GRCh38
rs1025076640 Chr.1: 247842004-247842004 on Build GRCh38
rs760857351 Chr.1: 247841869-247841869 on Build GRCh38
rs988115846 Chr.1: 247841994-247841994 on Build GRCh38
rs1017096802 Chr.1: 247842090-247842090 on Build GRCh38
rs752930802 Chr.1: 247841718-247841718 on Build GRCh38
rs762551636 Chr.1: 247841885-247841885 on Build GRCh38
rs754943806 Chr.1: 247841919-247841919 on Build GRCh38
rs773572668 Chr.1: 247841666-247841666 on Build GRCh38
rs367831469 Chr.1: 247841697-247841697 on Build GRCh38
rs778595732 Chr.1: 247841726-247841726 on Build GRCh38
rs953844023 Chr.1: 247841827-247841827 on Build GRCh38
rs766171383 Chr.1: 247841915-247841915 on Build GRCh38
rs371395265 Chr.1: 247841946-247841946 on Build GRCh38
rs570469821 Chr.1: 247841865-247841865 on Build GRCh38
rs202005304 Chr.1: 247841756-247841756 on Build GRCh38
rs1053275887 Chr.1: 247842011-247842011 on Build GRCh38
rs776975762 Chr.1: 247841892-247841892 on Build GRCh38
rs371909713 Chr.1: 247841755-247841755 on Build GRCh38
rs919139122 Chr.1: 247842075-247842075 on Build GRCh38
rs556413423 Chr.1: 247841680-247841680 on Build GRCh38
rs767711381 Chr.1: 247841778-247841778 on Build GRCh38
rs760117271 Chr.1: 247841776-247841776 on Build GRCh38
rs763908015 Chr.1: 247841896-247841896 on Build GRCh38
rs569345550 Chr.1: 247841769-247841769 on Build GRCh38
rs867990793 Chr.1: 247841850-247841850 on Build GRCh38
rs189115009 Chr.1: 247842044-247842044 on Build GRCh38
rs767534351 Chr.1: 247841920-247841920 on Build GRCh38
rs575579497 Chr.1: 247842016-247842016 on Build GRCh38
rs76040612 Chr.1: 247842012-247842012 on Build GRCh38
rs755155451 Chr.1: 247841698-247841698 on Build GRCh38
rs972575296 Chr.1: 247842105-247842105 on Build GRCh38
rs780586103 Chr.1: 247841820-247841820 on Build GRCh38
rs865872393 Chr.1: 247841889-247841889 on Build GRCh38
rs768218744 Chr.1: 247841748-247841748 on Build GRCh38
rs1005195301 Chr.1: 247842077-247842077 on Build GRCh38
rs764416147 Chr.1: 247841799-247841799 on Build GRCh38
rs941868443 Chr.1: 247842051-247842051 on Build GRCh38
rs777146149 Chr.1: 247841945-247841945 on Build GRCh38
rs369772281 Chr.1: 247841757-247841757 on Build GRCh38
rs78126803 Chr.1: 247841742-247841742 on Build GRCh38
rs571959936 Chr.1: 247842054-247842054 on Build GRCh38
rs751302022 Chr.1: 247841918-247841918 on Build GRCh38
rs567908878 Chr.1: 247841770-247841770 on Build GRCh38
rs765527806 Chr.1: 247841657-247841657 on Build GRCh38
rs138609459 Chr.1: 247841738-247841738 on Build GRCh38
rs372016660 Chr.1: 247841707-247841707 on Build GRCh38
rs961118919 Chr.1: 247842095-247842095 on Build GRCh38
rs748937976 Chr.1: 247841842-247841842 on Build GRCh38

View Full Product Details