Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 248812888-248813379 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH3BP5L
Entrez Gene ID
80851
Gene Name
SH3 binding domain protein 5 like
Gene Aliases
-
UniGene
Hs.298573
Gene Chromosome Location
Chr.1: 248810446-248826285 on Build GRCh38
Transcripts
NM_001322462.1 NM_001322463.1 NM_001322464.1 NM_030645.1 XM_017002404.1 XM_017002405.1 XM_017002406.1 XM_017002407.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4030801 c.656T>G p.I219S Chr.1: 248813044-248813044 on Build GRCh38
4030803 c.610G>A p.A204T Chr.1: 248813090-248813090 on Build GRCh38
6440285 c.709G>A p.E237K Chr.1: 248812991-248812991 on Build GRCh38
4397389 c.521C>T p.A174V Chr.1: 248813083-248813083 on Build GRCh38
6440286 c.613G>A p.E205K Chr.1: 248812991-248812991 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1024913491 Chr.1: 248812924-248812924 on Build GRCh38
rs777843948 Chr.1: 248813148-248813148 on Build GRCh38
rs779053402 Chr.1: 248813173-248813173 on Build GRCh38
rs1053777157 Chr.1: 248813309-248813309 on Build GRCh38
rs773269872 Chr.1: 248813032-248813032 on Build GRCh38
rs751280700 Chr.1: 248813135-248813135 on Build GRCh38
rs868194300 Chr.1: 248813028-248813028 on Build GRCh38
rs771245444 Chr.1: 248813170-248813170 on Build GRCh38
rs147597154 Chr.1: 248813054-248813054 on Build GRCh38
rs529944717 Chr.1: 248813248-248813248 on Build GRCh38
rs781745875 Chr.1: 248813079-248813079 on Build GRCh38
rs747460082 Chr.1: 248812992-248812992 on Build GRCh38
rs1009435735 Chr.1: 248812922-248812922 on Build GRCh38
rs1002121217 Chr.1: 248813282-248813282 on Build GRCh38
rs374016845 Chr.1: 248813212-248813212 on Build GRCh38
rs147282536 Chr.1: 248813352-248813352 on Build GRCh38
rs371089082 Chr.1: 248813033-248813033 on Build GRCh38
rs772176517 Chr.1: 248813217-248813217 on Build GRCh38
rs759676523 Chr.1: 248813043-248813043 on Build GRCh38
rs754586080 Chr.1: 248813077-248813077 on Build GRCh38
rs549306552 Chr.1: 248813048-248813048 on Build GRCh38
rs572452753 Chr.1: 248813243-248813243 on Build GRCh38
rs1016292574 Chr.1: 248813301-248813301 on Build GRCh38
rs771182505 Chr.1: 248813097-248813097 on Build GRCh38
rs571643456 Chr.1: 248813195-248813195 on Build GRCh38
rs905146046 Chr.1: 248813253-248813253 on Build GRCh38
rs753202290 Chr.1: 248813147-248813147 on Build GRCh38
rs147847671 Chr.1: 248813053-248813053 on Build GRCh38
rs530277665 Chr.1: 248813344-248813344 on Build GRCh38
rs903497885 Chr.1: 248813196-248813196 on Build GRCh38
rs938049546 Chr.1: 248813256-248813256 on Build GRCh38
rs938116739 Chr.1: 248812914-248812914 on Build GRCh38
rs527837306 Chr.1: 248812921-248812921 on Build GRCh38
rs776685418 Chr.1: 248813104-248813105 on Build GRCh38
rs769283474 Chr.1: 248812994-248812994 on Build GRCh38
rs750218043 Chr.1: 248813125-248813125 on Build GRCh38
rs987576804 Chr.1: 248813152-248813152 on Build GRCh38
rs910718530 Chr.1: 248813322-248813322 on Build GRCh38
rs779618597 Chr.1: 248812967-248812967 on Build GRCh38
rs775559770 Chr.1: 248813198-248813198 on Build GRCh38
rs554983579 Chr.1: 248813126-248813126 on Build GRCh38
rs766314992 Chr.1: 248813037-248813037 on Build GRCh38
rs375608111 Chr.1: 248813083-248813083 on Build GRCh38
rs756993487 Chr.1: 248813052-248813052 on Build GRCh38
rs756633294 Chr.1: 248813151-248813151 on Build GRCh38
rs536791785 Chr.1: 248813310-248813310 on Build GRCh38
rs149049712 Chr.1: 248813105-248813105 on Build GRCh38
rs746487476 Chr.1: 248813071-248813071 on Build GRCh38
rs764880415 Chr.1: 248813113-248813113 on Build GRCh38
rs969256206 Chr.1: 248812930-248812930 on Build GRCh38
rs572905746 Chr.1: 248813172-248813173 on Build GRCh38
rs770217668 Chr.1: 248813030-248813030 on Build GRCh38
rs1042261357 Chr.1: 248813321-248813321 on Build GRCh38
rs116192941 Chr.1: 248813315-248813315 on Build GRCh38
rs573807015 Chr.1: 248813327-248813327 on Build GRCh38
rs192395341 Chr.1: 248813188-248813188 on Build GRCh38
rs777320393 Chr.1: 248813008-248813008 on Build GRCh38
rs538525157 Chr.1: 248813059-248813059 on Build GRCh38
rs892190188 Chr.1: 248813239-248813239 on Build GRCh38
rs767602867 Chr.1: 248813104-248813104 on Build GRCh38
rs773644142 Chr.1: 248813085-248813085 on Build GRCh38
rs759586568 Chr.1: 248813100-248813100 on Build GRCh38
rs778720975 Chr.1: 248812919-248812919 on Build GRCh38
rs377372429 Chr.1: 248813094-248813094 on Build GRCh38
rs754498353 Chr.1: 248813140-248813140 on Build GRCh38
rs890266977 Chr.1: 248813331-248813331 on Build GRCh38
rs780857674 Chr.1: 248813141-248813141 on Build GRCh38
rs774594178 Chr.1: 248813099-248813099 on Build GRCh38
rs982736282 Chr.1: 248813218-248813218 on Build GRCh38
rs748696860 Chr.1: 248813080-248813080 on Build GRCh38
rs11205412 Chr.1: 248813215-248813215 on Build GRCh38
rs945423289 Chr.1: 248813320-248813320 on Build GRCh38
rs963695358 Chr.1: 248813302-248813302 on Build GRCh38
rs972061395 Chr.1: 248813307-248813307 on Build GRCh38
rs745555905 Chr.1: 248812952-248812952 on Build GRCh38
rs757805866 Chr.1: 248812958-248812958 on Build GRCh38
rs774404618 Chr.1: 248813042-248813042 on Build GRCh38
rs753818825 Chr.1: 248813051-248813051 on Build GRCh38
rs778403707 Chr.1: 248812942-248812942 on Build GRCh38
rs748502436 Chr.1: 248813026-248813026 on Build GRCh38
rs554198832 Chr.1: 248813225-248813225 on Build GRCh38
rs749631274 Chr.1: 248813162-248813162 on Build GRCh38
rs772064009 Chr.1: 248813118-248813118 on Build GRCh38

View Full Product Details