Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 5393793-5394289 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TUBAL3
Entrez Gene ID
79861
Gene Name
tubulin alpha like 3
Gene Aliases
-
UniGene
Hs.163079
Gene Chromosome Location
Chr.10: 5393098-5404830 on Build GRCh38
Transcripts
NM_001171864.1 NM_024803.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3727135 c.757G>A p.G253R Chr.10: 5394101-5394101 on Build GRCh38
1702007 c.995C>T p.P332L Chr.10: 5393863-5393863 on Build GRCh38
6129497 c.984G>A p.G328G Chr.10: 5393874-5393874 on Build GRCh38
6814020 c.809C>T p.P270L Chr.10: 5394049-5394049 on Build GRCh38
3981034 c.647T>C p.V216A Chr.10: 5394211-5394211 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376671619 Chr.10: 5394031-5394031 on Build GRCh38
rs377599023 Chr.10: 5394173-5394173 on Build GRCh38
rs782780114 Chr.10: 5394042-5394042 on Build GRCh38
rs148029829 Chr.10: 5393818-5393818 on Build GRCh38
rs781805478 Chr.10: 5394225-5394225 on Build GRCh38
rs537901396 Chr.10: 5394149-5394149 on Build GRCh38
rs947999143 Chr.10: 5393865-5393865 on Build GRCh38
rs782339556 Chr.10: 5394266-5394266 on Build GRCh38
rs375383101 Chr.10: 5393936-5393936 on Build GRCh38
rs782333333 Chr.10: 5393978-5393978 on Build GRCh38
rs369990671 Chr.10: 5393993-5393993 on Build GRCh38
rs782589068 Chr.10: 5394261-5394261 on Build GRCh38
rs927836076 Chr.10: 5394169-5394169 on Build GRCh38
rs76052898 Chr.10: 5393860-5393860 on Build GRCh38
rs1025640290 Chr.10: 5394105-5394105 on Build GRCh38
rs372651229 Chr.10: 5393863-5393863 on Build GRCh38
rs782051685 Chr.10: 5393819-5393819 on Build GRCh38
rs782226532 Chr.10: 5394263-5394263 on Build GRCh38
rs781880312 Chr.10: 5394203-5394203 on Build GRCh38
rs782811947 Chr.10: 5393932-5393932 on Build GRCh38
rs782540943 Chr.10: 5393838-5393838 on Build GRCh38
rs144912794 Chr.10: 5393914-5393914 on Build GRCh38
rs142943186 Chr.10: 5394012-5394012 on Build GRCh38
rs372608934 Chr.10: 5394252-5394252 on Build GRCh38
rs534834682 Chr.10: 5393915-5393915 on Build GRCh38
rs577160496 Chr.10: 5393883-5393883 on Build GRCh38
rs782531228 Chr.10: 5394175-5394175 on Build GRCh38
rs782261479 Chr.10: 5393846-5393846 on Build GRCh38
rs149499697 Chr.10: 5394194-5394194 on Build GRCh38
rs145539086 Chr.10: 5393864-5393864 on Build GRCh38
rs782257792 Chr.10: 5394071-5394071 on Build GRCh38
rs782724522 Chr.10: 5394058-5394058 on Build GRCh38
rs751991687 Chr.10: 5394167-5394167 on Build GRCh38
rs782284577 Chr.10: 5394197-5394197 on Build GRCh38
rs782519938 Chr.10: 5394241-5394241 on Build GRCh38
rs781976987 Chr.10: 5394218-5394218 on Build GRCh38
rs782630053 Chr.10: 5393961-5393961 on Build GRCh38
rs200013500 Chr.10: 5394240-5394240 on Build GRCh38
rs782678357 Chr.10: 5394089-5394089 on Build GRCh38
rs377212178 Chr.10: 5394186-5394186 on Build GRCh38
rs41289265 Chr.10: 5394020-5394020 on Build GRCh38
rs781998041 Chr.10: 5394000-5394000 on Build GRCh38
rs988254391 Chr.10: 5394184-5394184 on Build GRCh38
rs782600093 Chr.10: 5394188-5394188 on Build GRCh38
rs200569900 Chr.10: 5393820-5393820 on Build GRCh38
rs149687567 Chr.10: 5393833-5393833 on Build GRCh38
rs782444461 Chr.10: 5393977-5393977 on Build GRCh38
rs143134074 Chr.10: 5394219-5394219 on Build GRCh38
rs782762172 Chr.10: 5393894-5393894 on Build GRCh38
rs781816034 Chr.10: 5393826-5393826 on Build GRCh38
rs782260699 Chr.10: 5394193-5394193 on Build GRCh38
rs781923152 Chr.10: 5394100-5394100 on Build GRCh38
rs1018515261 Chr.10: 5394048-5394048 on Build GRCh38
rs782754306 Chr.10: 5394171-5394171 on Build GRCh38
rs782428043 Chr.10: 5394016-5394016 on Build GRCh38
rs782328118 Chr.10: 5394098-5394098 on Build GRCh38
rs150848733 Chr.10: 5393835-5393835 on Build GRCh38
rs782339778 Chr.10: 5393996-5393996 on Build GRCh38
rs145537374 Chr.10: 5394139-5394139 on Build GRCh38
rs782444674 Chr.10: 5393940-5393940 on Build GRCh38
rs200240096 Chr.10: 5394046-5394046 on Build GRCh38
rs782814459 Chr.10: 5394121-5394121 on Build GRCh38
rs782525329 Chr.10: 5393827-5393827 on Build GRCh38
rs782611202 Chr.10: 5393845-5393845 on Build GRCh38
rs7097775 Chr.10: 5393955-5393955 on Build GRCh38
rs781971389 Chr.10: 5393873-5393873 on Build GRCh38
rs782598783 Chr.10: 5394066-5394066 on Build GRCh38
rs782698202 Chr.10: 5394021-5394021 on Build GRCh38
rs781968869 Chr.10: 5394208-5394208 on Build GRCh38
rs782498344 Chr.10: 5394081-5394081 on Build GRCh38
rs199531744 Chr.10: 5394212-5394212 on Build GRCh38
rs146670201 Chr.10: 5394126-5394126 on Build GRCh38
rs140777769 Chr.10: 5394049-5394049 on Build GRCh38
rs532114046 Chr.10: 5394182-5394182 on Build GRCh38
rs774325532 Chr.10: 5393861-5393861 on Build GRCh38
rs782085561 Chr.10: 5394035-5394035 on Build GRCh38
rs115615637 Chr.10: 5394134-5394134 on Build GRCh38
rs782232187 Chr.10: 5394108-5394108 on Build GRCh38
rs782078146 Chr.10: 5393888-5393888 on Build GRCh38
rs200952820 Chr.10: 5393832-5393832 on Build GRCh38
rs782292095 Chr.10: 5393967-5393967 on Build GRCh38
rs35744127 Chr.10: 5393841-5393841 on Build GRCh38
rs782139432 Chr.10: 5394033-5394033 on Build GRCh38
rs560849745 Chr.10: 5394109-5394109 on Build GRCh38
rs148559873 Chr.10: 5393957-5393957 on Build GRCh38
rs782789211 Chr.10: 5394222-5394222 on Build GRCh38
rs368148096 Chr.10: 5394267-5394267 on Build GRCh38
rs370241724 Chr.10: 5394185-5394185 on Build GRCh38
rs34080891 Chr.10: 5394110-5394110 on Build GRCh38
rs782308313 Chr.10: 5394207-5394207 on Build GRCh38
rs138109258 Chr.10: 5394163-5394163 on Build GRCh38
rs1016746088 Chr.10: 5394111-5394111 on Build GRCh38
rs781822090 Chr.10: 5394144-5394144 on Build GRCh38
rs139311920 Chr.10: 5393840-5393840 on Build GRCh38
rs782538763 Chr.10: 5394258-5394258 on Build GRCh38
rs782565558 Chr.10: 5393992-5393992 on Build GRCh38
rs782512176 Chr.10: 5394147-5394147 on Build GRCh38
rs772829697 Chr.10: 5393839-5393839 on Build GRCh38
rs782687548 Chr.10: 5394195-5394195 on Build GRCh38
rs148106464 Chr.10: 5394244-5394244 on Build GRCh38
rs113581631 Chr.10: 5393916-5393916 on Build GRCh38
rs782268673 Chr.10: 5394096-5394096 on Build GRCh38
rs782626582 Chr.10: 5393952-5393952 on Build GRCh38

View Full Product Details