Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 6216025-6216531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PFKFB3
Entrez Gene ID
5209
Gene Name
6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 3
Gene Aliases
IPFK2, PFK2, iPFK-2
UniGene
Hs.195471
Gene Chromosome Location
Chr.10: 6144802-6254648 on Build GRCh38
Transcripts
NM_001145443.2 NM_001282630.2 NM_001314063.1 NM_001323016.1 NM_001323017.1 NM_004566.3 NR_136554.1 XM_005252464.1 XM_011519493.1 XM_017016325.1 XM_017016326.1 XM_017016327.1 XM_017016328.1 XM_017016329.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6744332 c.278A>G p.Y93C Chr.10: 6216163-6216163 on Build GRCh38
326126 c.314C>G p.A105G Chr.10: 6216139-6216139 on Build GRCh38
7109527 c.261G>T p.L87F Chr.10: 6216146-6216146 on Build GRCh38
4796172 c.289G>A p.E97K Chr.10: 6216174-6216174 on Build GRCh38
5473030 c.328G>A p.V110I Chr.10: 6216153-6216153 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1012918733 Chr.10: 6216107-6216107 on Build GRCh38
rs776249437 Chr.10: 6216104-6216104 on Build GRCh38
rs775425578 Chr.10: 6216055-6216055 on Build GRCh38
rs78894631 Chr.10: 6216485-6216485 on Build GRCh38
rs200359095 Chr.10: 6216079-6216079 on Build GRCh38
rs1006186286 Chr.10: 6216353-6216353 on Build GRCh38
rs754181310 Chr.10: 6216085-6216085 on Build GRCh38
rs1043413469 Chr.10: 6216464-6216464 on Build GRCh38
rs559720198 Chr.10: 6216272-6216272 on Build GRCh38
rs371395204 Chr.10: 6216236-6216236 on Build GRCh38
rs372840282 Chr.10: 6216073-6216073 on Build GRCh38
rs2516620 Chr.10: 6216260-6216260 on Build GRCh38
rs377231341 Chr.10: 6216075-6216075 on Build GRCh38
rs750805171 Chr.10: 6216170-6216170 on Build GRCh38
rs576378564 Chr.10: 6216088-6216088 on Build GRCh38
rs975220693 Chr.10: 6216145-6216145 on Build GRCh38
rs191208974 Chr.10: 6216051-6216051 on Build GRCh38
rs1057086965 Chr.10: 6216366-6216366 on Build GRCh38
rs968320753 Chr.10: 6216486-6216486 on Build GRCh38
rs548405989 Chr.10: 6216361-6216361 on Build GRCh38
rs926252759 Chr.10: 6216465-6216465 on Build GRCh38
rs963949213 Chr.10: 6216396-6216396 on Build GRCh38
rs954282148 Chr.10: 6216474-6216474 on Build GRCh38
rs2148293 Chr.10: 6216052-6216052 on Build GRCh38
rs1017520112 Chr.10: 6216367-6216367 on Build GRCh38
rs569707437 Chr.10: 6216385-6216385 on Build GRCh38
rs757197649 Chr.10: 6216191-6216191 on Build GRCh38
rs975271796 Chr.10: 6216430-6216430 on Build GRCh38
rs753980509 Chr.10: 6216156-6216156 on Build GRCh38
rs756200144 Chr.10: 6216080-6216080 on Build GRCh38
rs748517372 Chr.10: 6216212-6216212 on Build GRCh38
rs35710132 Chr.10: 6216053-6216053 on Build GRCh38
rs111352523 Chr.10: 6216124-6216124 on Build GRCh38
rs112006058 Chr.10: 6216063-6216063 on Build GRCh38
rs747900698 Chr.10: 6216090-6216090 on Build GRCh38
rs759306510 Chr.10: 6216280-6216280 on Build GRCh38
rs745410022 Chr.10: 6216202-6216202 on Build GRCh38
rs576133477 Chr.10: 6216056-6216056 on Build GRCh38
rs978878730 Chr.10: 6216316-6216316 on Build GRCh38
rs766680450 Chr.10: 6216082-6216082 on Build GRCh38
rs74114570 Chr.10: 6216251-6216251 on Build GRCh38
rs765641319 Chr.10: 6216157-6216157 on Build GRCh38
rs745463544 Chr.10: 6216105-6216105 on Build GRCh38
rs998759558 Chr.10: 6216462-6216462 on Build GRCh38
rs985648493 Chr.10: 6216483-6216483 on Build GRCh38
rs2516623 Chr.10: 6216348-6216348 on Build GRCh38
rs762111869 Chr.10: 6216114-6216114 on Build GRCh38
rs779626957 Chr.10: 6216211-6216211 on Build GRCh38
rs988377130 Chr.10: 6216453-6216453 on Build GRCh38
rs542439439 Chr.10: 6216134-6216134 on Build GRCh38
rs769392153 Chr.10: 6216210-6216210 on Build GRCh38
rs765399775 Chr.10: 6216169-6216169 on Build GRCh38
rs369059195 Chr.10: 6216091-6216091 on Build GRCh38
rs2516622 Chr.10: 6216337-6216337 on Build GRCh38
rs954418685 Chr.10: 6216252-6216252 on Build GRCh38
rs2516621 Chr.10: 6216283-6216283 on Build GRCh38
rs769357876 Chr.10: 6216078-6216089 on Build GRCh38
rs576445073 Chr.10: 6216457-6216457 on Build GRCh38
rs766625430 Chr.10: 6216141-6216141 on Build GRCh38
rs537150412 Chr.10: 6216403-6216403 on Build GRCh38
rs955257933 Chr.10: 6216452-6216452 on Build GRCh38
rs994055849 Chr.10: 6216288-6216288 on Build GRCh38
rs760834564 Chr.10: 6216139-6216139 on Build GRCh38
rs769437140 Chr.10: 6216106-6216106 on Build GRCh38
rs111691148 Chr.10: 6216053-6216054 on Build GRCh38
rs910332125 Chr.10: 6216266-6216266 on Build GRCh38
rs576782752 Chr.10: 6216429-6216430 on Build GRCh38
rs115959316 Chr.10: 6216092-6216092 on Build GRCh38
rs771806677 Chr.10: 6216285-6216285 on Build GRCh38
rs775179934 Chr.10: 6216397-6216397 on Build GRCh38
rs2251469 Chr.10: 6216470-6216470 on Build GRCh38
rs1005643690 Chr.10: 6216444-6216444 on Build GRCh38
rs765466995 Chr.10: 6216232-6216232 on Build GRCh38
rs1016062684 Chr.10: 6216450-6216450 on Build GRCh38
rs373022015 Chr.10: 6216217-6216217 on Build GRCh38
rs759737654 Chr.10: 6216230-6216230 on Build GRCh38
rs763367903 Chr.10: 6216174-6216174 on Build GRCh38
rs774793354 Chr.10: 6216080-6216080 on Build GRCh38
rs552664159 Chr.10: 6216243-6216243 on Build GRCh38
rs774741824 Chr.10: 6216108-6216108 on Build GRCh38
rs558199624 Chr.10: 6216407-6216407 on Build GRCh38
rs546999821 Chr.10: 6216271-6216271 on Build GRCh38
rs1002415846 Chr.10: 6216269-6216269 on Build GRCh38
rs528468769 Chr.10: 6216267-6216267 on Build GRCh38
rs1056743834 Chr.10: 6216478-6216479 on Build GRCh38
rs773635607 Chr.10: 6216136-6216136 on Build GRCh38
rs185998775 Chr.10: 6216050-6216050 on Build GRCh38
rs776813160 Chr.10: 6216226-6216226 on Build GRCh38
rs888428027 Chr.10: 6216428-6216428 on Build GRCh38
rs769070410 Chr.10: 6216047-6216047 on Build GRCh38
rs1052219587 Chr.10: 6216206-6216206 on Build GRCh38
rs371347480 Chr.10: 6216060-6216060 on Build GRCh38
rs961077157 Chr.10: 6216067-6216067 on Build GRCh38
rs369478234 Chr.10: 6216213-6216213 on Build GRCh38
rs376785737 Chr.10: 6216218-6216218 on Build GRCh38
rs780609733 Chr.10: 6216197-6216197 on Build GRCh38
rs34276031 Chr.10: 6216077-6216077 on Build GRCh38
rs748109191 Chr.10: 6216470-6216472 on Build GRCh38
rs978568063 Chr.10: 6216507-6216507 on Build GRCh38
rs375498227 Chr.10: 6216101-6216101 on Build GRCh38
rs922497746 Chr.10: 6216442-6216449 on Build GRCh38
rs544663576 Chr.10: 6216102-6216102 on Build GRCh38
rs985743078 Chr.10: 6216264-6216264 on Build GRCh38
rs2148294 Chr.10: 6216089-6216089 on Build GRCh38
rs770569919 Chr.10: 6216081-6216081 on Build GRCh38
rs561268096 Chr.10: 6216460-6216460 on Build GRCh38

View Full Product Details