Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 7563038-7563537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITIH5
Entrez Gene ID
80760
Gene Name
inter-alpha-trypsin inhibitor heavy chain family member 5
Gene Aliases
ITI-HC5, PP14776
UniGene
Hs.498586
Gene Chromosome Location
Chr.10: 7559270-7666998 on Build GRCh38
Transcripts
NM_030569.6 NM_032817.5 XM_011519713.2

SNP Detail

SNP ID Chromosome Location
rs374656410 Chr.10: 7563398-7563398 on Build GRCh38
rs1009067126 Chr.10: 7563145-7563145 on Build GRCh38
rs138658798 Chr.10: 7563280-7563280 on Build GRCh38
rs771365875 Chr.10: 7563188-7563188 on Build GRCh38
rs766313151 Chr.10: 7563295-7563298 on Build GRCh38
rs772162750 Chr.10: 7563432-7563432 on Build GRCh38
rs778436441 Chr.10: 7563353-7563353 on Build GRCh38
rs953766760 Chr.10: 7563159-7563159 on Build GRCh38
rs767819986 Chr.10: 7563077-7563077 on Build GRCh38
rs1028132377 Chr.10: 7563315-7563315 on Build GRCh38
rs371983728 Chr.10: 7563382-7563382 on Build GRCh38
rs781406958 Chr.10: 7563296-7563296 on Build GRCh38
rs753375922 Chr.10: 7563349-7563349 on Build GRCh38
rs750537149 Chr.10: 7563085-7563086 on Build GRCh38
rs10795551 Chr.10: 7563138-7563138 on Build GRCh38
rs370505197 Chr.10: 7563434-7563434 on Build GRCh38
rs149543607 Chr.10: 7563212-7563212 on Build GRCh38
rs373788153 Chr.10: 7563131-7563131 on Build GRCh38
rs768712021 Chr.10: 7563412-7563412 on Build GRCh38
rs746207276 Chr.10: 7563388-7563388 on Build GRCh38
rs761618755 Chr.10: 7563220-7563220 on Build GRCh38
rs746436027 Chr.10: 7563297-7563297 on Build GRCh38
rs750171430 Chr.10: 7563334-7563334 on Build GRCh38
rs745730124 Chr.10: 7563069-7563069 on Build GRCh38
rs113706314 Chr.10: 7563085-7563085 on Build GRCh38
rs779597051 Chr.10: 7563272-7563272 on Build GRCh38
rs977810057 Chr.10: 7563407-7563407 on Build GRCh38
rs982512423 Chr.10: 7563271-7563271 on Build GRCh38
rs374247037 Chr.10: 7563326-7563326 on Build GRCh38
rs769874866 Chr.10: 7563247-7563247 on Build GRCh38
rs1047971291 Chr.10: 7563341-7563341 on Build GRCh38
rs781142532 Chr.10: 7563162-7563162 on Build GRCh38
rs865835899 Chr.10: 7563180-7563180 on Build GRCh38
rs398102295 Chr.10: 7563116-7563116 on Build GRCh38
rs747349397 Chr.10: 7563292-7563292 on Build GRCh38
rs367911074 Chr.10: 7563433-7563433 on Build GRCh38
rs368073507 Chr.10: 7563374-7563374 on Build GRCh38
rs550819053 Chr.10: 7563488-7563488 on Build GRCh38
rs10737022 Chr.10: 7563494-7563494 on Build GRCh38
rs143775699 Chr.10: 7563164-7563164 on Build GRCh38
rs767367042 Chr.10: 7563221-7563221 on Build GRCh38
rs770406264 Chr.10: 7563299-7563299 on Build GRCh38
rs760653731 Chr.10: 7563342-7563342 on Build GRCh38
rs3824658 Chr.10: 7563105-7563105 on Build GRCh38
rs770056084 Chr.10: 7563418-7563418 on Build GRCh38
rs1047442457 Chr.10: 7563119-7563119 on Build GRCh38
rs767409400 Chr.10: 7563332-7563332 on Build GRCh38
rs751349610 Chr.10: 7563149-7563149 on Build GRCh38
rs774287532 Chr.10: 7563414-7563414 on Build GRCh38
rs193920741 Chr.10: 7563367-7563367 on Build GRCh38
rs778586237 Chr.10: 7563281-7563281 on Build GRCh38
rs150365694 Chr.10: 7563351-7563351 on Build GRCh38
rs529180522 Chr.10: 7563470-7563470 on Build GRCh38
rs368675896 Chr.10: 7563355-7563355 on Build GRCh38
rs948760624 Chr.10: 7563222-7563222 on Build GRCh38
rs757973663 Chr.10: 7563371-7563371 on Build GRCh38
rs995064325 Chr.10: 7563116-7563116 on Build GRCh38
rs149848698 Chr.10: 7563100-7563100 on Build GRCh38
rs751345630 Chr.10: 7563219-7563219 on Build GRCh38
rs938075780 Chr.10: 7563284-7563284 on Build GRCh38
rs755560452 Chr.10: 7563163-7563163 on Build GRCh38
rs61731872 Chr.10: 7563300-7563300 on Build GRCh38
rs769019728 Chr.10: 7563304-7563304 on Build GRCh38
rs1020191547 Chr.10: 7563509-7563509 on Build GRCh38
rs768283912 Chr.10: 7563178-7563178 on Build GRCh38
rs137874246 Chr.10: 7563183-7563183 on Build GRCh38
rs766271604 Chr.10: 7563346-7563346 on Build GRCh38
rs553296913 Chr.10: 7563106-7563106 on Build GRCh38
rs763097885 Chr.10: 7563311-7563311 on Build GRCh38
rs770066744 Chr.10: 7563209-7563209 on Build GRCh38
rs774572211 Chr.10: 7563321-7563321 on Build GRCh38
rs1048391365 Chr.10: 7563376-7563376 on Build GRCh38
rs776612261 Chr.10: 7563122-7563122 on Build GRCh38
rs777139283 Chr.10: 7563192-7563192 on Build GRCh38
rs750304119 Chr.10: 7563231-7563231 on Build GRCh38
rs369184382 Chr.10: 7563431-7563431 on Build GRCh38
rs181306299 Chr.10: 7563216-7563216 on Build GRCh38
rs769360539 Chr.10: 7563066-7563066 on Build GRCh38
rs372816798 Chr.10: 7563179-7563179 on Build GRCh38
rs140047376 Chr.10: 7563087-7563087 on Build GRCh38
rs566798696 Chr.10: 7563504-7563504 on Build GRCh38
rs759540997 Chr.10: 7563197-7563197 on Build GRCh38
rs543699458 Chr.10: 7563290-7563290 on Build GRCh38
rs757184944 Chr.10: 7563155-7563155 on Build GRCh38
rs907204199 Chr.10: 7563092-7563092 on Build GRCh38
rs188458658 Chr.10: 7563140-7563140 on Build GRCh38
rs147579573 Chr.10: 7563170-7563170 on Build GRCh38
rs759614175 Chr.10: 7563125-7563125 on Build GRCh38
rs765571107 Chr.10: 7563129-7563129 on Build GRCh38
rs780661368 Chr.10: 7563408-7563408 on Build GRCh38
rs749390531 Chr.10: 7563410-7563410 on Build GRCh38

View Full Product Details