Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 11742244-11742751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECHDC3
Entrez Gene ID
79746
Gene Name
enoyl-CoA hydratase domain containing 3
Gene Aliases
-
UniGene
Hs.22242
Gene Chromosome Location
Chr.10: 11742357-11764070 on Build GRCh38
Transcripts
NM_024693.4 XM_011519689.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4966737 c.162C>T p.D54D Chr.10: 11742738-11742738 on Build GRCh38
5826615 c.58C>T p.R20C Chr.10: 11742634-11742634 on Build GRCh38
4656496 c.25G>A p.A9T Chr.10: 11742601-11742601 on Build GRCh38
1321513 c.58C>T p.R20C Chr.10: 11742634-11742634 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs922836663 Chr.10: 11742575-11742575 on Build GRCh38
rs928714364 Chr.10: 11742572-11742572 on Build GRCh38
rs991094607 Chr.10: 11742584-11742584 on Build GRCh38
rs544949429 Chr.10: 11742494-11742494 on Build GRCh38
rs902168805 Chr.10: 11742454-11742454 on Build GRCh38
rs1056415970 Chr.10: 11742683-11742683 on Build GRCh38
rs748653173 Chr.10: 11742660-11742660 on Build GRCh38
rs1015507881 Chr.10: 11742459-11742460 on Build GRCh38
rs575825221 Chr.10: 11742489-11742489 on Build GRCh38
rs985597442 Chr.10: 11742585-11742585 on Build GRCh38
rs961343355 Chr.10: 11742460-11742469 on Build GRCh38
rs11558855 Chr.10: 11742634-11742634 on Build GRCh38
rs936301846 Chr.10: 11742361-11742361 on Build GRCh38
rs554911891 Chr.10: 11742417-11742417 on Build GRCh38
rs866770710 Chr.10: 11742321-11742321 on Build GRCh38
rs951375576 Chr.10: 11742533-11742534 on Build GRCh38
rs1042090913 Chr.10: 11742351-11742351 on Build GRCh38
rs76272655 Chr.10: 11742481-11742481 on Build GRCh38
rs946817310 Chr.10: 11742606-11742606 on Build GRCh38
rs560916612 Chr.10: 11742710-11742710 on Build GRCh38
rs768047715 Chr.10: 11742390-11742390 on Build GRCh38
rs946627433 Chr.10: 11742439-11742439 on Build GRCh38
rs866046731 Chr.10: 11742674-11742674 on Build GRCh38
rs973032398 Chr.10: 11742550-11742550 on Build GRCh38
rs1006631076 Chr.10: 11742684-11742685 on Build GRCh38
rs961991173 Chr.10: 11742546-11742546 on Build GRCh38
rs935318431 Chr.10: 11742651-11742651 on Build GRCh38
rs749677462 Chr.10: 11742628-11742628 on Build GRCh38
rs866458082 Chr.10: 11742290-11742290 on Build GRCh38
rs924937775 Chr.10: 11742356-11742356 on Build GRCh38
rs898685781 Chr.10: 11742705-11742705 on Build GRCh38
rs981407523 Chr.10: 11742549-11742549 on Build GRCh38
rs1005939606 Chr.10: 11742518-11742518 on Build GRCh38
rs1050743352 Chr.10: 11742666-11742666 on Build GRCh38
rs779219240 Chr.10: 11742644-11742644 on Build GRCh38
rs1025880279 Chr.10: 11742528-11742528 on Build GRCh38
rs529785223 Chr.10: 11742721-11742721 on Build GRCh38
rs534715744 Chr.10: 11742401-11742401 on Build GRCh38
rs574861452 Chr.10: 11742427-11742427 on Build GRCh38
rs540691785 Chr.10: 11742699-11742699 on Build GRCh38
rs751079767 Chr.10: 11742592-11742592 on Build GRCh38
rs1039270522 Chr.10: 11742376-11742376 on Build GRCh38
rs566327905 Chr.10: 11742396-11742396 on Build GRCh38
rs564839457 Chr.10: 11742522-11742522 on Build GRCh38
rs896398554 Chr.10: 11742694-11742694 on Build GRCh38
rs890907207 Chr.10: 11742675-11742675 on Build GRCh38
rs939190099 Chr.10: 11742267-11742267 on Build GRCh38
rs373050915 Chr.10: 11742622-11742622 on Build GRCh38
rs916525106 Chr.10: 11742429-11742429 on Build GRCh38
rs376112463 Chr.10: 11742281-11742281 on Build GRCh38
rs951777221 Chr.10: 11742516-11742516 on Build GRCh38
rs763566323 Chr.10: 11742586-11742586 on Build GRCh38
rs758329858 Chr.10: 11742578-11742578 on Build GRCh38
rs543879294 Chr.10: 11742442-11742442 on Build GRCh38
rs756911422 Chr.10: 11742599-11742599 on Build GRCh38
rs752614044 Chr.10: 11742563-11742563 on Build GRCh38
rs760484004 Chr.10: 11742695-11742695 on Build GRCh38
rs2763202 Chr.10: 11742341-11742341 on Build GRCh38

View Full Product Details