Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 13161447-13161946 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCM10
Entrez Gene ID
55388
Gene Name
minichromosome maintenance 10 replication initiation factor
Gene Aliases
CNA43, DNA43, PRO2249
UniGene
Hs.198363
Gene Chromosome Location
Chr.10: 13161554-13211110 on Build GRCh38
Transcripts
NM_018518.4 NM_182751.2

SNP Detail

SNP ID Chromosome Location
rs771981418 Chr.10: 13161643-13161643 on Build GRCh38
rs1047800801 Chr.10: 13161692-13161692 on Build GRCh38
rs900430812 Chr.10: 13161541-13161542 on Build GRCh38
rs927134564 Chr.10: 13161852-13161852 on Build GRCh38
rs1039253700 Chr.10: 13161540-13161540 on Build GRCh38
rs1016509504 Chr.10: 13161531-13161531 on Build GRCh38
rs200895482 Chr.10: 13161892-13161892 on Build GRCh38
rs12416029 Chr.10: 13161576-13161576 on Build GRCh38
rs954415196 Chr.10: 13161553-13161553 on Build GRCh38
rs147704803 Chr.10: 13161663-13161663 on Build GRCh38
rs60738279 Chr.10: 13161915-13161918 on Build GRCh38
rs142646485 Chr.10: 13161892-13161903 on Build GRCh38
rs937275791 Chr.10: 13161628-13161628 on Build GRCh38
rs925990924 Chr.10: 13161623-13161623 on Build GRCh38
rs996443700 Chr.10: 13161790-13161790 on Build GRCh38
rs1048999181 Chr.10: 13161751-13161751 on Build GRCh38
rs529257432 Chr.10: 13161820-13161820 on Build GRCh38
rs67050062 Chr.10: 13161892-13161895 on Build GRCh38
rs1023129885 Chr.10: 13161828-13161828 on Build GRCh38
rs1007679529 Chr.10: 13161735-13161764 on Build GRCh38
rs960507802 Chr.10: 13161605-13161605 on Build GRCh38
rs759194218 Chr.10: 13161900-13161903 on Build GRCh38
rs373636165 Chr.10: 13161537-13161537 on Build GRCh38
rs542669194 Chr.10: 13161821-13161821 on Build GRCh38
rs974386888 Chr.10: 13161542-13161542 on Build GRCh38
rs184833678 Chr.10: 13161711-13161711 on Build GRCh38
rs949434975 Chr.10: 13161681-13161681 on Build GRCh38
rs979034152 Chr.10: 13161615-13161615 on Build GRCh38
rs1055022043 Chr.10: 13161632-13161632 on Build GRCh38
rs556356033 Chr.10: 13161546-13161546 on Build GRCh38
rs993025084 Chr.10: 13161608-13161608 on Build GRCh38
rs187690999 Chr.10: 13161834-13161834 on Build GRCh38
rs74122549 Chr.10: 13161903-13161903 on Build GRCh38
rs571969909 Chr.10: 13161698-13161698 on Build GRCh38
rs112666276 Chr.10: 13161627-13161627 on Build GRCh38
rs140032614 Chr.10: 13161908-13161908 on Build GRCh38
rs900296547 Chr.10: 13161714-13161714 on Build GRCh38
rs896553033 Chr.10: 13161474-13161474 on Build GRCh38
rs77359387 Chr.10: 13161831-13161831 on Build GRCh38
rs988567448 Chr.10: 13161585-13161585 on Build GRCh38
rs947236549 Chr.10: 13161612-13161612 on Build GRCh38
rs954364137 Chr.10: 13161802-13161802 on Build GRCh38
rs749481103 Chr.10: 13161585-13161587 on Build GRCh38
rs1027323920 Chr.10: 13161551-13161551 on Build GRCh38
rs1026410004 Chr.10: 13161494-13161494 on Build GRCh38
rs956467886 Chr.10: 13161891-13161892 on Build GRCh38
rs1004534205 Chr.10: 13161784-13161784 on Build GRCh38
rs748975481 Chr.10: 13161480-13161480 on Build GRCh38
rs978980960 Chr.10: 13161618-13161618 on Build GRCh38
rs908272191 Chr.10: 13161572-13161572 on Build GRCh38
rs527469651 Chr.10: 13161510-13161510 on Build GRCh38
rs1006840959 Chr.10: 13161529-13161529 on Build GRCh38
rs1053203711 Chr.10: 13161490-13161490 on Build GRCh38
rs576146978 Chr.10: 13161598-13161598 on Build GRCh38
rs958617216 Chr.10: 13161883-13161883 on Build GRCh38
rs144730289 Chr.10: 13161907-13161907 on Build GRCh38
rs1006376943 Chr.10: 13161728-13161728 on Build GRCh38
rs1033209152 Chr.10: 13161844-13161844 on Build GRCh38
rs1029968144 Chr.10: 13161793-13161793 on Build GRCh38
rs1038301166 Chr.10: 13161770-13161770 on Build GRCh38
rs1535857 Chr.10: 13161772-13161772 on Build GRCh38
rs912001270 Chr.10: 13161649-13161650 on Build GRCh38
rs150151707 Chr.10: 13161911-13161911 on Build GRCh38
rs57090063 Chr.10: 13161907-13161918 on Build GRCh38
rs944538989 Chr.10: 13161511-13161511 on Build GRCh38
rs913027962 Chr.10: 13161599-13161599 on Build GRCh38

View Full Product Details