Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 14519911-14520420 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM107B
Entrez Gene ID
83641
Gene Name
family with sequence similarity 107 member B
Gene Aliases
C10orf45
UniGene
Hs.446315
Gene Chromosome Location
Chr.10: 14518557-14774897 on Build GRCh38
Transcripts
NM_001282695.1 NM_001282696.1 NM_001282697.1 NM_001282698.1 NM_001282699.1 NM_001282700.1 NM_001282701.1 NM_001282702.1 NM_001282703.1 NM_001320735.1 NM_001320736.1 NM_001320737.1 NM_001320738.1 NM_001320739.1 NM_001320740.1 NM_001320741.1 NM_031453.3 XM_005252616.3 XM_011519735.1 XM_017016746.1 XM_017016747.1 XM_017016748.1 XM_017016749.1

SNP Detail

SNP ID Chromosome Location
rs1021044606 Chr.10: 14520283-14520283 on Build GRCh38
rs917718050 Chr.10: 14520103-14520103 on Build GRCh38
rs544554283 Chr.10: 14520094-14520094 on Build GRCh38
rs759100767 Chr.10: 14520020-14520020 on Build GRCh38
rs183967595 Chr.10: 14520353-14520353 on Build GRCh38
rs926353664 Chr.10: 14520029-14520029 on Build GRCh38
rs941692024 Chr.10: 14520175-14520175 on Build GRCh38
rs74772433 Chr.10: 14519975-14519975 on Build GRCh38
rs1049939227 Chr.10: 14520248-14520248 on Build GRCh38
rs979720492 Chr.10: 14520305-14520305 on Build GRCh38
rs914100567 Chr.10: 14520287-14520287 on Build GRCh38
rs897652196 Chr.10: 14520139-14520139 on Build GRCh38
rs925180223 Chr.10: 14520076-14520076 on Build GRCh38
rs1018758951 Chr.10: 14520110-14520110 on Build GRCh38
rs945519698 Chr.10: 14520289-14520289 on Build GRCh38
rs529236889 Chr.10: 14520218-14520218 on Build GRCh38
rs113186348 Chr.10: 14520068-14520069 on Build GRCh38
rs775936930 Chr.10: 14519992-14519992 on Build GRCh38
rs540063731 Chr.10: 14520194-14520194 on Build GRCh38
rs993411678 Chr.10: 14520162-14520162 on Build GRCh38
rs746786800 Chr.10: 14520031-14520031 on Build GRCh38
rs72784927 Chr.10: 14520202-14520202 on Build GRCh38
rs373857407 Chr.10: 14520008-14520008 on Build GRCh38
rs557843253 Chr.10: 14520134-14520134 on Build GRCh38
rs191312494 Chr.10: 14520373-14520373 on Build GRCh38
rs750408745 Chr.10: 14520388-14520388 on Build GRCh38
rs949175127 Chr.10: 14520109-14520109 on Build GRCh38
rs992299026 Chr.10: 14520228-14520228 on Build GRCh38
rs907587630 Chr.10: 14520166-14520166 on Build GRCh38
rs775279915 Chr.10: 14520044-14520044 on Build GRCh38
rs534245582 Chr.10: 14520094-14520094 on Build GRCh38
rs1033860847 Chr.10: 14520210-14520210 on Build GRCh38
rs990669868 Chr.10: 14520085-14520085 on Build GRCh38
rs571331521 Chr.10: 14519937-14519937 on Build GRCh38
rs577685799 Chr.10: 14520150-14520150 on Build GRCh38
rs949116013 Chr.10: 14519959-14519959 on Build GRCh38
rs1000874335 Chr.10: 14520313-14520313 on Build GRCh38
rs925484408 Chr.10: 14520326-14520326 on Build GRCh38
rs902547609 Chr.10: 14520294-14520294 on Build GRCh38
rs550136029 Chr.10: 14520037-14520037 on Build GRCh38
rs575551726 Chr.10: 14520059-14520059 on Build GRCh38
rs375761538 Chr.10: 14520058-14520058 on Build GRCh38
rs536249568 Chr.10: 14520121-14520121 on Build GRCh38
rs1008702250 Chr.10: 14520055-14520055 on Build GRCh38
rs920291069 Chr.10: 14519978-14519978 on Build GRCh38
rs540061038 Chr.10: 14519950-14519950 on Build GRCh38
rs979380419 Chr.10: 14520073-14520073 on Build GRCh38
rs966684301 Chr.10: 14520064-14520064 on Build GRCh38
rs148860744 Chr.10: 14519954-14519954 on Build GRCh38
rs542562880 Chr.10: 14520253-14520253 on Build GRCh38
rs1028871114 Chr.10: 14520041-14520041 on Build GRCh38
rs1008336880 Chr.10: 14520279-14520279 on Build GRCh38
rs1008690026 Chr.10: 14519965-14519965 on Build GRCh38
rs769317618 Chr.10: 14520026-14520026 on Build GRCh38
rs188870107 Chr.10: 14520372-14520372 on Build GRCh38
rs891278183 Chr.10: 14520266-14520266 on Build GRCh38
rs80031831 Chr.10: 14520384-14520384 on Build GRCh38
rs1038587553 Chr.10: 14520017-14520017 on Build GRCh38
rs551513965 Chr.10: 14519935-14519935 on Build GRCh38
rs41284444 Chr.10: 14519958-14519958 on Build GRCh38
rs959515442 Chr.10: 14520080-14520080 on Build GRCh38
rs11542181 Chr.10: 14519976-14519976 on Build GRCh38

View Full Product Details