Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 14520318-14520831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM107B
Entrez Gene ID
83641
Gene Name
family with sequence similarity 107 member B
Gene Aliases
C10orf45
UniGene
Hs.446315
Gene Chromosome Location
Chr.10: 14518557-14774897 on Build GRCh38
Transcripts
NM_001282695.1 NM_001282696.1 NM_001282697.1 NM_001282698.1 NM_001282699.1 NM_001282700.1 NM_001282701.1 NM_001282702.1 NM_001282703.1 NM_001320735.1 NM_001320736.1 NM_001320737.1 NM_001320738.1 NM_001320739.1 NM_001320740.1 NM_001320741.1 NM_031453.3 XM_005252616.3 XM_011519735.1 XM_017016746.1 XM_017016747.1 XM_017016748.1 XM_017016749.1

SNP Detail

SNP ID Chromosome Location
rs183967595 Chr.10: 14520353-14520353 on Build GRCh38
rs755274111 Chr.10: 14520592-14520592 on Build GRCh38
rs912600155 Chr.10: 14520664-14520664 on Build GRCh38
rs146424194 Chr.10: 14520450-14520450 on Build GRCh38
rs79140130 Chr.10: 14520493-14520493 on Build GRCh38
rs921642360 Chr.10: 14520776-14520776 on Build GRCh38
rs1033376107 Chr.10: 14520479-14520479 on Build GRCh38
rs11259151 Chr.10: 14520794-14520794 on Build GRCh38
rs1050416144 Chr.10: 14520595-14520595 on Build GRCh38
rs756287766 Chr.10: 14520404-14520404 on Build GRCh38
rs577747280 Chr.10: 14520640-14520640 on Build GRCh38
rs746804318 Chr.10: 14520642-14520642 on Build GRCh38
rs1020713742 Chr.10: 14520519-14520519 on Build GRCh38
rs970948559 Chr.10: 14520441-14520441 on Build GRCh38
rs111575617 Chr.10: 14520784-14520784 on Build GRCh38
rs983251667 Chr.10: 14520723-14520723 on Build GRCh38
rs140702051 Chr.10: 14520634-14520634 on Build GRCh38
rs770935541 Chr.10: 14520672-14520672 on Build GRCh38
rs918637976 Chr.10: 14520732-14520732 on Build GRCh38
rs576265339 Chr.10: 14520798-14520798 on Build GRCh38
rs553804437 Chr.10: 14520704-14520704 on Build GRCh38
rs1037082307 Chr.10: 14520698-14520698 on Build GRCh38
rs973062155 Chr.10: 14520485-14520485 on Build GRCh38
rs960638609 Chr.10: 14520490-14520490 on Build GRCh38
rs540318580 Chr.10: 14520703-14520703 on Build GRCh38
rs145197329 Chr.10: 14520418-14520418 on Build GRCh38
rs979558927 Chr.10: 14520553-14520553 on Build GRCh38
rs896191590 Chr.10: 14520772-14520772 on Build GRCh38
rs547256992 Chr.10: 14520425-14520425 on Build GRCh38
rs191312494 Chr.10: 14520373-14520373 on Build GRCh38
rs750408745 Chr.10: 14520388-14520388 on Build GRCh38
rs932948981 Chr.10: 14520498-14520498 on Build GRCh38
rs993254442 Chr.10: 14520423-14520423 on Build GRCh38
rs944193485 Chr.10: 14520622-14520622 on Build GRCh38
rs1014688387 Chr.10: 14520739-14520739 on Build GRCh38
rs780112949 Chr.10: 14520454-14520454 on Build GRCh38
rs1000926079 Chr.10: 14520643-14520643 on Build GRCh38
rs1055443788 Chr.10: 14520768-14520768 on Build GRCh38
rs780843520 Chr.10: 14520694-14520694 on Build GRCh38
rs777672293 Chr.10: 14520617-14520617 on Build GRCh38
rs966867968 Chr.10: 14520525-14520525 on Build GRCh38
rs537868662 Chr.10: 14520527-14520527 on Build GRCh38
rs1032356135 Chr.10: 14520665-14520665 on Build GRCh38
rs182171487 Chr.10: 14520740-14520740 on Build GRCh38
rs377141261 Chr.10: 14520459-14520459 on Build GRCh38
rs73599361 Chr.10: 14520399-14520399 on Build GRCh38
rs912811645 Chr.10: 14520805-14520805 on Build GRCh38
rs748723196 Chr.10: 14520752-14520752 on Build GRCh38
rs756862549 Chr.10: 14520797-14520798 on Build GRCh38
rs188870107 Chr.10: 14520372-14520372 on Build GRCh38
rs538931394 Chr.10: 14520468-14520468 on Build GRCh38
rs80031831 Chr.10: 14520384-14520384 on Build GRCh38
rs895124617 Chr.10: 14520671-14520671 on Build GRCh38
rs970637530 Chr.10: 14520712-14520712 on Build GRCh38

View Full Product Details