Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 15073318-15073821 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OLAH
Entrez Gene ID
55301
Gene Name
oleoyl-ACP hydrolase
Gene Aliases
AURA1, SAST, THEDC1
UniGene
Hs.24309
Gene Chromosome Location
Chr.10: 15043896-15073852 on Build GRCh38
Transcripts
NM_001039702.2 NM_018324.2 XM_017016376.1

SNP Detail

SNP ID Chromosome Location
rs539096868 Chr.10: 15073456-15073456 on Build GRCh38
rs967625023 Chr.10: 15073614-15073614 on Build GRCh38
rs1002112386 Chr.10: 15073783-15073783 on Build GRCh38
rs202218155 Chr.10: 15073490-15073490 on Build GRCh38
rs924199045 Chr.10: 15073410-15073410 on Build GRCh38
rs780452199 Chr.10: 15073471-15073471 on Build GRCh38
rs923467368 Chr.10: 15073638-15073638 on Build GRCh38
rs889314512 Chr.10: 15073457-15073457 on Build GRCh38
rs1031204672 Chr.10: 15073487-15073487 on Build GRCh38
rs572470516 Chr.10: 15073478-15073478 on Build GRCh38
rs1040570580 Chr.10: 15073546-15073546 on Build GRCh38
rs538417835 Chr.10: 15073443-15073443 on Build GRCh38
rs1022139506 Chr.10: 15073570-15073570 on Build GRCh38
rs969432337 Chr.10: 15073418-15073418 on Build GRCh38
rs555332312 Chr.10: 15073772-15073772 on Build GRCh38
rs11259458 Chr.10: 15073701-15073701 on Build GRCh38
rs71485544 Chr.10: 15073463-15073463 on Build GRCh38
rs369902691 Chr.10: 15073485-15073485 on Build GRCh38
rs557980742 Chr.10: 15073528-15073528 on Build GRCh38
rs4748140 Chr.10: 15073720-15073720 on Build GRCh38
rs960816773 Chr.10: 15073584-15073584 on Build GRCh38
rs985062375 Chr.10: 15073533-15073533 on Build GRCh38
rs535698393 Chr.10: 15073435-15073435 on Build GRCh38
rs552620432 Chr.10: 15073734-15073734 on Build GRCh38
rs919287734 Chr.10: 15073525-15073525 on Build GRCh38
rs905127622 Chr.10: 15073575-15073575 on Build GRCh38
rs760962109 Chr.10: 15073660-15073662 on Build GRCh38
rs754223039 Chr.10: 15073473-15073473 on Build GRCh38
rs946352438 Chr.10: 15073729-15073729 on Build GRCh38
rs543698729 Chr.10: 15073521-15073521 on Build GRCh38
rs532642733 Chr.10: 15073349-15073349 on Build GRCh38
rs991776517 Chr.10: 15073761-15073761 on Build GRCh38
rs7917664 Chr.10: 15073516-15073516 on Build GRCh38
rs561450171 Chr.10: 15073505-15073505 on Build GRCh38
rs933742466 Chr.10: 15073680-15073680 on Build GRCh38
rs568300116 Chr.10: 15073609-15073609 on Build GRCh38
rs1022123037 Chr.10: 15073417-15073417 on Build GRCh38
rs905117884 Chr.10: 15073748-15073748 on Build GRCh38
rs199827897 Chr.10: 15073660-15073661 on Build GRCh38
rs952116021 Chr.10: 15073598-15073598 on Build GRCh38
rs771095293 Chr.10: 15073661-15073662 on Build GRCh38
rs183131004 Chr.10: 15073563-15073563 on Build GRCh38
rs969171408 Chr.10: 15073492-15073492 on Build GRCh38
rs984800929 Chr.10: 15073608-15073608 on Build GRCh38
rs148267877 Chr.10: 15073367-15073367 on Build GRCh38
rs1046420232 Chr.10: 15073789-15073789 on Build GRCh38
rs534927849 Chr.10: 15073371-15073371 on Build GRCh38
rs185725238 Chr.10: 15073462-15073462 on Build GRCh38
rs900883697 Chr.10: 15073552-15073552 on Build GRCh38
rs1052098721 Chr.10: 15073554-15073554 on Build GRCh38
rs1038133612 Chr.10: 15073476-15073476 on Build GRCh38
rs910824031 Chr.10: 15073536-15073536 on Build GRCh38
rs1010590046 Chr.10: 15073564-15073564 on Build GRCh38
rs190938250 Chr.10: 15073553-15073553 on Build GRCh38
rs939327191 Chr.10: 15073432-15073432 on Build GRCh38
rs550502344 Chr.10: 15073655-15073655 on Build GRCh38
rs558312045 Chr.10: 15073477-15073477 on Build GRCh38
rs961080377 Chr.10: 15073520-15073520 on Build GRCh38
rs548543187 Chr.10: 15073602-15073602 on Build GRCh38
rs7081383 Chr.10: 15073591-15073591 on Build GRCh38
rs970884192 Chr.10: 15073377-15073377 on Build GRCh38
rs767186061 Chr.10: 15073431-15073431 on Build GRCh38
rs929759458 Chr.10: 15073517-15073517 on Build GRCh38
rs546818499 Chr.10: 15073368-15073368 on Build GRCh38
rs7902459 Chr.10: 15073580-15073580 on Build GRCh38
rs188222843 Chr.10: 15073760-15073760 on Build GRCh38
rs530826917 Chr.10: 15073615-15073615 on Build GRCh38
rs568537287 Chr.10: 15073387-15073387 on Build GRCh38
rs989141374 Chr.10: 15073489-15073489 on Build GRCh38
rs11292797 Chr.10: 15073679-15073679 on Build GRCh38
rs114542587 Chr.10: 15073383-15073383 on Build GRCh38
rs150802704 Chr.10: 15073427-15073427 on Build GRCh38
rs892380099 Chr.10: 15073479-15073479 on Build GRCh38
rs942406047 Chr.10: 15073461-15073461 on Build GRCh38
rs948083004 Chr.10: 15073560-15073572 on Build GRCh38
rs145445602 Chr.10: 15073747-15073747 on Build GRCh38
rs915839284 Chr.10: 15073769-15073769 on Build GRCh38
rs397847013 Chr.10: 15073678-15073678 on Build GRCh38
rs572035250 Chr.10: 15073592-15073592 on Build GRCh38
rs976421341 Chr.10: 15073699-15073699 on Build GRCh38
rs141322592 Chr.10: 15073398-15073398 on Build GRCh38
rs557106837 Chr.10: 15073442-15073442 on Build GRCh38
rs943486736 Chr.10: 15073542-15073542 on Build GRCh38
rs976244696 Chr.10: 15073617-15073617 on Build GRCh38
rs909690515 Chr.10: 15073740-15073740 on Build GRCh38
rs1001955222 Chr.10: 15073576-15073576 on Build GRCh38
rs145268129 Chr.10: 15073488-15073488 on Build GRCh38

View Full Product Details