Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 16436851-16437331 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTER
Entrez Gene ID
9317
Gene Name
phosphotriesterase related
Gene Aliases
HPHRP, RPR-1
UniGene
Hs.444321
Gene Chromosome Location
Chr.10: 16436943-16517962 on Build GRCh38
Transcripts
NM_001001484.2 NM_001261836.1 NM_001261837.1 NM_001261838.1 NM_030664.4 XM_017016929.1

SNP Detail

SNP ID Chromosome Location
rs576321320 Chr.10: 16436890-16436890 on Build GRCh38
rs925897395 Chr.10: 16437131-16437131 on Build GRCh38
rs905413231 Chr.10: 16437286-16437286 on Build GRCh38
rs898001815 Chr.10: 16437089-16437089 on Build GRCh38
rs386741377 Chr.10: 16437252-16437257 on Build GRCh38
rs999848316 Chr.10: 16437283-16437283 on Build GRCh38
rs534676315 Chr.10: 16436954-16436954 on Build GRCh38
rs897984935 Chr.10: 16437253-16437253 on Build GRCh38
rs537698791 Chr.10: 16436895-16436895 on Build GRCh38
rs952389643 Chr.10: 16436974-16436974 on Build GRCh38
rs907891767 Chr.10: 16437189-16437189 on Build GRCh38
rs1001229980 Chr.10: 16437168-16437171 on Build GRCh38
rs565700811 Chr.10: 16436871-16436871 on Build GRCh38
rs531503355 Chr.10: 16437080-16437080 on Build GRCh38
rs967239633 Chr.10: 16437100-16437100 on Build GRCh38
rs543604144 Chr.10: 16437133-16437133 on Build GRCh38
rs913020618 Chr.10: 16437193-16437193 on Build GRCh38
rs940693184 Chr.10: 16437191-16437191 on Build GRCh38
rs139098984 Chr.10: 16437233-16437233 on Build GRCh38
rs749667696 Chr.10: 16437168-16437168 on Build GRCh38
rs916651174 Chr.10: 16437147-16437147 on Build GRCh38
rs958520391 Chr.10: 16437138-16437138 on Build GRCh38
rs7894216 Chr.10: 16437252-16437252 on Build GRCh38
rs1055600971 Chr.10: 16436952-16436952 on Build GRCh38
rs17138611 Chr.10: 16437187-16437187 on Build GRCh38
rs932010055 Chr.10: 16437051-16437051 on Build GRCh38
rs541527942 Chr.10: 16436982-16436982 on Build GRCh38
rs539427995 Chr.10: 16436876-16436876 on Build GRCh38
rs565821708 Chr.10: 16437206-16437206 on Build GRCh38
rs913136462 Chr.10: 16436894-16436894 on Build GRCh38
rs948782606 Chr.10: 16437287-16437287 on Build GRCh38
rs945912509 Chr.10: 16437020-16437020 on Build GRCh38
rs570974458 Chr.10: 16436912-16436912 on Build GRCh38
rs768264650 Chr.10: 16437104-16437104 on Build GRCh38
rs561690411 Chr.10: 16437156-16437156 on Build GRCh38
rs765948469 Chr.10: 16437038-16437038 on Build GRCh38
rs927052251 Chr.10: 16436994-16436994 on Build GRCh38
rs73593456 Chr.10: 16437218-16437218 on Build GRCh38
rs553109785 Chr.10: 16437002-16437002 on Build GRCh38
rs146686576 Chr.10: 16437205-16437205 on Build GRCh38
rs978531253 Chr.10: 16436909-16436909 on Build GRCh38
rs551360924 Chr.10: 16437223-16437223 on Build GRCh38
rs1032884929 Chr.10: 16437295-16437295 on Build GRCh38
rs7910712 Chr.10: 16437257-16437257 on Build GRCh38
rs1008941137 Chr.10: 16437079-16437079 on Build GRCh38
rs991373555 Chr.10: 16437144-16437144 on Build GRCh38
rs758965236 Chr.10: 16436915-16436915 on Build GRCh38
rs1046433636 Chr.10: 16437005-16437005 on Build GRCh38
rs1043014648 Chr.10: 16437032-16437033 on Build GRCh38
rs7922050 Chr.10: 16437008-16437008 on Build GRCh38
rs979845174 Chr.10: 16436975-16436975 on Build GRCh38
rs916431789 Chr.10: 16437279-16437279 on Build GRCh38
rs978667218 Chr.10: 16437129-16437129 on Build GRCh38
rs1019955717 Chr.10: 16437086-16437086 on Build GRCh38
rs572098559 Chr.10: 16437009-16437009 on Build GRCh38
rs74125099 Chr.10: 16437063-16437063 on Build GRCh38
rs986071483 Chr.10: 16437230-16437230 on Build GRCh38
rs1008660650 Chr.10: 16437248-16437248 on Build GRCh38
rs535474700 Chr.10: 16437066-16437066 on Build GRCh38
rs558066537 Chr.10: 16436886-16436886 on Build GRCh38
rs143963673 Chr.10: 16437264-16437264 on Build GRCh38
rs552711803 Chr.10: 16436903-16436903 on Build GRCh38
rs959752370 Chr.10: 16437170-16437170 on Build GRCh38
rs1037944103 Chr.10: 16437194-16437194 on Build GRCh38
rs952339293 Chr.10: 16437167-16437167 on Build GRCh38
rs899447601 Chr.10: 16437200-16437200 on Build GRCh38
rs190715956 Chr.10: 16437039-16437039 on Build GRCh38
rs778250174 Chr.10: 16436937-16436937 on Build GRCh38
rs1037847465 Chr.10: 16437037-16437037 on Build GRCh38

View Full Product Details