Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 16521668-16522187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QL3
Entrez Gene ID
389941
Gene Name
complement C1q like 3
Gene Aliases
C1QTNF13, C1ql, CTRP13, K100
UniGene
Hs.676792
Gene Chromosome Location
Chr.10: 16513743-16522005 on Build GRCh38
Transcripts
NM_001010908.1

SNP Detail

SNP ID Chromosome Location
rs994083644 Chr.10: 16521977-16521977 on Build GRCh38
rs758162302 Chr.10: 16521931-16521931 on Build GRCh38
rs964737690 Chr.10: 16522120-16522120 on Build GRCh38
rs1040881773 Chr.10: 16521966-16521966 on Build GRCh38
rs934915370 Chr.10: 16521800-16521800 on Build GRCh38
rs1057069449 Chr.10: 16521987-16521987 on Build GRCh38
rs1041525740 Chr.10: 16521897-16521897 on Build GRCh38
rs971148516 Chr.10: 16522097-16522097 on Build GRCh38
rs10610563 Chr.10: 16522016-16522019 on Build GRCh38
rs576442449 Chr.10: 16521745-16521745 on Build GRCh38
rs114297869 Chr.10: 16521960-16521960 on Build GRCh38
rs1006442197 Chr.10: 16522009-16522009 on Build GRCh38
rs909507214 Chr.10: 16521826-16521826 on Build GRCh38
rs895675340 Chr.10: 16521724-16521725 on Build GRCh38
rs780664765 Chr.10: 16522106-16522106 on Build GRCh38
rs994096091 Chr.10: 16522007-16522007 on Build GRCh38
rs1026792933 Chr.10: 16522061-16522061 on Build GRCh38
rs550610290 Chr.10: 16521751-16521751 on Build GRCh38
rs983805318 Chr.10: 16521819-16521819 on Build GRCh38
rs189357616 Chr.10: 16521774-16521774 on Build GRCh38
rs901912910 Chr.10: 16521973-16521973 on Build GRCh38
rs935829585 Chr.10: 16522111-16522111 on Build GRCh38
rs756578815 Chr.10: 16522156-16522156 on Build GRCh38
rs368966277 Chr.10: 16522091-16522091 on Build GRCh38
rs1032952119 Chr.10: 16521752-16521752 on Build GRCh38
rs748671695 Chr.10: 16521983-16521983 on Build GRCh38
rs879003894 Chr.10: 16522016-16522016 on Build GRCh38
rs1054393190 Chr.10: 16521946-16521946 on Build GRCh38
rs991141708 Chr.10: 16522079-16522079 on Build GRCh38
rs893307307 Chr.10: 16521863-16521863 on Build GRCh38
rs61844132 Chr.10: 16521925-16521925 on Build GRCh38
rs952673419 Chr.10: 16522078-16522078 on Build GRCh38
rs949954173 Chr.10: 16521886-16521886 on Build GRCh38
rs952299792 Chr.10: 16521809-16521809 on Build GRCh38
rs1055635661 Chr.10: 16521706-16521706 on Build GRCh38
rs59639125 Chr.10: 16522028-16522030 on Build GRCh38
rs1053181656 Chr.10: 16521814-16521814 on Build GRCh38
rs751155319 Chr.10: 16521962-16521962 on Build GRCh38
rs555337219 Chr.10: 16521796-16521796 on Build GRCh38
rs1007565351 Chr.10: 16521754-16521754 on Build GRCh38
rs190395798 Chr.10: 16522150-16522150 on Build GRCh38
rs35860209 Chr.10: 16522030-16522031 on Build GRCh38
rs577705116 Chr.10: 16522060-16522060 on Build GRCh38
rs976295655 Chr.10: 16521783-16521783 on Build GRCh38
rs765241199 Chr.10: 16522152-16522152 on Build GRCh38
rs1000167046 Chr.10: 16521749-16521749 on Build GRCh38
rs949913985 Chr.10: 16521737-16521737 on Build GRCh38
rs796257300 Chr.10: 16522016-16522017 on Build GRCh38
rs1042051188 Chr.10: 16521739-16521739 on Build GRCh38
rs555979797 Chr.10: 16522018-16522019 on Build GRCh38
rs186576057 Chr.10: 16522104-16522104 on Build GRCh38
rs923280357 Chr.10: 16522161-16522161 on Build GRCh38
rs897035676 Chr.10: 16521994-16521994 on Build GRCh38
rs958385596 Chr.10: 16522071-16522071 on Build GRCh38
rs1018978210 Chr.10: 16521758-16521758 on Build GRCh38
rs943391427 Chr.10: 16521753-16521753 on Build GRCh38
rs756825554 Chr.10: 16522098-16522098 on Build GRCh38
rs924435292 Chr.10: 16522107-16522107 on Build GRCh38
rs182407082 Chr.10: 16521939-16521939 on Build GRCh38
rs972239559 Chr.10: 16521797-16521797 on Build GRCh38
rs1054339763 Chr.10: 16522144-16522144 on Build GRCh38
rs991279021 Chr.10: 16521884-16521884 on Build GRCh38
rs958449400 Chr.10: 16521870-16521870 on Build GRCh38
rs1002574577 Chr.10: 16521961-16521961 on Build GRCh38
rs947132158 Chr.10: 16521885-16521885 on Build GRCh38
rs148898321 Chr.10: 16521984-16521984 on Build GRCh38
rs1040345903 Chr.10: 16522164-16522164 on Build GRCh38
rs889148648 Chr.10: 16521948-16521948 on Build GRCh38

View Full Product Details