Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 20780258-20780801 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEBL
Entrez Gene ID
10529
Gene Name
nebulette
Gene Aliases
LASP2, LNEBL
UniGene
Hs.5025
Gene Chromosome Location
Chr.10: 20779973-21174187 on Build GRCh38
Transcripts
NM_001173484.1 NM_006393.2 NM_213569.2 XM_005252342.4 XM_005252343.4 XM_005252344.4 XM_011519291.2 XM_017015467.1 XM_017015468.1

SNP Detail

SNP ID Chromosome Location
rs1021526812 Chr.10: 20780625-20780625 on Build GRCh38
rs895324008 Chr.10: 20780521-20780521 on Build GRCh38
rs45454397 Chr.10: 20780685-20780685 on Build GRCh38
rs1052385180 Chr.10: 20780421-20780421 on Build GRCh38
rs939430407 Chr.10: 20780519-20780519 on Build GRCh38
rs923940217 Chr.10: 20780648-20780648 on Build GRCh38
rs948173256 Chr.10: 20780531-20780531 on Build GRCh38
rs913837175 Chr.10: 20780647-20780647 on Build GRCh38
rs532667000 Chr.10: 20780571-20780571 on Build GRCh38
rs746198163 Chr.10: 20780374-20780374 on Build GRCh38
rs993806215 Chr.10: 20780399-20780399 on Build GRCh38
rs1004187624 Chr.10: 20780748-20780748 on Build GRCh38
rs767761348 Chr.10: 20780593-20780593 on Build GRCh38
rs980967500 Chr.10: 20780613-20780613 on Build GRCh38
rs920207258 Chr.10: 20780723-20780723 on Build GRCh38
rs943922173 Chr.10: 20780673-20780673 on Build GRCh38
rs749736936 Chr.10: 20780387-20780387 on Build GRCh38
rs957719875 Chr.10: 20780627-20780627 on Build GRCh38
rs751845908 Chr.10: 20780771-20780771 on Build GRCh38
rs960228548 Chr.10: 20780292-20780292 on Build GRCh38
rs892375271 Chr.10: 20780440-20780440 on Build GRCh38
rs761643122 Chr.10: 20780683-20780683 on Build GRCh38
rs866432002 Chr.10: 20780420-20780420 on Build GRCh38
rs149868974 Chr.10: 20780577-20780577 on Build GRCh38
rs558401705 Chr.10: 20780464-20780464 on Build GRCh38
rs895434593 Chr.10: 20780373-20780373 on Build GRCh38
rs960706544 Chr.10: 20780665-20780665 on Build GRCh38
rs943874115 Chr.10: 20780460-20780460 on Build GRCh38
rs1025422694 Chr.10: 20780407-20780407 on Build GRCh38
rs898182567 Chr.10: 20780398-20780398 on Build GRCh38
rs987785661 Chr.10: 20780640-20780640 on Build GRCh38
rs1015825410 Chr.10: 20780557-20780557 on Build GRCh38
rs1041251697 Chr.10: 20780461-20780461 on Build GRCh38
rs993387761 Chr.10: 20780316-20780316 on Build GRCh38
rs997304255 Chr.10: 20780475-20780475 on Build GRCh38
rs61153696 Chr.10: 20780313-20780313 on Build GRCh38
rs1036964483 Chr.10: 20780767-20780767 on Build GRCh38
rs902580627 Chr.10: 20780367-20780367 on Build GRCh38
rs1040889297 Chr.10: 20780674-20780674 on Build GRCh38
rs528628389 Chr.10: 20780690-20780690 on Build GRCh38
rs543920601 Chr.10: 20780565-20780565 on Build GRCh38
rs78728646 Chr.10: 20780643-20780643 on Build GRCh38
rs563236052 Chr.10: 20780590-20780590 on Build GRCh38
rs41277358 Chr.10: 20780375-20780375 on Build GRCh38
rs546776032 Chr.10: 20780752-20780752 on Build GRCh38
rs756290903 Chr.10: 20780406-20780407 on Build GRCh38
rs1017159589 Chr.10: 20780379-20780379 on Build GRCh38
rs1038650926 Chr.10: 20780541-20780542 on Build GRCh38
rs1017953419 Chr.10: 20780614-20780614 on Build GRCh38
rs930216500 Chr.10: 20780743-20780743 on Build GRCh38
rs946743929 Chr.10: 20780493-20780493 on Build GRCh38
rs1048472122 Chr.10: 20780483-20780483 on Build GRCh38
rs150687182 Chr.10: 20780331-20780331 on Build GRCh38
rs939558387 Chr.10: 20780368-20780368 on Build GRCh38
rs967260896 Chr.10: 20780635-20780636 on Build GRCh38
rs181186058 Chr.10: 20780479-20780479 on Build GRCh38
rs924085088 Chr.10: 20780506-20780506 on Build GRCh38
rs914468477 Chr.10: 20780484-20780484 on Build GRCh38
rs978486341 Chr.10: 20780497-20780497 on Build GRCh38
rs888482794 Chr.10: 20780520-20780520 on Build GRCh38
rs992664513 Chr.10: 20780669-20780669 on Build GRCh38
rs774488421 Chr.10: 20780556-20780556 on Build GRCh38
rs898729986 Chr.10: 20780569-20780569 on Build GRCh38
rs888514848 Chr.10: 20780699-20780699 on Build GRCh38
rs552545448 Chr.10: 20780676-20780676 on Build GRCh38
rs776001038 Chr.10: 20780377-20780377 on Build GRCh38
rs1007872231 Chr.10: 20780777-20780777 on Build GRCh38
rs993875512 Chr.10: 20780575-20780575 on Build GRCh38
rs947767571 Chr.10: 20780370-20780370 on Build GRCh38
rs768622883 Chr.10: 20780471-20780471 on Build GRCh38
rs185544685 Chr.10: 20780616-20780616 on Build GRCh38
rs969168683 Chr.10: 20780612-20780612 on Build GRCh38

View Full Product Details