Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 22535954-22536461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIP4K2A
Entrez Gene ID
5305
Gene Name
phosphatidylinositol-5-phosphate 4-kinase type 2 alpha
Gene Aliases
PI5P4KA, PIP5K2A, PIP5KII-alpha, PIP5KIIA, PIPK
UniGene
Hs.57079
Gene Chromosome Location
Chr.10: 22534837-22714574 on Build GRCh38
Transcripts
NM_005028.4 XM_006717450.2 XM_011519496.1 XM_017016330.1 XM_017016331.1 XM_017016332.1

SNP Detail

SNP ID Chromosome Location
rs1159 Chr.10: 22536424-22536424 on Build GRCh38
rs528294023 Chr.10: 22536166-22536166 on Build GRCh38
rs966342721 Chr.10: 22536401-22536401 on Build GRCh38
rs920992911 Chr.10: 22536249-22536249 on Build GRCh38
rs752789959 Chr.10: 22535979-22535979 on Build GRCh38
rs544838642 Chr.10: 22536180-22536180 on Build GRCh38
rs988352874 Chr.10: 22536205-22536205 on Build GRCh38
rs932356949 Chr.10: 22536262-22536262 on Build GRCh38
rs989216755 Chr.10: 22536361-22536361 on Build GRCh38
rs143766959 Chr.10: 22536428-22536428 on Build GRCh38
rs1014486783 Chr.10: 22536363-22536363 on Build GRCh38
rs3215826 Chr.10: 22536359-22536359 on Build GRCh38
rs960122229 Chr.10: 22536056-22536056 on Build GRCh38
rs910261653 Chr.10: 22536151-22536151 on Build GRCh38
rs895150085 Chr.10: 22535989-22535989 on Build GRCh38
rs1048321423 Chr.10: 22536021-22536021 on Build GRCh38
rs542265429 Chr.10: 22536414-22536415 on Build GRCh38
rs954062655 Chr.10: 22536430-22536430 on Build GRCh38
rs939141075 Chr.10: 22536218-22536218 on Build GRCh38
rs567701965 Chr.10: 22536354-22536354 on Build GRCh38
rs397719702 Chr.10: 22536360-22536360 on Build GRCh38
rs543036149 Chr.10: 22536017-22536017 on Build GRCh38
rs943805078 Chr.10: 22535980-22535980 on Build GRCh38
rs1037327890 Chr.10: 22536340-22536340 on Build GRCh38
rs996453040 Chr.10: 22536020-22536020 on Build GRCh38
rs1025467437 Chr.10: 22536106-22536106 on Build GRCh38
rs939961424 Chr.10: 22536339-22536339 on Build GRCh38
rs113084433 Chr.10: 22536233-22536233 on Build GRCh38
rs994368267 Chr.10: 22536029-22536029 on Build GRCh38
rs148948464 Chr.10: 22536003-22536003 on Build GRCh38
rs1048059290 Chr.10: 22536266-22536266 on Build GRCh38
rs764023973 Chr.10: 22535996-22535996 on Build GRCh38
rs944172175 Chr.10: 22536229-22536229 on Build GRCh38
rs895679749 Chr.10: 22536352-22536352 on Build GRCh38
rs745636639 Chr.10: 22536332-22536332 on Build GRCh38
rs994399355 Chr.10: 22536301-22536305 on Build GRCh38
rs539069932 Chr.10: 22536180-22536181 on Build GRCh38
rs758495715 Chr.10: 22535992-22535992 on Build GRCh38
rs1018442449 Chr.10: 22536355-22536355 on Build GRCh38
rs757144513 Chr.10: 22536220-22536220 on Build GRCh38
rs916395689 Chr.10: 22536424-22536425 on Build GRCh38
rs1013685091 Chr.10: 22536083-22536083 on Build GRCh38
rs1158 Chr.10: 22536359-22536359 on Build GRCh38
rs990343917 Chr.10: 22536125-22536125 on Build GRCh38
rs891303620 Chr.10: 22536346-22536346 on Build GRCh38
rs542553400 Chr.10: 22536104-22536104 on Build GRCh38
rs905269224 Chr.10: 22536409-22536409 on Build GRCh38
rs185951018 Chr.10: 22536139-22536139 on Build GRCh38
rs1025097379 Chr.10: 22536382-22536382 on Build GRCh38
rs987242132 Chr.10: 22536140-22536140 on Build GRCh38
rs76630921 Chr.10: 22536027-22536027 on Build GRCh38
rs757196516 Chr.10: 22536211-22536213 on Build GRCh38
rs76451424 Chr.10: 22536054-22536054 on Build GRCh38
rs969439744 Chr.10: 22536145-22536145 on Build GRCh38
rs116492026 Chr.10: 22536224-22536224 on Build GRCh38
rs1032627796 Chr.10: 22536196-22536196 on Build GRCh38
rs888076386 Chr.10: 22536283-22536283 on Build GRCh38
rs1038355990 Chr.10: 22536271-22536271 on Build GRCh38
rs571618758 Chr.10: 22536023-22536023 on Build GRCh38
rs8341 Chr.10: 22536204-22536204 on Build GRCh38
rs941450441 Chr.10: 22536256-22536256 on Build GRCh38

View Full Product Details