Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 26720133-26720631 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDSS1
Entrez Gene ID
23590
Gene Name
prenyl (decaprenyl) diphosphate synthase, subunit 1
Gene Aliases
COQ1, COQ10D2, DPS, SPS, TPRT, TPT, TPT 1, hDPS1
UniGene
Hs.558468
Gene Chromosome Location
Chr.10: 26697052-26746798 on Build GRCh38
Transcripts
NM_001321978.1 NM_001321979.1 NM_014317.4 XM_011519437.2 XM_017016011.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
917347 c.489C>A p.R163R Chr.10: 26720239-26720239 on Build GRCh38
3436891 c.537C>T p.H179H Chr.10: 26720287-26720287 on Build GRCh38
4823958 c.601G>A p.E201K Chr.10: 26720351-26720351 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754510996 Chr.10: 26720449-26720449 on Build GRCh38
rs748266908 Chr.10: 26720278-26720278 on Build GRCh38
rs985371706 Chr.10: 26720452-26720452 on Build GRCh38
rs540043853 Chr.10: 26720330-26720330 on Build GRCh38
rs745689033 Chr.10: 26720221-26720221 on Build GRCh38
rs552242296 Chr.10: 26720338-26720338 on Build GRCh38
rs189215146 Chr.10: 26720381-26720381 on Build GRCh38
rs914000422 Chr.10: 26720573-26720573 on Build GRCh38
rs1002472641 Chr.10: 26720236-26720236 on Build GRCh38
rs758072939 Chr.10: 26720366-26720366 on Build GRCh38
rs370492173 Chr.10: 26720185-26720185 on Build GRCh38
rs897499362 Chr.10: 26720171-26720171 on Build GRCh38
rs910576192 Chr.10: 26720161-26720161 on Build GRCh38
rs921287700 Chr.10: 26720602-26720602 on Build GRCh38
rs199585120 Chr.10: 26720373-26720373 on Build GRCh38
rs773495425 Chr.10: 26720287-26720287 on Build GRCh38
rs944695053 Chr.10: 26720174-26720174 on Build GRCh38
rs545634593 Chr.10: 26720417-26720417 on Build GRCh38
rs1027272149 Chr.10: 26720438-26720438 on Build GRCh38
rs976148633 Chr.10: 26720178-26720178 on Build GRCh38
rs968003045 Chr.10: 26720557-26720557 on Build GRCh38
rs913868781 Chr.10: 26720305-26720305 on Build GRCh38
rs547415481 Chr.10: 26720547-26720547 on Build GRCh38
rs780198984 Chr.10: 26720238-26720238 on Build GRCh38
rs770030240 Chr.10: 26720281-26720281 on Build GRCh38
rs144149122 Chr.10: 26720303-26720303 on Build GRCh38
rs1027853662 Chr.10: 26720205-26720205 on Build GRCh38
rs116424900 Chr.10: 26720339-26720339 on Build GRCh38
rs146170591 Chr.10: 26720290-26720290 on Build GRCh38
rs777578120 Chr.10: 26720374-26720374 on Build GRCh38
rs940217264 Chr.10: 26720403-26720403 on Build GRCh38
rs1044251533 Chr.10: 26720199-26720199 on Build GRCh38
rs749651097 Chr.10: 26720411-26720411 on Build GRCh38
rs572597825 Chr.10: 26720412-26720412 on Build GRCh38
rs532941777 Chr.10: 26720419-26720419 on Build GRCh38
rs1055367473 Chr.10: 26720300-26720300 on Build GRCh38
rs1009429716 Chr.10: 26720532-26720532 on Build GRCh38
rs148661087 Chr.10: 26720357-26720357 on Build GRCh38
rs904971603 Chr.10: 26720416-26720416 on Build GRCh38
rs969248593 Chr.10: 26720227-26720227 on Build GRCh38
rs78311898 Chr.10: 26720462-26720462 on Build GRCh38
rs763562554 Chr.10: 26720309-26720309 on Build GRCh38
rs753472393 Chr.10: 26720184-26720184 on Build GRCh38
rs569304206 Chr.10: 26720325-26720325 on Build GRCh38
rs370623507 Chr.10: 26720402-26720402 on Build GRCh38
rs754963544 Chr.10: 26720389-26720389 on Build GRCh38
rs560057978 Chr.10: 26720394-26720394 on Build GRCh38
rs764994361 Chr.10: 26720358-26720358 on Build GRCh38
rs1780196 Chr.10: 26720405-26720405 on Build GRCh38
rs750229694 Chr.10: 26720363-26720363 on Build GRCh38
rs769950262 Chr.10: 26720404-26720404 on Build GRCh38
rs747109562 Chr.10: 26720239-26720239 on Build GRCh38
rs755634985 Chr.10: 26720181-26720181 on Build GRCh38
rs771418338 Chr.10: 26720295-26720295 on Build GRCh38
rs953884469 Chr.10: 26720441-26720441 on Build GRCh38
rs976013222 Chr.10: 26720562-26720562 on Build GRCh38
rs374748493 Chr.10: 26720240-26720240 on Build GRCh38
rs139094994 Chr.10: 26720302-26720302 on Build GRCh38
rs546764434 Chr.10: 26720499-26720499 on Build GRCh38
rs772887446 Chr.10: 26720182-26720182 on Build GRCh38
rs3802606 Chr.10: 26720578-26720578 on Build GRCh38
rs780107336 Chr.10: 26720380-26720380 on Build GRCh38
rs753411855 Chr.10: 26720332-26720332 on Build GRCh38
rs1001975942 Chr.10: 26720428-26720428 on Build GRCh38
rs758101398 Chr.10: 26720294-26720294 on Build GRCh38
rs750746851 Chr.10: 26720405-26720406 on Build GRCh38
rs1051671197 Chr.10: 26720383-26720383 on Build GRCh38
rs758353274 Chr.10: 26720237-26720237 on Build GRCh38
rs778801305 Chr.10: 26720193-26720193 on Build GRCh38
rs754963280 Chr.10: 26720450-26720450 on Build GRCh38

View Full Product Details