Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 26748497-26749003 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABI1
Entrez Gene ID
10006
Gene Name
abl interactor 1
Gene Aliases
ABI-1, ABLBP4, E3B1, NAP1BP, SSH3BP, SSH3BP1
UniGene
Hs.508148
Gene Chromosome Location
Chr.10: 26746596-26861087 on Build GRCh38
Transcripts
NM_001012750.2 NM_001012751.2 NM_001012752.2 NM_001178116.1 NM_001178119.1 NM_001178120.1 NM_001178121.1 NM_001178122.1 NM_001178123.1 NM_001178124.1 NM_001178125.1 NM_005470.3 XM_005252332.3 XM_005252333.3 XM_006717359.2 XM_006717360.2 XM_006717361.2 XM_017015458.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
295113 c.1461C>T p.V487V Chr.10: 26748636-26748636 on Build GRCh38
6590934 c.1456G>T p.G486* Chr.10: 26748641-26748641 on Build GRCh38
6048394 c.1364_1365delAT p.Y455fs*1 Chr.10: 26748732-26748733 on Build GRCh38
3748501 c.1480C>A p.L494M Chr.10: 26748617-26748617 on Build GRCh38
6590938 c.1411G>A p.A471T Chr.10: 26748686-26748686 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs943777352 Chr.10: 26748822-26748825 on Build GRCh38
rs772738804 Chr.10: 26748681-26748681 on Build GRCh38
rs756274226 Chr.10: 26748631-26748631 on Build GRCh38
rs768975419 Chr.10: 26748533-26748536 on Build GRCh38
rs770706196 Chr.10: 26748775-26748775 on Build GRCh38
rs939854812 Chr.10: 26748972-26748972 on Build GRCh38
rs1034214220 Chr.10: 26748702-26748702 on Build GRCh38
rs903881514 Chr.10: 26748590-26748590 on Build GRCh38
rs769783682 Chr.10: 26748541-26748543 on Build GRCh38
rs932353429 Chr.10: 26748813-26748814 on Build GRCh38
rs767457879 Chr.10: 26748787-26748787 on Build GRCh38
rs777696418 Chr.10: 26748927-26748927 on Build GRCh38
rs1022308692 Chr.10: 26748838-26748838 on Build GRCh38
rs373977143 Chr.10: 26748785-26748785 on Build GRCh38
rs1002212935 Chr.10: 26748871-26748871 on Build GRCh38
rs891962286 Chr.10: 26748811-26748811 on Build GRCh38
rs748620918 Chr.10: 26748600-26748600 on Build GRCh38
rs752780903 Chr.10: 26748628-26748628 on Build GRCh38
rs3839922 Chr.10: 26748871-26748871 on Build GRCh38
rs778646055 Chr.10: 26748816-26748817 on Build GRCh38
rs368535118 Chr.10: 26748878-26748878 on Build GRCh38
rs868639534 Chr.10: 26748836-26748836 on Build GRCh38
rs3802612 Chr.10: 26748964-26748964 on Build GRCh38
rs146846937 Chr.10: 26748817-26748818 on Build GRCh38
rs1013699864 Chr.10: 26748933-26748933 on Build GRCh38
rs762630640 Chr.10: 26748769-26748769 on Build GRCh38
rs1002991140 Chr.10: 26748743-26748743 on Build GRCh38
rs763675140 Chr.10: 26748559-26748560 on Build GRCh38
rs767541582 Chr.10: 26748569-26748569 on Build GRCh38
rs368838171 Chr.10: 26748957-26748957 on Build GRCh38
rs570868988 Chr.10: 26748577-26748577 on Build GRCh38
rs757579163 Chr.10: 26748870-26748871 on Build GRCh38
rs1033647697 Chr.10: 26748919-26748919 on Build GRCh38
rs779266640 Chr.10: 26748669-26748669 on Build GRCh38
rs539883149 Chr.10: 26748828-26748828 on Build GRCh38
rs369605742 Chr.10: 26748818-26748819 on Build GRCh38
rs775560926 Chr.10: 26748558-26748559 on Build GRCh38
rs376485217 Chr.10: 26748534-26748534 on Build GRCh38
rs750450988 Chr.10: 26748817-26748820 on Build GRCh38
rs113435131 Chr.10: 26748559-26748559 on Build GRCh38
rs757681575 Chr.10: 26748650-26748650 on Build GRCh38
rs912357213 Chr.10: 26748969-26748969 on Build GRCh38
rs979115543 Chr.10: 26748848-26748848 on Build GRCh38
rs774042408 Chr.10: 26748777-26748777 on Build GRCh38
rs769565941 Chr.10: 26748727-26748727 on Build GRCh38
rs75446996 Chr.10: 26748559-26748559 on Build GRCh38
rs780790043 Chr.10: 26748683-26748683 on Build GRCh38
rs1015739448 Chr.10: 26748766-26748766 on Build GRCh38
rs947188248 Chr.10: 26748850-26748850 on Build GRCh38
rs369813692 Chr.10: 26748526-26748526 on Build GRCh38
rs372879838 Chr.10: 26748760-26748760 on Build GRCh38
rs778031305 Chr.10: 26748638-26748638 on Build GRCh38
rs775227312 Chr.10: 26748819-26748822 on Build GRCh38
rs7086847 Chr.10: 26748567-26748567 on Build GRCh38
rs749614071 Chr.10: 26748644-26748644 on Build GRCh38
rs747717030 Chr.10: 26748721-26748721 on Build GRCh38
rs898459890 Chr.10: 26748546-26748546 on Build GRCh38
rs3802611 Chr.10: 26748839-26748839 on Build GRCh38
rs746338683 Chr.10: 26748674-26748674 on Build GRCh38
rs1044441816 Chr.10: 26748615-26748615 on Build GRCh38
rs762872038 Chr.10: 26748535-26748535 on Build GRCh38
rs186037177 Chr.10: 26748926-26748926 on Build GRCh38
rs1028062746 Chr.10: 26748866-26748866 on Build GRCh38
rs569995035 Chr.10: 26748846-26748846 on Build GRCh38
rs766192413 Chr.10: 26748562-26748562 on Build GRCh38
rs377616132 Chr.10: 26748566-26748566 on Build GRCh38
rs752764466 Chr.10: 26748789-26748789 on Build GRCh38
rs373159403 Chr.10: 26748773-26748774 on Build GRCh38

View Full Product Details