Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 27734337-27734867 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MKX
Entrez Gene ID
283078
Gene Name
mohawk homeobox
Gene Aliases
C10orf48, IFRX, IRXL1
UniGene
Hs.128193
Gene Chromosome Location
Chr.10: 27672874-27745849 on Build GRCh38
Transcripts
NM_001242702.1 NM_173576.2 XM_017016105.1 XM_017016106.1 XM_017016107.1 XM_017016108.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
166671 c.648C>G p.P216P Chr.10: 27734646-27734646 on Build GRCh38
6480368 c.767C>A p.S256Y Chr.10: 27734527-27734527 on Build GRCh38
1627458 c.791T>G p.V264G Chr.10: 27734503-27734503 on Build GRCh38
4702331 c.534C>T p.N178N Chr.10: 27734760-27734760 on Build GRCh38
3436973 c.582G>A p.E194E Chr.10: 27734712-27734712 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763693197 Chr.10: 27734559-27734559 on Build GRCh38
rs1013627570 Chr.10: 27734673-27734673 on Build GRCh38
rs765624213 Chr.10: 27734440-27734440 on Build GRCh38
rs750371730 Chr.10: 27734573-27734573 on Build GRCh38
rs760341032 Chr.10: 27734553-27734553 on Build GRCh38
rs558586786 Chr.10: 27734734-27734734 on Build GRCh38
rs564441402 Chr.10: 27734385-27734385 on Build GRCh38
rs374332156 Chr.10: 27734469-27734469 on Build GRCh38
rs939575741 Chr.10: 27734548-27734548 on Build GRCh38
rs773650203 Chr.10: 27734521-27734521 on Build GRCh38
rs750113734 Chr.10: 27734464-27734464 on Build GRCh38
rs371872351 Chr.10: 27734503-27734503 on Build GRCh38
rs370469899 Chr.10: 27734434-27734434 on Build GRCh38
rs748367959 Chr.10: 27734591-27734591 on Build GRCh38
rs372650648 Chr.10: 27734648-27734648 on Build GRCh38
rs765795299 Chr.10: 27734388-27734388 on Build GRCh38
rs764994992 Chr.10: 27734649-27734649 on Build GRCh38
rs371518228 Chr.10: 27734542-27734542 on Build GRCh38
rs781507547 Chr.10: 27734587-27734587 on Build GRCh38
rs767754453 Chr.10: 27734747-27734747 on Build GRCh38
rs768306454 Chr.10: 27734631-27734631 on Build GRCh38
rs138902206 Chr.10: 27734688-27734688 on Build GRCh38
rs35775843 Chr.10: 27734653-27734653 on Build GRCh38
rs985155856 Chr.10: 27734738-27734738 on Build GRCh38
rs201511812 Chr.10: 27734695-27734695 on Build GRCh38
rs141896248 Chr.10: 27734652-27734652 on Build GRCh38
rs774497750 Chr.10: 27734740-27734740 on Build GRCh38
rs201497413 Chr.10: 27734669-27734669 on Build GRCh38
rs143892033 Chr.10: 27734706-27734706 on Build GRCh38
rs757214275 Chr.10: 27734468-27734468 on Build GRCh38
rs761382120 Chr.10: 27734836-27734836 on Build GRCh38
rs375003775 Chr.10: 27734541-27734541 on Build GRCh38
rs1015551221 Chr.10: 27734457-27734457 on Build GRCh38
rs748888546 Chr.10: 27734827-27734827 on Build GRCh38
rs764307692 Chr.10: 27734765-27734765 on Build GRCh38
rs767039772 Chr.10: 27734449-27734449 on Build GRCh38
rs760315971 Chr.10: 27734601-27734601 on Build GRCh38
rs147198302 Chr.10: 27734571-27734571 on Build GRCh38
rs754164534 Chr.10: 27734766-27734766 on Build GRCh38
rs780600706 Chr.10: 27734809-27734809 on Build GRCh38
rs267602459 Chr.10: 27734614-27734614 on Build GRCh38
rs778169038 Chr.10: 27734611-27734611 on Build GRCh38
rs368236674 Chr.10: 27734498-27734498 on Build GRCh38
rs770890670 Chr.10: 27734833-27734833 on Build GRCh38
rs755604261 Chr.10: 27734804-27734804 on Build GRCh38
rs753632136 Chr.10: 27734569-27734569 on Build GRCh38
rs772891422 Chr.10: 27734707-27734707 on Build GRCh38
rs547780445 Chr.10: 27734671-27734671 on Build GRCh38
rs765063362 Chr.10: 27734572-27734572 on Build GRCh38
rs760287864 Chr.10: 27734461-27734461 on Build GRCh38
rs774216352 Chr.10: 27734833-27734833 on Build GRCh38
rs775009289 Chr.10: 27734627-27734627 on Build GRCh38
rs758858814 Chr.10: 27734808-27734808 on Build GRCh38
rs758914580 Chr.10: 27734448-27734448 on Build GRCh38
rs375810471 Chr.10: 27734823-27734823 on Build GRCh38
rs748459500 Chr.10: 27734504-27734504 on Build GRCh38
rs200413424 Chr.10: 27734658-27734658 on Build GRCh38
rs762433649 Chr.10: 27734438-27734438 on Build GRCh38
rs867727269 Chr.10: 27734510-27734510 on Build GRCh38
rs139722718 Chr.10: 27734703-27734703 on Build GRCh38
rs764110595 Chr.10: 27734609-27734609 on Build GRCh38
rs896486759 Chr.10: 27734655-27734655 on Build GRCh38
rs756336977 Chr.10: 27734684-27734684 on Build GRCh38
rs761498565 Chr.10: 27734637-27734637 on Build GRCh38
rs778019547 Chr.10: 27734686-27734686 on Build GRCh38
rs759513838 Chr.10: 27734741-27734741 on Build GRCh38
rs779624616 Chr.10: 27734657-27734657 on Build GRCh38
rs760108159 Chr.10: 27734630-27734630 on Build GRCh38
rs757672109 Chr.10: 27734771-27734771 on Build GRCh38
rs771419318 Chr.10: 27734621-27734621 on Build GRCh38
rs776101097 Chr.10: 27734699-27734699 on Build GRCh38
rs780022176 Chr.10: 27734583-27734583 on Build GRCh38
rs953815283 Chr.10: 27734489-27734489 on Build GRCh38
rs920367946 Chr.10: 27734458-27734458 on Build GRCh38
rs370707355 Chr.10: 27734424-27734424 on Build GRCh38
rs552664981 Chr.10: 27734384-27734384 on Build GRCh38
rs762753004 Chr.10: 27734714-27734714 on Build GRCh38
rs377522552 Chr.10: 27734425-27734425 on Build GRCh38
rs779241706 Chr.10: 27734796-27734796 on Build GRCh38
rs774956262 Chr.10: 27734547-27734547 on Build GRCh38
rs1041023861 Chr.10: 27734567-27734567 on Build GRCh38
rs973131909 Chr.10: 27734760-27734760 on Build GRCh38
rs779331895 Chr.10: 27734694-27734694 on Build GRCh38
rs148104265 Chr.10: 27734584-27734584 on Build GRCh38
rs1043920392 Chr.10: 27734575-27734575 on Build GRCh38
rs576493288 Chr.10: 27734690-27734690 on Build GRCh38
rs138096382 Chr.10: 27734462-27734462 on Build GRCh38
rs747286454 Chr.10: 27734496-27734496 on Build GRCh38
rs749807102 Chr.10: 27734620-27734620 on Build GRCh38
rs770122927 Chr.10: 27734592-27734592 on Build GRCh38
rs752413011 Chr.10: 27734459-27734459 on Build GRCh38
rs758478667 Chr.10: 27734493-27734493 on Build GRCh38
rs758337934 Chr.10: 27734579-27734579 on Build GRCh38
rs267602458 Chr.10: 27734605-27734605 on Build GRCh38
rs528660133 Chr.10: 27734406-27734406 on Build GRCh38
rs752902262 Chr.10: 27734675-27734675 on Build GRCh38
rs773917993 Chr.10: 27734447-27734447 on Build GRCh38
rs143655800 Chr.10: 27734494-27734494 on Build GRCh38
rs148956781 Chr.10: 27734759-27734759 on Build GRCh38
rs763510823 Chr.10: 27734522-27734522 on Build GRCh38
rs146403733 Chr.10: 27734753-27734753 on Build GRCh38
rs745892674 Chr.10: 27734405-27734405 on Build GRCh38
rs750975434 Chr.10: 27734554-27734554 on Build GRCh38
rs746426904 Chr.10: 27734797-27734797 on Build GRCh38
rs776319124 Chr.10: 27734635-27734635 on Build GRCh38
rs925423954 Chr.10: 27734829-27734829 on Build GRCh38
rs770348215 Chr.10: 27734515-27734515 on Build GRCh38
rs12218027 Chr.10: 27734818-27734818 on Build GRCh38

View Full Product Details