Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 32015285-32015793 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF5B
Entrez Gene ID
3799
Gene Name
kinesin family member 5B
Gene Aliases
HEL-S-61, KINH, KNS, KNS1, UKHC
UniGene
Hs.327736
Gene Chromosome Location
Chr.10: 32009010-32056443 on Build GRCh38
Transcripts
NM_004521.2 XM_017016224.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4696171 c.2847C>T p.N949N Chr.10: 32015574-32015574 on Build GRCh38
1470275 c.2840T>C p.V947A Chr.10: 32015581-32015581 on Build GRCh38
6461124 c.2850C>T p.S950S Chr.10: 32015571-32015571 on Build GRCh38
7266851 c.2799T>C p.S933S Chr.10: 32015622-32015622 on Build GRCh38
3358740 c.2837T>G p.F946C Chr.10: 32015584-32015584 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751006235 Chr.10: 32015480-32015480 on Build GRCh38
rs748505646 Chr.10: 32015678-32015678 on Build GRCh38
rs775591282 Chr.10: 32015566-32015566 on Build GRCh38
rs11539713 Chr.10: 32015512-32015512 on Build GRCh38
rs923427440 Chr.10: 32015414-32015414 on Build GRCh38
rs770073696 Chr.10: 32015688-32015688 on Build GRCh38
rs764031233 Chr.10: 32015632-32015632 on Build GRCh38
rs574882765 Chr.10: 32015714-32015714 on Build GRCh38
rs182913371 Chr.10: 32015324-32015324 on Build GRCh38
rs770970409 Chr.10: 32015541-32015541 on Build GRCh38
rs1055055936 Chr.10: 32015372-32015372 on Build GRCh38
rs760906686 Chr.10: 32015572-32015572 on Build GRCh38
rs752313913 Chr.10: 32015613-32015613 on Build GRCh38
rs766966738 Chr.10: 32015706-32015706 on Build GRCh38
rs986284647 Chr.10: 32015755-32015755 on Build GRCh38
rs533600588 Chr.10: 32015590-32015590 on Build GRCh38
rs745764054 Chr.10: 32015646-32015646 on Build GRCh38
rs757332683 Chr.10: 32015642-32015642 on Build GRCh38
rs937804168 Chr.10: 32015357-32015357 on Build GRCh38
rs375595563 Chr.10: 32015729-32015729 on Build GRCh38
rs368618931 Chr.10: 32015631-32015631 on Build GRCh38
rs1013381334 Chr.10: 32015635-32015635 on Build GRCh38
rs754715270 Chr.10: 32015493-32015493 on Build GRCh38
rs758433680 Chr.10: 32015648-32015648 on Build GRCh38
rs752049537 Chr.10: 32015512-32015513 on Build GRCh38
rs541699828 Chr.10: 32015413-32015413 on Build GRCh38
rs769044593 Chr.10: 32015583-32015583 on Build GRCh38
rs201609384 Chr.10: 32015691-32015691 on Build GRCh38
rs757449614 Chr.10: 32015390-32015390 on Build GRCh38
rs201874978 Chr.10: 32015524-32015524 on Build GRCh38
rs1000501651 Chr.10: 32015337-32015337 on Build GRCh38
rs551287583 Chr.10: 32015424-32015424 on Build GRCh38
rs757854232 Chr.10: 32015670-32015673 on Build GRCh38
rs1013908930 Chr.10: 32015318-32015318 on Build GRCh38
rs956451157 Chr.10: 32015381-32015381 on Build GRCh38
rs199676662 Chr.10: 32015675-32015675 on Build GRCh38
rs201380973 Chr.10: 32015555-32015555 on Build GRCh38
rs200653435 Chr.10: 32015663-32015663 on Build GRCh38
rs779191916 Chr.10: 32015545-32015545 on Build GRCh38
rs141695177 Chr.10: 32015643-32015643 on Build GRCh38
rs755872429 Chr.10: 32015522-32015522 on Build GRCh38
rs138770073 Chr.10: 32015600-32015600 on Build GRCh38
rs144552798 Chr.10: 32015651-32015651 on Build GRCh38
rs772124743 Chr.10: 32015558-32015558 on Build GRCh38
rs750217497 Chr.10: 32015608-32015608 on Build GRCh38
rs768173987 Chr.10: 32015340-32015343 on Build GRCh38
rs777033538 Chr.10: 32015677-32015677 on Build GRCh38
rs942287029 Chr.10: 32015583-32015583 on Build GRCh38
rs188811574 Chr.10: 32015346-32015346 on Build GRCh38
rs1029111777 Chr.10: 32015557-32015557 on Build GRCh38
rs755787052 Chr.10: 32015615-32015615 on Build GRCh38
rs758972432 Chr.10: 32015603-32015603 on Build GRCh38
rs895903322 Chr.10: 32015618-32015618 on Build GRCh38
rs375094831 Chr.10: 32015637-32015637 on Build GRCh38
rs780138182 Chr.10: 32015754-32015754 on Build GRCh38
rs569386112 Chr.10: 32015426-32015426 on Build GRCh38
rs749267836 Chr.10: 32015527-32015527 on Build GRCh38
rs112836367 Chr.10: 32015705-32015705 on Build GRCh38
rs756563235 Chr.10: 32015442-32015442 on Build GRCh38
rs374849264 Chr.10: 32015666-32015666 on Build GRCh38
rs993636339 Chr.10: 32015574-32015574 on Build GRCh38

View Full Product Details