Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 35011736-35012269 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUL2
Entrez Gene ID
8453
Gene Name
cullin 2
Gene Aliases
-
UniGene
Hs.82919
Gene Chromosome Location
Chr.10: 35008551-35126899 on Build GRCh38
Transcripts
NM_001198777.1 NM_001198778.1 NM_001198779.1 NM_001324375.1 NM_001324376.1 NM_003591.3 XM_011519743.1 XM_011519744.1 XM_011519745.1 XM_011519747.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5825634 c.2113C>T p.R705C Chr.10: 35011898-35011898 on Build GRCh38
4013944 c.2026C>T p.R676W Chr.10: 35011928-35011928 on Build GRCh38
4651787 c.2028G>A p.R676R Chr.10: 35011926-35011926 on Build GRCh38
1955902 c.2056C>T p.R686C Chr.10: 35011898-35011898 on Build GRCh38
917866 c.2089A>T p.N697Y Chr.10: 35011865-35011865 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146565928 Chr.10: 35012119-35012119 on Build GRCh38
rs935258082 Chr.10: 35011971-35011971 on Build GRCh38
rs761150131 Chr.10: 35011823-35011823 on Build GRCh38
rs528961582 Chr.10: 35011797-35011797 on Build GRCh38
rs776084154 Chr.10: 35011983-35011984 on Build GRCh38
rs1020449499 Chr.10: 35012072-35012072 on Build GRCh38
rs74712298 Chr.10: 35012232-35012232 on Build GRCh38
rs1018707768 Chr.10: 35011765-35011765 on Build GRCh38
rs149984910 Chr.10: 35011869-35011869 on Build GRCh38
rs1006361201 Chr.10: 35012109-35012109 on Build GRCh38
rs995938595 Chr.10: 35012045-35012045 on Build GRCh38
rs1003110026 Chr.10: 35012185-35012185 on Build GRCh38
rs775927225 Chr.10: 35011814-35011814 on Build GRCh38
rs921789085 Chr.10: 35011831-35011831 on Build GRCh38
rs754853022 Chr.10: 35011861-35011861 on Build GRCh38
rs144406968 Chr.10: 35012025-35012025 on Build GRCh38
rs934683661 Chr.10: 35011914-35011914 on Build GRCh38
rs752451742 Chr.10: 35011863-35011863 on Build GRCh38
rs772665111 Chr.10: 35011898-35011898 on Build GRCh38
rs1044441596 Chr.10: 35012055-35012055 on Build GRCh38
rs1050601644 Chr.10: 35012054-35012061 on Build GRCh38
rs1053026711 Chr.10: 35012011-35012011 on Build GRCh38
rs779075074 Chr.10: 35011881-35011881 on Build GRCh38
rs766070174 Chr.10: 35011962-35011962 on Build GRCh38
rs533771692 Chr.10: 35012153-35012153 on Build GRCh38
rs961992607 Chr.10: 35011762-35011762 on Build GRCh38
rs979241269 Chr.10: 35012243-35012243 on Build GRCh38
rs759079979 Chr.10: 35011978-35011978 on Build GRCh38
rs770287417 Chr.10: 35011968-35011969 on Build GRCh38
rs748186127 Chr.10: 35012131-35012131 on Build GRCh38
rs1040047558 Chr.10: 35011993-35011993 on Build GRCh38
rs184002650 Chr.10: 35011997-35011997 on Build GRCh38
rs995406259 Chr.10: 35012130-35012130 on Build GRCh38
rs777120601 Chr.10: 35011941-35011941 on Build GRCh38
rs901541834 Chr.10: 35012032-35012032 on Build GRCh38
rs773785894 Chr.10: 35011965-35011965 on Build GRCh38
rs777675857 Chr.10: 35011870-35011870 on Build GRCh38
rs776161868 Chr.10: 35011905-35011905 on Build GRCh38
rs568801813 Chr.10: 35011916-35011916 on Build GRCh38
rs73256993 Chr.10: 35011790-35011790 on Build GRCh38
rs751128379 Chr.10: 35011860-35011860 on Build GRCh38
rs752612909 Chr.10: 35011991-35011991 on Build GRCh38
rs541636598 Chr.10: 35011767-35011767 on Build GRCh38
rs567673098 Chr.10: 35012212-35012212 on Build GRCh38
rs767396435 Chr.10: 35011983-35011983 on Build GRCh38
rs115148655 Chr.10: 35012236-35012236 on Build GRCh38
rs765802200 Chr.10: 35011855-35011855 on Build GRCh38
rs373965981 Chr.10: 35011882-35011882 on Build GRCh38
rs397845751 Chr.10: 35012071-35012071 on Build GRCh38
rs772716457 Chr.10: 35011809-35011809 on Build GRCh38
rs958806392 Chr.10: 35011772-35011774 on Build GRCh38
rs771239317 Chr.10: 35011876-35011876 on Build GRCh38
rs967883460 Chr.10: 35012242-35012242 on Build GRCh38
rs5784438 Chr.10: 35012054-35012054 on Build GRCh38
rs1049326000 Chr.10: 35012141-35012141 on Build GRCh38
rs779288812 Chr.10: 35011945-35011945 on Build GRCh38
rs950167050 Chr.10: 35012053-35012053 on Build GRCh38
rs886649657 Chr.10: 35012197-35012197 on Build GRCh38
rs968265535 Chr.10: 35011839-35011839 on Build GRCh38
rs778685967 Chr.10: 35011940-35011940 on Build GRCh38
rs762707681 Chr.10: 35011840-35011840 on Build GRCh38
rs772266081 Chr.10: 35012182-35012182 on Build GRCh38
rs750020937 Chr.10: 35011833-35011833 on Build GRCh38
rs764893153 Chr.10: 35011827-35011827 on Build GRCh38
rs887918082 Chr.10: 35012087-35012087 on Build GRCh38
rs570409902 Chr.10: 35012226-35012226 on Build GRCh38
rs762500449 Chr.10: 35011943-35011943 on Build GRCh38
rs749443608 Chr.10: 35011871-35011871 on Build GRCh38
rs141320035 Chr.10: 35012210-35012210 on Build GRCh38
rs754850217 Chr.10: 35011872-35011872 on Build GRCh38
rs141060026 Chr.10: 35011903-35011903 on Build GRCh38
rs756121965 Chr.10: 35012002-35012002 on Build GRCh38

View Full Product Details