Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 37144675-37145176 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD30A
Entrez Gene ID
91074
Gene Name
ankyrin repeat domain 30A
Gene Aliases
NY-BR-1
UniGene
Hs.373787
Gene Chromosome Location
Chr.10: 37125754-37235831 on Build GRCh38
Transcripts
NM_052997.2 XM_011519757.2 XM_011519758.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3733811 c.1285C>G p.R429G Chr.10: 37145054-37145054 on Build GRCh38
4524250 c.1261G>A p.D421N Chr.10: 37145030-37145030 on Build GRCh38
5027208 c.1251A>G p.K417K Chr.10: 37145020-37145020 on Build GRCh38
3733809 c.1278T>G p.C426W Chr.10: 37145047-37145047 on Build GRCh38
3437540 c.1287G>A p.R429R Chr.10: 37145056-37145056 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769173020 Chr.10: 37145106-37145106 on Build GRCh38
rs72787523 Chr.10: 37145120-37145120 on Build GRCh38
rs1047046199 Chr.10: 37144813-37144813 on Build GRCh38
rs758243246 Chr.10: 37144962-37144962 on Build GRCh38
rs865883800 Chr.10: 37144888-37144888 on Build GRCh38
rs796408081 Chr.10: 37144897-37144897 on Build GRCh38
rs200478378 Chr.10: 37144971-37144971 on Build GRCh38
rs867286949 Chr.10: 37145027-37145027 on Build GRCh38
rs796178169 Chr.10: 37144895-37144895 on Build GRCh38
rs749506604 Chr.10: 37145074-37145074 on Build GRCh38
rs878978673 Chr.10: 37145035-37145035 on Build GRCh38
rs747174830 Chr.10: 37144947-37144949 on Build GRCh38
rs202194104 Chr.10: 37145075-37145076 on Build GRCh38
rs770982262 Chr.10: 37145081-37145081 on Build GRCh38
rs200164455 Chr.10: 37145055-37145055 on Build GRCh38
rs887390506 Chr.10: 37144779-37144779 on Build GRCh38
rs775746277 Chr.10: 37145150-37145150 on Build GRCh38
rs751480777 Chr.10: 37144964-37144964 on Build GRCh38
rs910434281 Chr.10: 37145059-37145059 on Build GRCh38
rs200974283 Chr.10: 37145054-37145054 on Build GRCh38
rs190697085 Chr.10: 37144738-37144738 on Build GRCh38
rs115413281 Chr.10: 37145149-37145149 on Build GRCh38
rs200315308 Chr.10: 37145047-37145047 on Build GRCh38
rs752549907 Chr.10: 37145062-37145062 on Build GRCh38
rs202023860 Chr.10: 37145098-37145098 on Build GRCh38
rs772495428 Chr.10: 37145011-37145011 on Build GRCh38
rs796989199 Chr.10: 37144904-37144904 on Build GRCh38
rs267602479 Chr.10: 37145012-37145012 on Build GRCh38
rs370818573 Chr.10: 37144967-37144967 on Build GRCh38
rs371611794 Chr.10: 37145089-37145089 on Build GRCh38
rs755971264 Chr.10: 37144747-37144750 on Build GRCh38
rs568629478 Chr.10: 37145118-37145118 on Build GRCh38
rs532590546 Chr.10: 37144912-37144912 on Build GRCh38
rs367822107 Chr.10: 37145064-37145064 on Build GRCh38
rs771314539 Chr.10: 37144996-37144996 on Build GRCh38
rs201385549 Chr.10: 37145097-37145097 on Build GRCh38
rs537729584 Chr.10: 37145144-37145144 on Build GRCh38
rs200031600 Chr.10: 37145060-37145060 on Build GRCh38
rs202149101 Chr.10: 37145001-37145001 on Build GRCh38
rs374001360 Chr.10: 37145057-37145057 on Build GRCh38
rs796136359 Chr.10: 37144905-37144905 on Build GRCh38
rs767794156 Chr.10: 37144945-37144945 on Build GRCh38
rs371191023 Chr.10: 37145071-37145071 on Build GRCh38
rs199603021 Chr.10: 37144976-37144976 on Build GRCh38
rs759312955 Chr.10: 37145022-37145022 on Build GRCh38
rs762483091 Chr.10: 37145049-37145049 on Build GRCh38
rs750007128 Chr.10: 37144959-37144959 on Build GRCh38
rs796330786 Chr.10: 37144867-37144867 on Build GRCh38
rs561793492 Chr.10: 37144771-37144771 on Build GRCh38
rs550628884 Chr.10: 37144789-37144789 on Build GRCh38
rs201289483 Chr.10: 37144975-37144975 on Build GRCh38
rs368316641 Chr.10: 37145032-37145032 on Build GRCh38
rs372822532 Chr.10: 37144954-37144954 on Build GRCh38
rs572202700 Chr.10: 37144842-37144842 on Build GRCh38
rs1047889176 Chr.10: 37145145-37145146 on Build GRCh38
rs563683797 Chr.10: 37144855-37144855 on Build GRCh38
rs200410911 Chr.10: 37144986-37144986 on Build GRCh38
rs764834907 Chr.10: 37144956-37144956 on Build GRCh38
rs759970739 Chr.10: 37145007-37145007 on Build GRCh38
rs375078975 Chr.10: 37145023-37145023 on Build GRCh38
rs73242779 Chr.10: 37144783-37144783 on Build GRCh38
rs182730096 Chr.10: 37144770-37144770 on Build GRCh38
rs72787522 Chr.10: 37144925-37144925 on Build GRCh38
rs548813357 Chr.10: 37145107-37145107 on Build GRCh38
rs780589149 Chr.10: 37144899-37144899 on Build GRCh38
rs552700946 Chr.10: 37144981-37144981 on Build GRCh38
rs929032632 Chr.10: 37145138-37145138 on Build GRCh38
rs770715522 Chr.10: 37144802-37144802 on Build GRCh38
rs1032404896 Chr.10: 37144926-37144926 on Build GRCh38
rs796912404 Chr.10: 37144889-37144889 on Build GRCh38
rs753481934 Chr.10: 37144950-37144950 on Build GRCh38

View Full Product Details