Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 38114498-38115003 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF37A
Entrez Gene ID
7587
Gene Name
zinc finger protein 37A
Gene Aliases
KOX21, ZNF37
UniGene
Hs.292575
Gene Chromosome Location
Chr.10: 38094327-38150293 on Build GRCh38
Transcripts
NM_001007094.2 NM_001178101.1 NM_001324245.1 NM_001324246.1 NM_001324247.1 NM_001324248.1 NM_001324249.1 NM_001324250.1 NM_001324251.1 NM_001324252.1 NM_001324253.1 NM_001324254.1 NM_001324256.1 NM_001324257.1 NM_001324258.1 NM_001324259.1 NM_001324260.1 NM_001324261.1 NM_001324262.1 NM_003421.2 XM_011519656.2 XM_011519657.2 XM_011519658.2 XM_017016618.1 XM_017016619.1 XM_017016620.1 XM_017016621.1 XM_017016622.1 XM_017016623.1 XM_017016624.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1180109 c.122A>C p.Y41S Chr.10: 38114861-38114861 on Build GRCh38
7247757 c.47G>T p.G16V Chr.10: 38114786-38114786 on Build GRCh38
6576768 c.73C>T p.L25L Chr.10: 38114812-38114812 on Build GRCh38
6065724 c.16-2A>G p.? Chr.10: 38114753-38114753 on Build GRCh38
4969582 c.47G>A p.G16D Chr.10: 38114786-38114786 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562663995 Chr.10: 38114721-38114721 on Build GRCh38
rs757197561 Chr.10: 38114629-38114629 on Build GRCh38
rs149074366 Chr.10: 38114790-38114790 on Build GRCh38
rs995454950 Chr.10: 38114880-38114880 on Build GRCh38
rs202054917 Chr.10: 38114923-38114923 on Build GRCh38
rs775814730 Chr.10: 38114820-38114820 on Build GRCh38
rs970870857 Chr.10: 38114618-38114618 on Build GRCh38
rs771770464 Chr.10: 38114818-38114818 on Build GRCh38
rs771346484 Chr.10: 38114877-38114880 on Build GRCh38
rs769070940 Chr.10: 38114933-38114933 on Build GRCh38
rs528035859 Chr.10: 38114860-38114860 on Build GRCh38
rs780762946 Chr.10: 38114752-38114752 on Build GRCh38
rs777231606 Chr.10: 38114891-38114891 on Build GRCh38
rs774082946 Chr.10: 38114655-38114655 on Build GRCh38
rs529848660 Chr.10: 38114626-38114626 on Build GRCh38
rs775475448 Chr.10: 38114922-38114922 on Build GRCh38
rs752294302 Chr.10: 38114887-38114887 on Build GRCh38
rs759318179 Chr.10: 38114808-38114808 on Build GRCh38
rs762226514 Chr.10: 38114830-38114830 on Build GRCh38
rs143951612 Chr.10: 38114821-38114821 on Build GRCh38
rs769249464 Chr.10: 38114766-38114766 on Build GRCh38
rs921977079 Chr.10: 38114673-38114673 on Build GRCh38
rs777286666 Chr.10: 38114772-38114772 on Build GRCh38
rs748895503 Chr.10: 38114892-38114892 on Build GRCh38
rs752681795 Chr.10: 38114746-38114747 on Build GRCh38
rs750903877 Chr.10: 38114852-38114852 on Build GRCh38
rs764301228 Chr.10: 38114724-38114724 on Build GRCh38
rs12766043 Chr.10: 38114613-38114613 on Build GRCh38
rs760951136 Chr.10: 38114929-38114929 on Build GRCh38
rs757835213 Chr.10: 38114739-38114739 on Build GRCh38
rs766039002 Chr.10: 38114704-38114704 on Build GRCh38
rs758718933 Chr.10: 38114853-38114853 on Build GRCh38
rs771945449 Chr.10: 38114913-38114913 on Build GRCh38
rs746313977 Chr.10: 38114746-38114746 on Build GRCh38
rs774251701 Chr.10: 38114705-38114705 on Build GRCh38
rs779119032 Chr.10: 38114742-38114742 on Build GRCh38
rs752983437 Chr.10: 38114720-38114720 on Build GRCh38
rs754351145 Chr.10: 38114736-38114736 on Build GRCh38
rs376611638 Chr.10: 38114824-38114824 on Build GRCh38
rs958311584 Chr.10: 38114650-38114650 on Build GRCh38
rs770760514 Chr.10: 38114898-38114898 on Build GRCh38
rs551788907 Chr.10: 38114765-38114765 on Build GRCh38
rs745823188 Chr.10: 38114904-38114904 on Build GRCh38
rs149919662 Chr.10: 38114541-38114541 on Build GRCh38
rs781046299 Chr.10: 38114671-38114671 on Build GRCh38
rs779403288 Chr.10: 38114719-38114720 on Build GRCh38
rs1024595077 Chr.10: 38114939-38114939 on Build GRCh38
rs113864358 Chr.10: 38114710-38114710 on Build GRCh38
rs371950324 Chr.10: 38114706-38114706 on Build GRCh38
rs780425673 Chr.10: 38114871-38114871 on Build GRCh38
rs978271144 Chr.10: 38114624-38114624 on Build GRCh38
rs747226349 Chr.10: 38114838-38114839 on Build GRCh38
rs199622384 Chr.10: 38114843-38114843 on Build GRCh38
rs531562323 Chr.10: 38114751-38114751 on Build GRCh38
rs755811873 Chr.10: 38114889-38114889 on Build GRCh38
rs749159052 Chr.10: 38114788-38114788 on Build GRCh38
rs758530431 Chr.10: 38114787-38114795 on Build GRCh38
rs894818012 Chr.10: 38114902-38114902 on Build GRCh38
rs774011581 Chr.10: 38114802-38114802 on Build GRCh38
rs777769001 Chr.10: 38114820-38114820 on Build GRCh38
rs764250268 Chr.10: 38114822-38114822 on Build GRCh38
rs1024645469 Chr.10: 38114582-38114582 on Build GRCh38
rs778843904 Chr.10: 38114901-38114901 on Build GRCh38

View Full Product Details