Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 42786910-42787458 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMS1
Entrez Gene ID
9790
Gene Name
BMS1, ribosome biogenesis factor
Gene Aliases
ACC, BMS1L
UniGene
Hs.10848
Gene Chromosome Location
Chr.10: 42782506-42834937 on Build GRCh38
Transcripts
NM_014753.3 XM_005271846.3 XM_005271848.3 XM_005271849.3 XM_011540402.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
918084 c.376C>T p.R126C Chr.10: 42787176-42787176 on Build GRCh38
259169 c.383T>G p.L128R Chr.10: 42787183-42787183 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs142984592 Chr.10: 42787380-42787380 on Build GRCh38
rs184477582 Chr.10: 42787371-42787371 on Build GRCh38
rs17429415 Chr.10: 42787419-42787419 on Build GRCh38
rs17159071 Chr.10: 42787374-42787374 on Build GRCh38
rs867586150 Chr.10: 42786962-42786962 on Build GRCh38
rs916334894 Chr.10: 42787206-42787206 on Build GRCh38
rs927621326 Chr.10: 42787293-42787293 on Build GRCh38
rs777296580 Chr.10: 42787340-42787342 on Build GRCh38
rs756531858 Chr.10: 42787184-42787184 on Build GRCh38
rs780540295 Chr.10: 42787174-42787175 on Build GRCh38
rs763067853 Chr.10: 42787328-42787328 on Build GRCh38
rs774809970 Chr.10: 42787236-42787236 on Build GRCh38
rs192388236 Chr.10: 42786971-42786971 on Build GRCh38
rs191919925 Chr.10: 42787114-42787114 on Build GRCh38
rs972624226 Chr.10: 42787120-42787120 on Build GRCh38
rs746538548 Chr.10: 42787340-42787340 on Build GRCh38
rs763036596 Chr.10: 42787267-42787267 on Build GRCh38
rs781697087 Chr.10: 42787178-42787178 on Build GRCh38
rs765532200 Chr.10: 42787165-42787165 on Build GRCh38
rs561943045 Chr.10: 42787118-42787118 on Build GRCh38
rs11239772 Chr.10: 42787080-42787080 on Build GRCh38
rs564212508 Chr.10: 42787084-42787084 on Build GRCh38
rs776525557 Chr.10: 42787147-42787147 on Build GRCh38
rs766314874 Chr.10: 42787174-42787174 on Build GRCh38
rs531447303 Chr.10: 42787102-42787102 on Build GRCh38
rs542205059 Chr.10: 42787334-42787334 on Build GRCh38
rs10900269 Chr.10: 42787315-42787315 on Build GRCh38
rs918462638 Chr.10: 42787142-42787142 on Build GRCh38
rs761685443 Chr.10: 42787256-42787256 on Build GRCh38
rs574221530 Chr.10: 42786981-42786981 on Build GRCh38
rs189317234 Chr.10: 42787415-42787415 on Build GRCh38
rs570005174 Chr.10: 42787305-42787305 on Build GRCh38
rs769777591 Chr.10: 42787261-42787261 on Build GRCh38
rs771767050 Chr.10: 42787227-42787227 on Build GRCh38
rs750730619 Chr.10: 42787171-42787171 on Build GRCh38
rs778536019 Chr.10: 42787188-42787188 on Build GRCh38
rs1021706419 Chr.10: 42787050-42787050 on Build GRCh38
rs565731481 Chr.10: 42787242-42787242 on Build GRCh38
rs938960611 Chr.10: 42787314-42787314 on Build GRCh38
rs555929768 Chr.10: 42786980-42786980 on Build GRCh38
rs1003051329 Chr.10: 42786967-42786967 on Build GRCh38
rs761740448 Chr.10: 42787164-42787164 on Build GRCh38
rs773156244 Chr.10: 42787262-42787262 on Build GRCh38
rs891948819 Chr.10: 42787033-42787033 on Build GRCh38
rs766655500 Chr.10: 42787274-42787274 on Build GRCh38
rs755400315 Chr.10: 42787177-42787177 on Build GRCh38
rs533191255 Chr.10: 42787254-42787254 on Build GRCh38
rs551370809 Chr.10: 42787276-42787276 on Build GRCh38
rs547314695 Chr.10: 42787202-42787202 on Build GRCh38
rs529159115 Chr.10: 42787176-42787176 on Build GRCh38
rs545911521 Chr.10: 42787083-42787083 on Build GRCh38
rs73252895 Chr.10: 42787048-42787048 on Build GRCh38
rs907484345 Chr.10: 42786938-42786938 on Build GRCh38
rs748493682 Chr.10: 42787182-42787182 on Build GRCh38
rs1019648041 Chr.10: 42787396-42787396 on Build GRCh38
rs1019595836 Chr.10: 42787101-42787101 on Build GRCh38
rs1011757050 Chr.10: 42787363-42787363 on Build GRCh38
rs188002244 Chr.10: 42787081-42787081 on Build GRCh38
rs758651801 Chr.10: 42787172-42787172 on Build GRCh38

View Full Product Details