Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 43195880-43196414 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RASGEF1A
Entrez Gene ID
221002
Gene Name
RasGEF domain family member 1A
Gene Aliases
CG4853
UniGene
Hs.125293
Gene Chromosome Location
Chr.10: 43194533-43266919 on Build GRCh38
Transcripts
NM_001282862.1 NM_145313.3 XM_005271809.3 XM_011539500.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6216986 c.1263-9C>A p.? Chr.10: 43196277-43196277 on Build GRCh38
6216985 c.1422-9C>A p.? Chr.10: 43196277-43196277 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs906114917 Chr.10: 43196378-43196378 on Build GRCh38
rs963583398 Chr.10: 43196241-43196241 on Build GRCh38
rs143196643 Chr.10: 43196259-43196259 on Build GRCh38
rs534085953 Chr.10: 43196051-43196051 on Build GRCh38
rs184243450 Chr.10: 43195957-43195957 on Build GRCh38
rs947575563 Chr.10: 43196344-43196344 on Build GRCh38
rs370896202 Chr.10: 43196230-43196230 on Build GRCh38
rs529049640 Chr.10: 43196029-43196029 on Build GRCh38
rs367800733 Chr.10: 43196283-43196283 on Build GRCh38
rs750597019 Chr.10: 43196302-43196302 on Build GRCh38
rs760645333 Chr.10: 43196282-43196282 on Build GRCh38
rs757219745 Chr.10: 43196119-43196119 on Build GRCh38
rs1254968 Chr.10: 43196243-43196243 on Build GRCh38
rs111686790 Chr.10: 43196240-43196240 on Build GRCh38
rs377261048 Chr.10: 43196281-43196281 on Build GRCh38
rs139599170 Chr.10: 43196181-43196181 on Build GRCh38
rs758482277 Chr.10: 43196242-43196242 on Build GRCh38
rs771995687 Chr.10: 43196376-43196376 on Build GRCh38
rs770523436 Chr.10: 43196257-43196257 on Build GRCh38
rs566822195 Chr.10: 43196041-43196041 on Build GRCh38
rs958193263 Chr.10: 43196293-43196293 on Build GRCh38
rs1043741033 Chr.10: 43196182-43196182 on Build GRCh38
rs751669954 Chr.10: 43196312-43196312 on Build GRCh38
rs769205848 Chr.10: 43196206-43196206 on Build GRCh38
rs550696851 Chr.10: 43196348-43196348 on Build GRCh38
rs1051880077 Chr.10: 43196064-43196065 on Build GRCh38
rs780661592 Chr.10: 43196200-43196200 on Build GRCh38
rs542449479 Chr.10: 43196248-43196248 on Build GRCh38
rs568688652 Chr.10: 43195986-43195986 on Build GRCh38
rs558852608 Chr.10: 43196164-43196164 on Build GRCh38
rs963231901 Chr.10: 43195920-43195920 on Build GRCh38
rs978012686 Chr.10: 43195965-43195966 on Build GRCh38
rs765379171 Chr.10: 43196292-43196292 on Build GRCh38
rs201520636 Chr.10: 43196225-43196225 on Build GRCh38
rs763942490 Chr.10: 43196229-43196229 on Build GRCh38
rs752574052 Chr.10: 43196228-43196228 on Build GRCh38
rs907964970 Chr.10: 43196377-43196377 on Build GRCh38
rs747589189 Chr.10: 43196245-43196245 on Build GRCh38
rs944102502 Chr.10: 43196063-43196063 on Build GRCh38
rs375828754 Chr.10: 43196303-43196303 on Build GRCh38
rs777118344 Chr.10: 43196210-43196210 on Build GRCh38
rs959112755 Chr.10: 43195903-43195903 on Build GRCh38
rs912649602 Chr.10: 43196019-43196019 on Build GRCh38
rs78147207 Chr.10: 43195942-43195942 on Build GRCh38
rs940660824 Chr.10: 43196389-43196389 on Build GRCh38
rs548395547 Chr.10: 43196035-43196035 on Build GRCh38
rs1049683446 Chr.10: 43195991-43195991 on Build GRCh38
rs753595689 Chr.10: 43196285-43196285 on Build GRCh38
rs771628766 Chr.10: 43196275-43196275 on Build GRCh38
rs542466363 Chr.10: 43196224-43196224 on Build GRCh38
rs933386672 Chr.10: 43196025-43196025 on Build GRCh38
rs767447525 Chr.10: 43196227-43196227 on Build GRCh38
rs368536671 Chr.10: 43196311-43196311 on Build GRCh38
rs921803945 Chr.10: 43196024-43196024 on Build GRCh38
rs371049913 Chr.10: 43196211-43196211 on Build GRCh38
rs372426488 Chr.10: 43196205-43196205 on Build GRCh38
rs773105795 Chr.10: 43196304-43196323 on Build GRCh38
rs936484431 Chr.10: 43196005-43196005 on Build GRCh38
rs376421107 Chr.10: 43196008-43196008 on Build GRCh38
rs372453969 Chr.10: 43196077-43196077 on Build GRCh38
rs188454909 Chr.10: 43196089-43196089 on Build GRCh38
rs1053229539 Chr.10: 43196076-43196076 on Build GRCh38
rs371694944 Chr.10: 43196296-43196296 on Build GRCh38
rs781461991 Chr.10: 43196251-43196251 on Build GRCh38
rs758693848 Chr.10: 43196309-43196309 on Build GRCh38
rs913311874 Chr.10: 43196332-43196332 on Build GRCh38
rs1001830504 Chr.10: 43196216-43196216 on Build GRCh38
rs530900719 Chr.10: 43196313-43196313 on Build GRCh38
rs966371229 Chr.10: 43195943-43195943 on Build GRCh38
rs745931879 Chr.10: 43196198-43196198 on Build GRCh38
rs973753375 Chr.10: 43195938-43195938 on Build GRCh38
rs1004665771 Chr.10: 43196236-43196236 on Build GRCh38
rs779145141 Chr.10: 43196234-43196234 on Build GRCh38
rs925205516 Chr.10: 43195969-43195969 on Build GRCh38
rs1029333792 Chr.10: 43196272-43196272 on Build GRCh38
rs761625278 Chr.10: 43196289-43196289 on Build GRCh38
rs781586427 Chr.10: 43195954-43195954 on Build GRCh38
rs748498930 Chr.10: 43196252-43196252 on Build GRCh38

View Full Product Details