Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 43199538-43200034 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RASGEF1A
Entrez Gene ID
221002
Gene Name
RasGEF domain family member 1A
Gene Aliases
CG4853
UniGene
Hs.125293
Gene Chromosome Location
Chr.10: 43194533-43266919 on Build GRCh38
Transcripts
NM_001282862.1 NM_145313.3 XM_005271809.3 XM_011539500.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6899411 c.780C>T p.T260T Chr.10: 43199745-43199745 on Build GRCh38
7137524 c.767A>G p.D256G Chr.10: 43199758-43199758 on Build GRCh38
4753946 c.761G>A p.R254Q Chr.10: 43199764-43199764 on Build GRCh38
6763179 c.633G>A p.E211E Chr.10: 43199733-43199733 on Build GRCh38
4769566 c.612G>A p.L204L Chr.10: 43199754-43199754 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138086250 Chr.10: 43199832-43199832 on Build GRCh38
rs998030710 Chr.10: 43199936-43199936 on Build GRCh38
rs41301581 Chr.10: 43199664-43199664 on Build GRCh38
rs1046044120 Chr.10: 43199581-43199581 on Build GRCh38
rs965223704 Chr.10: 43199610-43199610 on Build GRCh38
rs775439278 Chr.10: 43199667-43199667 on Build GRCh38
rs374403441 Chr.10: 43199760-43199760 on Build GRCh38
rs756936174 Chr.10: 43199754-43199754 on Build GRCh38
rs189494581 Chr.10: 43199787-43199787 on Build GRCh38
rs773676627 Chr.10: 43199592-43199592 on Build GRCh38
rs1025936748 Chr.10: 43199937-43199937 on Build GRCh38
rs977210606 Chr.10: 43199624-43199624 on Build GRCh38
rs748412258 Chr.10: 43199746-43199746 on Build GRCh38
rs965171655 Chr.10: 43199930-43199934 on Build GRCh38
rs540807592 Chr.10: 43199894-43199894 on Build GRCh38
rs575901432 Chr.10: 43199831-43199831 on Build GRCh38
rs549159403 Chr.10: 43199634-43199634 on Build GRCh38
rs552886057 Chr.10: 43199900-43199900 on Build GRCh38
rs368852111 Chr.10: 43199652-43199652 on Build GRCh38
rs776503810 Chr.10: 43199809-43199809 on Build GRCh38
rs771626771 Chr.10: 43199780-43199780 on Build GRCh38
rs745393909 Chr.10: 43199764-43199764 on Build GRCh38
rs747007195 Chr.10: 43199797-43199797 on Build GRCh38
rs573524937 Chr.10: 43199893-43199893 on Build GRCh38
rs755412351 Chr.10: 43199735-43199735 on Build GRCh38
rs1047073780 Chr.10: 43199901-43199901 on Build GRCh38
rs1254964 Chr.10: 43199601-43199601 on Build GRCh38
rs201765175 Chr.10: 43199745-43199745 on Build GRCh38
rs766636567 Chr.10: 43199659-43199659 on Build GRCh38
rs766555953 Chr.10: 43199712-43199712 on Build GRCh38
rs904221156 Chr.10: 43199873-43199873 on Build GRCh38
rs765505444 Chr.10: 43199694-43199694 on Build GRCh38
rs762139835 Chr.10: 43199682-43199682 on Build GRCh38
rs947707195 Chr.10: 43199579-43199579 on Build GRCh38
rs555013287 Chr.10: 43199874-43199874 on Build GRCh38
rs376128525 Chr.10: 43199657-43199657 on Build GRCh38
rs1006847897 Chr.10: 43199581-43199582 on Build GRCh38
rs747351138 Chr.10: 43199632-43199632 on Build GRCh38
rs150387348 Chr.10: 43199564-43199564 on Build GRCh38
rs1010442338 Chr.10: 43199838-43199838 on Build GRCh38
rs185837886 Chr.10: 43199635-43199635 on Build GRCh38
rs776255974 Chr.10: 43199679-43199679 on Build GRCh38
rs747520896 Chr.10: 43199664-43199665 on Build GRCh38
rs939548687 Chr.10: 43199742-43199742 on Build GRCh38
rs760516252 Chr.10: 43199668-43199668 on Build GRCh38
rs373617588 Chr.10: 43199626-43199626 on Build GRCh38
rs758497018 Chr.10: 43199699-43199699 on Build GRCh38
rs989612572 Chr.10: 43199871-43199871 on Build GRCh38
rs768452835 Chr.10: 43199804-43199804 on Build GRCh38
rs767971155 Chr.10: 43199648-43199648 on Build GRCh38
rs968621773 Chr.10: 43199580-43199580 on Build GRCh38
rs139262265 Chr.10: 43199697-43199697 on Build GRCh38
rs374472625 Chr.10: 43199658-43199658 on Build GRCh38
rs752179523 Chr.10: 43199728-43199728 on Build GRCh38
rs76306135 Chr.10: 43199665-43199665 on Build GRCh38
rs764016510 Chr.10: 43199677-43199677 on Build GRCh38
rs927678224 Chr.10: 43200005-43200005 on Build GRCh38
rs749010259 Chr.10: 43199656-43199656 on Build GRCh38

View Full Product Details