Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 43606323-43606816 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF485
Entrez Gene ID
220992
Gene Name
zinc finger protein 485
Gene Aliases
-
UniGene
Hs.147440
Gene Chromosome Location
Chr.10: 43606407-43620173 on Build GRCh38
Transcripts
NM_001318140.1 NM_001318141.1 NM_001318142.1 NM_001318143.1 NM_145312.3

SNP Detail

SNP ID Chromosome Location
rs1027696587 Chr.10: 43606455-43606455 on Build GRCh38
rs984554830 Chr.10: 43606427-43606427 on Build GRCh38
rs12261773 Chr.10: 43606436-43606436 on Build GRCh38
rs754422421 Chr.10: 43606356-43606356 on Build GRCh38
rs944138450 Chr.10: 43606768-43606768 on Build GRCh38
rs1007819696 Chr.10: 43606459-43606459 on Build GRCh38
rs913631471 Chr.10: 43606367-43606367 on Build GRCh38
rs947412827 Chr.10: 43606463-43606463 on Build GRCh38
rs981580779 Chr.10: 43606644-43606644 on Build GRCh38
rs989072558 Chr.10: 43606700-43606700 on Build GRCh38
rs975818040 Chr.10: 43606366-43606366 on Build GRCh38
rs758986976 Chr.10: 43606484-43606484 on Build GRCh38
rs911283006 Chr.10: 43606751-43606751 on Build GRCh38
rs993983743 Chr.10: 43606454-43606454 on Build GRCh38
rs113735272 Chr.10: 43606612-43606612 on Build GRCh38
rs929411642 Chr.10: 43606715-43606715 on Build GRCh38
rs889876039 Chr.10: 43606485-43606485 on Build GRCh38
rs772835883 Chr.10: 43606420-43606420 on Build GRCh38
rs979587818 Chr.10: 43606529-43606529 on Build GRCh38
rs541598927 Chr.10: 43606468-43606468 on Build GRCh38
rs12261830 Chr.10: 43606465-43606465 on Build GRCh38
rs1055412526 Chr.10: 43606443-43606443 on Build GRCh38
rs901738675 Chr.10: 43606530-43606530 on Build GRCh38
rs917814172 Chr.10: 43606610-43606610 on Build GRCh38
rs1017731692 Chr.10: 43606350-43606350 on Build GRCh38
rs1046546363 Chr.10: 43606646-43606646 on Build GRCh38
rs865909826 Chr.10: 43606571-43606571 on Build GRCh38
rs56333360 Chr.10: 43606460-43606460 on Build GRCh38
rs966201518 Chr.10: 43606461-43606461 on Build GRCh38
rs949262630 Chr.10: 43606611-43606611 on Build GRCh38
rs554263526 Chr.10: 43606481-43606481 on Build GRCh38
rs537064926 Chr.10: 43606458-43606458 on Build GRCh38
rs945102354 Chr.10: 43606371-43606371 on Build GRCh38
rs1041138595 Chr.10: 43606777-43606777 on Build GRCh38
rs559737068 Chr.10: 43606518-43606518 on Build GRCh38
rs1025992060 Chr.10: 43606402-43606402 on Build GRCh38
rs979149106 Chr.10: 43606429-43606429 on Build GRCh38
rs765732142 Chr.10: 43606540-43606540 on Build GRCh38
rs924932651 Chr.10: 43606431-43606431 on Build GRCh38
rs1045866242 Chr.10: 43606452-43606452 on Build GRCh38
rs557831081 Chr.10: 43606357-43606357 on Build GRCh38
rs372436467 Chr.10: 43606547-43606547 on Build GRCh38
rs949169551 Chr.10: 43606451-43606451 on Build GRCh38
rs868227374 Chr.10: 43606650-43606650 on Build GRCh38
rs1021065449 Chr.10: 43606509-43606509 on Build GRCh38
rs140661694 Chr.10: 43606695-43606695 on Build GRCh38
rs896326449 Chr.10: 43606447-43606447 on Build GRCh38
rs985867224 Chr.10: 43606747-43606747 on Build GRCh38
rs1035848747 Chr.10: 43606679-43606679 on Build GRCh38
rs990747261 Chr.10: 43606609-43606609 on Build GRCh38
rs947373212 Chr.10: 43606731-43606731 on Build GRCh38
rs897933729 Chr.10: 43606453-43606453 on Build GRCh38
rs1007308110 Chr.10: 43606492-43606492 on Build GRCh38
rs1049580998 Chr.10: 43606780-43606780 on Build GRCh38

View Full Product Details