Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 68934684-68935217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX50
Entrez Gene ID
79009
Gene Name
DEAD-box helicase 50
Gene Aliases
GU2, GUB, RH-II/GuB, mcdrh
UniGene
Hs.522984
Gene Chromosome Location
Chr.10: 68901278-68956312 on Build GRCh38
Transcripts
NM_024045.1 XM_005270148.2 XM_011540144.2 XM_017016626.1 XM_017016627.1 XM_017016628.1 XM_017016629.1 XM_017016630.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
280343 c.1521+10G>T p.? Chr.10: 68934928-68934928 on Build GRCh38
4676863 c.1433G>A p.R478H Chr.10: 68934830-68934830 on Build GRCh38
5370995 c.1445C>T p.A482V Chr.10: 68934842-68934842 on Build GRCh38
4626680 c.1453A>G p.T485A Chr.10: 68934850-68934850 on Build GRCh38
274764 c.1424G>A p.R475H Chr.10: 68934821-68934821 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs574011544 Chr.10: 68934972-68934972 on Build GRCh38
rs368087423 Chr.10: 68934888-68934888 on Build GRCh38
rs758216487 Chr.10: 68934847-68934847 on Build GRCh38
rs538435431 Chr.10: 68934743-68934743 on Build GRCh38
rs768101576 Chr.10: 68935067-68935067 on Build GRCh38
rs113487970 Chr.10: 68935158-68935158 on Build GRCh38
rs115233762 Chr.10: 68934744-68934744 on Build GRCh38
rs549071330 Chr.10: 68935027-68935027 on Build GRCh38
rs1002628437 Chr.10: 68935119-68935119 on Build GRCh38
rs576110195 Chr.10: 68935168-68935168 on Build GRCh38
rs780015420 Chr.10: 68934862-68934862 on Build GRCh38
rs1056796830 Chr.10: 68935155-68935155 on Build GRCh38
rs773863221 Chr.10: 68935101-68935101 on Build GRCh38
rs187306208 Chr.10: 68934711-68934711 on Build GRCh38
rs751919136 Chr.10: 68934781-68934781 on Build GRCh38
rs769326738 Chr.10: 68934917-68934917 on Build GRCh38
rs759878528 Chr.10: 68934960-68934960 on Build GRCh38
rs777929772 Chr.10: 68934830-68934830 on Build GRCh38
rs1002574400 Chr.10: 68934722-68934722 on Build GRCh38
rs200806264 Chr.10: 68934766-68934766 on Build GRCh38
rs140372460 Chr.10: 68934966-68934966 on Build GRCh38
rs771386125 Chr.10: 68934949-68934949 on Build GRCh38
rs762503562 Chr.10: 68934940-68934940 on Build GRCh38
rs1021384019 Chr.10: 68935009-68935009 on Build GRCh38
rs1015312874 Chr.10: 68935147-68935147 on Build GRCh38
rs906831701 Chr.10: 68934721-68934721 on Build GRCh38
rs1012901654 Chr.10: 68935179-68935179 on Build GRCh38
rs1050393034 Chr.10: 68934985-68934985 on Build GRCh38
rs772662134 Chr.10: 68934929-68934929 on Build GRCh38
rs931870813 Chr.10: 68934970-68934970 on Build GRCh38
rs375122404 Chr.10: 68934829-68934829 on Build GRCh38
rs774921917 Chr.10: 68934959-68934959 on Build GRCh38
rs766788482 Chr.10: 68934779-68934779 on Build GRCh38
rs556470068 Chr.10: 68935097-68935097 on Build GRCh38
rs914372688 Chr.10: 68935025-68935025 on Build GRCh38
rs761377520 Chr.10: 68934763-68934763 on Build GRCh38
rs531070735 Chr.10: 68934804-68934804 on Build GRCh38
rs763041041 Chr.10: 68934777-68934777 on Build GRCh38
rs952268507 Chr.10: 68934783-68934783 on Build GRCh38
rs542546667 Chr.10: 68934975-68934975 on Build GRCh38
rs987396580 Chr.10: 68935144-68935144 on Build GRCh38
rs747738922 Chr.10: 68934903-68934903 on Build GRCh38
rs534152345 Chr.10: 68934771-68934771 on Build GRCh38
rs780877784 Chr.10: 68934897-68934897 on Build GRCh38
rs976014929 Chr.10: 68934944-68934944 on Build GRCh38
rs776234864 Chr.10: 68934761-68934761 on Build GRCh38
rs565193305 Chr.10: 68934792-68934792 on Build GRCh38
rs746818108 Chr.10: 68934874-68934874 on Build GRCh38
rs370419447 Chr.10: 68934821-68934821 on Build GRCh38
rs371119398 Chr.10: 68934894-68934894 on Build GRCh38
rs750403932 Chr.10: 68934838-68934838 on Build GRCh38

View Full Product Details