Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 70422360-70422868 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4EBP2
Entrez Gene ID
1979
Gene Name
eukaryotic translation initiation factor 4E binding protein 2
Gene Aliases
4EBP2, PHASII
UniGene
Hs.621200
Gene Chromosome Location
Chr.10: 70404105-70428618 on Build GRCh38
Transcripts
NM_004096.4

SNP Detail

SNP ID Chromosome Location
rs982769646 Chr.10: 70422791-70422791 on Build GRCh38
rs529984342 Chr.10: 70422521-70422521 on Build GRCh38
rs887182807 Chr.10: 70422526-70422526 on Build GRCh38
rs772108235 Chr.10: 70422585-70422585 on Build GRCh38
rs941429251 Chr.10: 70422528-70422528 on Build GRCh38
rs369685497 Chr.10: 70422716-70422716 on Build GRCh38
rs976037108 Chr.10: 70422498-70422498 on Build GRCh38
rs923207706 Chr.10: 70422510-70422510 on Build GRCh38
rs929248728 Chr.10: 70422767-70422767 on Build GRCh38
rs976343837 Chr.10: 70422744-70422744 on Build GRCh38
rs957154267 Chr.10: 70422411-70422411 on Build GRCh38
rs1013532269 Chr.10: 70422561-70422561 on Build GRCh38
rs957077498 Chr.10: 70422633-70422633 on Build GRCh38
rs1038451395 Chr.10: 70422836-70422836 on Build GRCh38
rs372647128 Chr.10: 70422629-70422629 on Build GRCh38
rs760547705 Chr.10: 70422631-70422631 on Build GRCh38
rs989909860 Chr.10: 70422418-70422418 on Build GRCh38
rs933988159 Chr.10: 70422511-70422511 on Build GRCh38
rs540369035 Chr.10: 70422456-70422456 on Build GRCh38
rs941398097 Chr.10: 70422807-70422807 on Build GRCh38
rs1031419390 Chr.10: 70422401-70422401 on Build GRCh38
rs908563502 Chr.10: 70422806-70422806 on Build GRCh38
rs180997209 Chr.10: 70422533-70422533 on Build GRCh38
rs375966357 Chr.10: 70422713-70422713 on Build GRCh38
rs542132313 Chr.10: 70422524-70422524 on Build GRCh38
rs964499873 Chr.10: 70422736-70422736 on Build GRCh38
rs549736568 Chr.10: 70422814-70422814 on Build GRCh38
rs923210281 Chr.10: 70422745-70422745 on Build GRCh38
rs1024543509 Chr.10: 70422576-70422576 on Build GRCh38
rs573462638 Chr.10: 70422442-70422442 on Build GRCh38
rs998526513 Chr.10: 70422620-70422620 on Build GRCh38
rs943264414 Chr.10: 70422470-70422470 on Build GRCh38
rs117563909 Chr.10: 70422396-70422396 on Build GRCh38
rs144402234 Chr.10: 70422556-70422556 on Build GRCh38
rs41277986 Chr.10: 70422613-70422613 on Build GRCh38
rs185997701 Chr.10: 70422671-70422671 on Build GRCh38
rs894660323 Chr.10: 70422560-70422560 on Build GRCh38

View Full Product Details