Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 70432645-70433148 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NODAL
Entrez Gene ID
4838
Gene Name
nodal growth differentiation factor
Gene Aliases
HTX5
UniGene
Hs.370414
Gene Chromosome Location
Chr.10: 70431936-70441709 on Build GRCh38
Transcripts
NM_018055.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7226684 c.1020C>G p.I340M Chr.10: 70432960-70432960 on Build GRCh38
5559235 c.898C>T p.L300L Chr.10: 70433082-70433082 on Build GRCh38
6432886 c.905G>T p.R302L Chr.10: 70433075-70433075 on Build GRCh38
7165273 c.941C>T p.A314V Chr.10: 70433039-70433039 on Build GRCh38
195595 c.960G>A p.P320P Chr.10: 70433020-70433020 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs898275148 Chr.10: 70432864-70432864 on Build GRCh38
rs142286836 Chr.10: 70432960-70432960 on Build GRCh38
rs749630247 Chr.10: 70432994-70432994 on Build GRCh38
rs1019007755 Chr.10: 70432750-70432750 on Build GRCh38
rs769824580 Chr.10: 70433026-70433026 on Build GRCh38
rs1021244749 Chr.10: 70432946-70432946 on Build GRCh38
rs148708629 Chr.10: 70433064-70433064 on Build GRCh38
rs528710830 Chr.10: 70433119-70433119 on Build GRCh38
rs150819707 Chr.10: 70433076-70433076 on Build GRCh38
rs755880329 Chr.10: 70432971-70432971 on Build GRCh38
rs746033726 Chr.10: 70433012-70433012 on Build GRCh38
rs756658835 Chr.10: 70433079-70433079 on Build GRCh38
rs748622757 Chr.10: 70432980-70432980 on Build GRCh38
rs952335495 Chr.10: 70432827-70432827 on Build GRCh38
rs1004084139 Chr.10: 70432846-70432846 on Build GRCh38
rs771199897 Chr.10: 70433001-70433001 on Build GRCh38
rs966160234 Chr.10: 70432783-70432784 on Build GRCh38
rs772060498 Chr.10: 70433017-70433017 on Build GRCh38
rs750203271 Chr.10: 70432921-70432921 on Build GRCh38
rs12269360 Chr.10: 70432809-70432809 on Build GRCh38
rs1045656582 Chr.10: 70432812-70432812 on Build GRCh38
rs369691106 Chr.10: 70432915-70432915 on Build GRCh38
rs578069296 Chr.10: 70432815-70432815 on Build GRCh38
rs536116678 Chr.10: 70432789-70432789 on Build GRCh38
rs773193349 Chr.10: 70433048-70433048 on Build GRCh38
rs553221767 Chr.10: 70432784-70432784 on Build GRCh38
rs778239524 Chr.10: 70433109-70433109 on Build GRCh38
rs778147685 Chr.10: 70432992-70432992 on Build GRCh38
rs767265637 Chr.10: 70432714-70432714 on Build GRCh38
rs758165167 Chr.10: 70432932-70432932 on Build GRCh38
rs949978867 Chr.10: 70432741-70432741 on Build GRCh38
rs768705222 Chr.10: 70433044-70433050 on Build GRCh38
rs1027919358 Chr.10: 70432871-70432871 on Build GRCh38
rs761753832 Chr.10: 70432909-70432909 on Build GRCh38
rs751176757 Chr.10: 70432959-70432959 on Build GRCh38
rs959419966 Chr.10: 70433020-70433020 on Build GRCh38
rs1016626408 Chr.10: 70432863-70432863 on Build GRCh38
rs765147574 Chr.10: 70432911-70432911 on Build GRCh38
rs113255026 Chr.10: 70433005-70433005 on Build GRCh38
rs774089644 Chr.10: 70432870-70432870 on Build GRCh38
rs757620898 Chr.10: 70433120-70433120 on Build GRCh38
rs985003990 Chr.10: 70432854-70432854 on Build GRCh38
rs949415691 Chr.10: 70433061-70433061 on Build GRCh38
rs535102533 Chr.10: 70432691-70432691 on Build GRCh38
rs987421678 Chr.10: 70432941-70432941 on Build GRCh38
rs372678936 Chr.10: 70433115-70433115 on Build GRCh38
rs750214155 Chr.10: 70433075-70433075 on Build GRCh38
rs112261582 Chr.10: 70432737-70432737 on Build GRCh38
rs188029435 Chr.10: 70433056-70433056 on Build GRCh38
rs200946243 Chr.10: 70432968-70432968 on Build GRCh38
rs143903715 Chr.10: 70433008-70433008 on Build GRCh38
rs1007600456 Chr.10: 70432729-70432729 on Build GRCh38
rs768727728 Chr.10: 70432818-70432818 on Build GRCh38
rs76889088 Chr.10: 70432874-70432874 on Build GRCh38
rs774499735 Chr.10: 70433090-70433091 on Build GRCh38
rs766110361 Chr.10: 70432957-70432957 on Build GRCh38
rs953356625 Chr.10: 70432718-70432720 on Build GRCh38
rs200233027 Chr.10: 70433117-70433117 on Build GRCh38
rs761688803 Chr.10: 70433025-70433025 on Build GRCh38
rs776435116 Chr.10: 70432894-70432894 on Build GRCh38
rs147251818 Chr.10: 70433021-70433021 on Build GRCh38
rs1056059105 Chr.10: 70432905-70432905 on Build GRCh38
rs756600339 Chr.10: 70432985-70432985 on Build GRCh38
rs759176326 Chr.10: 70433071-70433071 on Build GRCh38
rs766203379 Chr.10: 70433060-70433060 on Build GRCh38

View Full Product Details