Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 70478652-70479181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PALD1
Entrez Gene ID
27143
Gene Name
phosphatase domain containing, paladin 1
Gene Aliases
KIAA1274, PALD
UniGene
Hs.202351
Gene Chromosome Location
Chr.10: 70458542-70568450 on Build GRCh38
Transcripts
NM_014431.2 XM_011539638.2

SNP Detail

SNP ID Chromosome Location
rs576685426 Chr.10: 70479017-70479017 on Build GRCh38
rs992315261 Chr.10: 70478680-70478681 on Build GRCh38
rs943345339 Chr.10: 70478919-70478919 on Build GRCh38
rs1000930823 Chr.10: 70479056-70479056 on Build GRCh38
rs957851524 Chr.10: 70479015-70479015 on Build GRCh38
rs747299068 Chr.10: 70479159-70479159 on Build GRCh38
rs914009403 Chr.10: 70478676-70478676 on Build GRCh38
rs554824839 Chr.10: 70479122-70479122 on Build GRCh38
rs959382206 Chr.10: 70478907-70478907 on Build GRCh38
rs1038110530 Chr.10: 70478974-70478974 on Build GRCh38
rs929337266 Chr.10: 70479101-70479101 on Build GRCh38
rs967172303 Chr.10: 70478882-70478882 on Build GRCh38
rs1008779321 Chr.10: 70478863-70478863 on Build GRCh38
rs553447717 Chr.10: 70478980-70478980 on Build GRCh38
rs908385910 Chr.10: 70478950-70478950 on Build GRCh38
rs959034089 Chr.10: 70478677-70478677 on Build GRCh38
rs994005156 Chr.10: 70478780-70478780 on Build GRCh38
rs898164115 Chr.10: 70478976-70478976 on Build GRCh38
rs1032135982 Chr.10: 70479014-70479014 on Build GRCh38
rs551426642 Chr.10: 70478779-70478779 on Build GRCh38
rs925291491 Chr.10: 70478714-70478714 on Build GRCh38
rs936544565 Chr.10: 70478733-70478733 on Build GRCh38
rs1033931586 Chr.10: 70478817-70478817 on Build GRCh38
rs183948658 Chr.10: 70479036-70479036 on Build GRCh38
rs548898509 Chr.10: 70478841-70478841 on Build GRCh38
rs534768399 Chr.10: 70478918-70478918 on Build GRCh38
rs1040852452 Chr.10: 70478770-70478770 on Build GRCh38
rs1049683876 Chr.10: 70478753-70478753 on Build GRCh38
rs530865187 Chr.10: 70478834-70478834 on Build GRCh38
rs1021123899 Chr.10: 70478869-70478869 on Build GRCh38
rs990784764 Chr.10: 70479096-70479096 on Build GRCh38
rs895427350 Chr.10: 70478990-70478990 on Build GRCh38
rs762721648 Chr.10: 70478988-70478988 on Build GRCh38
rs1055746909 Chr.10: 70478731-70478731 on Build GRCh38
rs979386883 Chr.10: 70478707-70478707 on Build GRCh38
rs898198298 Chr.10: 70478829-70478829 on Build GRCh38
rs929370754 Chr.10: 70478969-70478969 on Build GRCh38
rs1047861199 Chr.10: 70478794-70478794 on Build GRCh38
rs1037974457 Chr.10: 70478825-70478825 on Build GRCh38
rs999247404 Chr.10: 70479013-70479013 on Build GRCh38
rs971693721 Chr.10: 70479117-70479117 on Build GRCh38
rs373999741 Chr.10: 70479141-70479141 on Build GRCh38
rs571278083 Chr.10: 70479002-70479002 on Build GRCh38
rs939763263 Chr.10: 70478973-70478973 on Build GRCh38
rs755453225 Chr.10: 70478723-70478723 on Build GRCh38
rs886821156 Chr.10: 70478799-70478799 on Build GRCh38
rs923276311 Chr.10: 70478965-70478965 on Build GRCh38
rs966828303 Chr.10: 70478680-70478680 on Build GRCh38
rs1014272970 Chr.10: 70479004-70479004 on Build GRCh38
rs1032465605 Chr.10: 70479063-70479063 on Build GRCh38
rs957862810 Chr.10: 70478899-70478899 on Build GRCh38
rs917707362 Chr.10: 70478706-70478706 on Build GRCh38
rs925127340 Chr.10: 70478725-70478725 on Build GRCh38
rs925261750 Chr.10: 70478903-70478903 on Build GRCh38
rs965293961 Chr.10: 70479036-70479036 on Build GRCh38
rs966463633 Chr.10: 70478850-70478850 on Build GRCh38
rs955560288 Chr.10: 70478852-70478852 on Build GRCh38
rs1003985853 Chr.10: 70478808-70478808 on Build GRCh38
rs943391732 Chr.10: 70478769-70478769 on Build GRCh38
rs910518365 Chr.10: 70478757-70478757 on Build GRCh38
rs567454859 Chr.10: 70478913-70478913 on Build GRCh38
rs925075571 Chr.10: 70478876-70478876 on Build GRCh38
rs983491741 Chr.10: 70479105-70479105 on Build GRCh38
rs949960737 Chr.10: 70478789-70478789 on Build GRCh38
rs1008497179 Chr.10: 70479021-70479021 on Build GRCh38
rs1045475945 Chr.10: 70478796-70478796 on Build GRCh38
rs979524512 Chr.10: 70478893-70478893 on Build GRCh38
rs997036599 Chr.10: 70478832-70478832 on Build GRCh38
rs991400092 Chr.10: 70478908-70478908 on Build GRCh38
rs538625999 Chr.10: 70479005-70479005 on Build GRCh38
rs950004610 Chr.10: 70478915-70478915 on Build GRCh38
rs1024994603 Chr.10: 70479111-70479111 on Build GRCh38
rs977081230 Chr.10: 70479072-70479072 on Build GRCh38
rs1055288780 Chr.10: 70478989-70478989 on Build GRCh38
rs1021512111 Chr.10: 70479035-70479035 on Build GRCh38
rs569947993 Chr.10: 70478813-70478813 on Build GRCh38
rs902708898 Chr.10: 70479049-70479049 on Build GRCh38
rs570855567 Chr.10: 70478875-70478875 on Build GRCh38
rs1006389280 Chr.10: 70478806-70478806 on Build GRCh38
rs7070313 Chr.10: 70479007-70479007 on Build GRCh38
rs959350226 Chr.10: 70479068-70479068 on Build GRCh38
rs908354204 Chr.10: 70479155-70479155 on Build GRCh38
rs1025127841 Chr.10: 70478837-70478837 on Build GRCh38
rs538276879 Chr.10: 70479097-70479097 on Build GRCh38

View Full Product Details