Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 73438238-73438736 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP3CB
Entrez Gene ID
5532
Gene Name
protein phosphatase 3 catalytic subunit beta
Gene Aliases
CALNA2, CALNB, CNA2, PP2Bbeta
UniGene
Hs.500067
Gene Chromosome Location
Chr.10: 73436428-73496024 on Build GRCh38
Transcripts
NM_001142353.2 NM_001142354.2 NM_021132.3 XM_005269944.1 XM_017016379.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1745885 c.505G>C p.D169H Chr.10: 73438328-73438328 on Build GRCh38
6754466 c.569C>T p.T190M Chr.10: 73438264-73438264 on Build GRCh38
6066418 c.493C>T p.P165S Chr.10: 73438340-73438340 on Build GRCh38
6129924 c.587A>T p.Q196L Chr.10: 73438246-73438246 on Build GRCh38
1349003 c.570G>A p.T190T Chr.10: 73438263-73438263 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148145548 Chr.10: 73438441-73438441 on Build GRCh38
rs1014038975 Chr.10: 73438445-73438445 on Build GRCh38
rs751336031 Chr.10: 73438311-73438311 on Build GRCh38
rs766155838 Chr.10: 73438308-73438308 on Build GRCh38
rs534698928 Chr.10: 73438593-73438593 on Build GRCh38
rs781230329 Chr.10: 73438327-73438327 on Build GRCh38
rs571444778 Chr.10: 73438615-73438615 on Build GRCh38
rs775907792 Chr.10: 73438268-73438268 on Build GRCh38
rs889517806 Chr.10: 73438460-73438460 on Build GRCh38
rs530036228 Chr.10: 73438507-73438507 on Build GRCh38
rs147391900 Chr.10: 73438269-73438269 on Build GRCh38
rs750093851 Chr.10: 73438296-73438296 on Build GRCh38
rs527245167 Chr.10: 73438625-73438625 on Build GRCh38
rs778959729 Chr.10: 73438259-73438259 on Build GRCh38
rs557564077 Chr.10: 73438694-73438694 on Build GRCh38
rs538756086 Chr.10: 73438622-73438622 on Build GRCh38
rs762654972 Chr.10: 73438299-73438299 on Build GRCh38
rs958280594 Chr.10: 73438604-73438604 on Build GRCh38
rs956064020 Chr.10: 73438317-73438317 on Build GRCh38
rs921992515 Chr.10: 73438667-73438668 on Build GRCh38
rs754983452 Chr.10: 73438323-73438323 on Build GRCh38
rs372419696 Chr.10: 73438277-73438277 on Build GRCh38
rs747315564 Chr.10: 73438451-73438451 on Build GRCh38
rs774900687 Chr.10: 73438642-73438642 on Build GRCh38
rs747497898 Chr.10: 73438647-73438647 on Build GRCh38
rs866711157 Chr.10: 73438592-73438592 on Build GRCh38
rs1008414784 Chr.10: 73438472-73438472 on Build GRCh38
rs753997606 Chr.10: 73438334-73438334 on Build GRCh38
rs1024461909 Chr.10: 73438504-73438504 on Build GRCh38
rs746068335 Chr.10: 73438263-73438263 on Build GRCh38
rs750020382 Chr.10: 73438468-73438469 on Build GRCh38
rs375807700 Chr.10: 73438395-73438395 on Build GRCh38
rs550798579 Chr.10: 73438283-73438283 on Build GRCh38
rs908076285 Chr.10: 73438628-73438628 on Build GRCh38
rs878903580 Chr.10: 73438469-73438469 on Build GRCh38
rs536823269 Chr.10: 73438709-73438709 on Build GRCh38
rs764774768 Chr.10: 73438290-73438290 on Build GRCh38
rs990812162 Chr.10: 73438612-73438612 on Build GRCh38
rs932061714 Chr.10: 73438668-73438668 on Build GRCh38
rs113303275 Chr.10: 73438576-73438576 on Build GRCh38
rs773768234 Chr.10: 73438501-73438504 on Build GRCh38
rs746120729 Chr.10: 73438264-73438264 on Build GRCh38
rs970771051 Chr.10: 73438620-73438620 on Build GRCh38
rs756307973 Chr.10: 73438289-73438289 on Build GRCh38
rs74983485 Chr.10: 73438584-73438584 on Build GRCh38
rs61749234 Chr.10: 73438414-73438414 on Build GRCh38
rs1043721455 Chr.10: 73438691-73438692 on Build GRCh38
rs112448442 Chr.10: 73438697-73438697 on Build GRCh38
rs1019781156 Chr.10: 73438513-73438513 on Build GRCh38
rs867079286 Chr.10: 73438463-73438463 on Build GRCh38
rs768616685 Chr.10: 73438608-73438608 on Build GRCh38
rs142001865 Chr.10: 73438613-73438613 on Build GRCh38
rs878977195 Chr.10: 73438468-73438468 on Build GRCh38
rs772332874 Chr.10: 73438431-73438431 on Build GRCh38
rs757317022 Chr.10: 73438338-73438338 on Build GRCh38

View Full Product Details