Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 73498243-73498771 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP3CB-AS1
Entrez Gene ID
101929145
Gene Name
PPP3CB antisense RNA 1 (head to head)
Gene Aliases
USP54-AS1
UniGene
Hs.434502
Gene Chromosome Location
Chr.10: 73495752-73507309 on Build GRCh38
Transcripts
NR_132103.1 NR_132104.1 NM_001320437.1 NM_152586.3 NR_135249.1 NR_135250.1 XM_005269582.2 XM_011539368.2 XM_017015759.1 XM_017015760.1 XM_017015761.1 XM_017015762.1 XM_017015763.1 XM_017015764.1 XM_017015765.1 XM_017015766.1 XM_017015767.1 XM_017015768.1 XM_017015769.1 XM_017015770.1 XM_017015771.1 XM_017015772.1 XM_017015773.1 XM_017015774.1 XM_017015775.1 XM_017015776.1 XM_017015777.1 XM_017015778.1 XM_017015779.1 XM_017015780.1 XM_017015781.1 XM_017015782.1 XM_017015783.1 XM_017015784.1 XM_017015785.1 XM_017015786.1

Gene Symbol
USP54
Entrez Gene ID
159195
Gene Name
ubiquitin specific peptidase 54
Gene Aliases
C10orf29, bA137L10.3, bA137L10.4
UniGene
Hs.657355
Gene Chromosome Location
Chr.10: 73497538-73626844 on Build GRCh38
Transcripts
NR_132103.1 NR_132104.1 NM_001320437.1 NM_152586.3 NR_135249.1 NR_135250.1 XM_005269582.2 XM_011539368.2 XM_017015759.1 XM_017015760.1 XM_017015761.1 XM_017015762.1 XM_017015763.1 XM_017015764.1 XM_017015765.1 XM_017015766.1 XM_017015767.1 XM_017015768.1 XM_017015769.1 XM_017015770.1 XM_017015771.1 XM_017015772.1 XM_017015773.1 XM_017015774.1 XM_017015775.1 XM_017015776.1 XM_017015777.1 XM_017015778.1 XM_017015779.1 XM_017015780.1 XM_017015781.1 XM_017015782.1 XM_017015783.1 XM_017015784.1 XM_017015785.1 XM_017015786.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
427886 c.4941T>C p.Y1647Y Chr.10: 73498743-73498743 on Build GRCh38
1474757 c.2271C>T p.I757I Chr.10: 73498662-73498662 on Build GRCh38
427887 c.2190T>C p.Y730Y Chr.10: 73498743-73498743 on Build GRCh38
5605311 c.5024G>A p.R1675K Chr.10: 73498660-73498660 on Build GRCh38
920312 c.4975G>A p.E1659K Chr.10: 73498709-73498709 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs900452471 Chr.10: 73498294-73498294 on Build GRCh38
rs920142190 Chr.10: 73498449-73498449 on Build GRCh38
rs979152155 Chr.10: 73498400-73498400 on Build GRCh38
rs758173085 Chr.10: 73498358-73498358 on Build GRCh38
rs530616694 Chr.10: 73498564-73498564 on Build GRCh38
rs778017475 Chr.10: 73498632-73498632 on Build GRCh38
rs1006615856 Chr.10: 73498465-73498465 on Build GRCh38
rs151120399 Chr.10: 73498676-73498676 on Build GRCh38
rs1039747995 Chr.10: 73498614-73498614 on Build GRCh38
rs781212824 Chr.10: 73498591-73498591 on Build GRCh38
rs1056999532 Chr.10: 73498407-73498407 on Build GRCh38
rs372951676 Chr.10: 73498638-73498638 on Build GRCh38
rs533065871 Chr.10: 73498426-73498426 on Build GRCh38
rs767717806 Chr.10: 73498694-73498694 on Build GRCh38
rs756018910 Chr.10: 73498681-73498681 on Build GRCh38
rs146862170 Chr.10: 73498735-73498735 on Build GRCh38
rs955092990 Chr.10: 73498309-73498309 on Build GRCh38
rs926852743 Chr.10: 73498401-73498401 on Build GRCh38
rs528324471 Chr.10: 73498577-73498577 on Build GRCh38
rs756316994 Chr.10: 73498707-73498707 on Build GRCh38
rs774886358 Chr.10: 73498650-73498650 on Build GRCh38
rs200991513 Chr.10: 73498582-73498582 on Build GRCh38
rs186234044 Chr.10: 73498432-73498432 on Build GRCh38
rs1023713094 Chr.10: 73498424-73498424 on Build GRCh38
rs2990138 Chr.10: 73498563-73498563 on Build GRCh38
rs2990139 Chr.10: 73498565-73498565 on Build GRCh38
rs749621251 Chr.10: 73498635-73498635 on Build GRCh38
rs541028822 Chr.10: 73498368-73498368 on Build GRCh38
rs562356083 Chr.10: 73498292-73498292 on Build GRCh38
rs537649468 Chr.10: 73498682-73498682 on Build GRCh38
rs975627784 Chr.10: 73498288-73498288 on Build GRCh38
rs779247821 Chr.10: 73498732-73498732 on Build GRCh38
rs772513150 Chr.10: 73498734-73498734 on Build GRCh38
rs921269117 Chr.10: 73498641-73498641 on Build GRCh38
rs751087693 Chr.10: 73498428-73498428 on Build GRCh38
rs746487888 Chr.10: 73498743-73498743 on Build GRCh38
rs766382599 Chr.10: 73498661-73498662 on Build GRCh38
rs996784817 Chr.10: 73498520-73498520 on Build GRCh38
rs771640964 Chr.10: 73498667-73498667 on Build GRCh38
rs758342922 Chr.10: 73498689-73498689 on Build GRCh38
rs900939277 Chr.10: 73498422-73498422 on Build GRCh38
rs769053365 Chr.10: 73498692-73498695 on Build GRCh38
rs913693722 Chr.10: 73498369-73498369 on Build GRCh38
rs574245381 Chr.10: 73498611-73498611 on Build GRCh38
rs938296717 Chr.10: 73498406-73498406 on Build GRCh38
rs750450548 Chr.10: 73498686-73498686 on Build GRCh38
rs58662799 Chr.10: 73498299-73498299 on Build GRCh38
rs942139319 Chr.10: 73498747-73498747 on Build GRCh38
rs1017622281 Chr.10: 73498467-73498467 on Build GRCh38
rs553102168 Chr.10: 73498714-73498714 on Build GRCh38
rs748322347 Chr.10: 73498610-73498610 on Build GRCh38
rs750356416 Chr.10: 73498600-73498600 on Build GRCh38
rs41315012 Chr.10: 73498679-73498679 on Build GRCh38
rs746363588 Chr.10: 73498665-73498665 on Build GRCh38
rs899719856 Chr.10: 73498470-73498470 on Build GRCh38
rs117429793 Chr.10: 73498685-73498685 on Build GRCh38
rs200429219 Chr.10: 73498727-73498727 on Build GRCh38
rs563528927 Chr.10: 73498433-73498433 on Build GRCh38
rs201563164 Chr.10: 73498716-73498716 on Build GRCh38
rs557967859 Chr.10: 73498671-73498671 on Build GRCh38
rs142765962 Chr.10: 73498670-73498670 on Build GRCh38
rs755165491 Chr.10: 73498601-73498601 on Build GRCh38
rs567234326 Chr.10: 73498567-73498567 on Build GRCh38
rs781415469 Chr.10: 73498609-73498609 on Build GRCh38
rs142582494 Chr.10: 73498371-73498371 on Build GRCh38
rs376126537 Chr.10: 73498443-73498443 on Build GRCh38

View Full Product Details