Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 73997892-73998381 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VCL
Entrez Gene ID
7414
Gene Name
vinculin
Gene Aliases
CMD1W, CMH15, HEL114, MV, MVCL
UniGene
Hs.643896
Gene Chromosome Location
Chr.10: 73998114-74120160 on Build GRCh38
Transcripts
NM_003373.3 NM_014000.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5988649 c.156C>A p.S52R Chr.10: 73998363-73998363 on Build GRCh38
4564136 c.100_101GG>AA p.G34N Chr.10: 73998307-73998308 on Build GRCh38
6345210 c.96G>T p.V32V Chr.10: 73998303-73998303 on Build GRCh38
5642458 c.13C>A p.H5N Chr.10: 73998220-73998220 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs976204657 Chr.10: 73997913-73997913 on Build GRCh38
rs368934359 Chr.10: 73998175-73998175 on Build GRCh38
rs759312175 Chr.10: 73998246-73998246 on Build GRCh38
rs918335824 Chr.10: 73998207-73998207 on Build GRCh38
rs978938406 Chr.10: 73998063-73998064 on Build GRCh38
rs397517240 Chr.10: 73998238-73998238 on Build GRCh38
rs888352872 Chr.10: 73998059-73998059 on Build GRCh38
rs144080529 Chr.10: 73998327-73998327 on Build GRCh38
rs1044698656 Chr.10: 73998108-73998108 on Build GRCh38
rs1019638297 Chr.10: 73998024-73998024 on Build GRCh38
rs554116863 Chr.10: 73998337-73998337 on Build GRCh38
rs568175141 Chr.10: 73998306-73998306 on Build GRCh38
rs780482320 Chr.10: 73998195-73998195 on Build GRCh38
rs749266193 Chr.10: 73998240-73998240 on Build GRCh38
rs1031820588 Chr.10: 73998333-73998333 on Build GRCh38
rs139329923 Chr.10: 73998310-73998310 on Build GRCh38
rs561721542 Chr.10: 73998096-73998096 on Build GRCh38
rs774195260 Chr.10: 73998243-73998243 on Build GRCh38
rs765704599 Chr.10: 73998180-73998180 on Build GRCh38
rs572664187 Chr.10: 73997960-73997960 on Build GRCh38
rs3812625 Chr.10: 73997944-73997944 on Build GRCh38
rs988923387 Chr.10: 73998107-73998107 on Build GRCh38
rs370879754 Chr.10: 73998148-73998148 on Build GRCh38
rs1030881347 Chr.10: 73997914-73997914 on Build GRCh38
rs930443014 Chr.10: 73998116-73998116 on Build GRCh38
rs761931436 Chr.10: 73998171-73998171 on Build GRCh38
rs771251552 Chr.10: 73997935-73997935 on Build GRCh38
rs146750460 Chr.10: 73998237-73998237 on Build GRCh38
rs886047215 Chr.10: 73998146-73998146 on Build GRCh38
rs992431086 Chr.10: 73998100-73998100 on Build GRCh38
rs1004201186 Chr.10: 73998023-73998023 on Build GRCh38
rs1030410681 Chr.10: 73998095-73998095 on Build GRCh38
rs770570643 Chr.10: 73998241-73998241 on Build GRCh38
rs914816340 Chr.10: 73997928-73997928 on Build GRCh38
rs750838330 Chr.10: 73998321-73998321 on Build GRCh38
rs1044200015 Chr.10: 73997948-73997948 on Build GRCh38
rs536893103 Chr.10: 73998158-73998158 on Build GRCh38
rs866262635 Chr.10: 73998328-73998328 on Build GRCh38
rs1039626542 Chr.10: 73998341-73998341 on Build GRCh38
rs764871020 Chr.10: 73998227-73998227 on Build GRCh38
rs1004316068 Chr.10: 73998151-73998151 on Build GRCh38
rs929741279 Chr.10: 73998211-73998211 on Build GRCh38
rs200733607 Chr.10: 73998288-73998288 on Build GRCh38
rs547337176 Chr.10: 73998104-73998104 on Build GRCh38
rs1006836590 Chr.10: 73998065-73998065 on Build GRCh38
rs934309901 Chr.10: 73997926-73997926 on Build GRCh38
rs1048292068 Chr.10: 73998036-73998036 on Build GRCh38
rs529010073 Chr.10: 73998097-73998097 on Build GRCh38
rs997969604 Chr.10: 73998089-73998089 on Build GRCh38
rs900508032 Chr.10: 73998079-73998079 on Build GRCh38
rs111776449 Chr.10: 73998068-73998069 on Build GRCh38
rs750972760 Chr.10: 73998182-73998182 on Build GRCh38
rs887160032 Chr.10: 73998140-73998140 on Build GRCh38
rs546209689 Chr.10: 73998025-73998025 on Build GRCh38
rs776882033 Chr.10: 73998274-73998274 on Build GRCh38
rs576751733 Chr.10: 73998048-73998048 on Build GRCh38
rs548768602 Chr.10: 73998199-73998199 on Build GRCh38
rs988975510 Chr.10: 73997925-73997925 on Build GRCh38
rs938365596 Chr.10: 73998149-73998149 on Build GRCh38
rs992545879 Chr.10: 73998343-73998343 on Build GRCh38
rs886047216 Chr.10: 73998362-73998362 on Build GRCh38
rs924352537 Chr.10: 73998086-73998086 on Build GRCh38
rs887212237 Chr.10: 73998021-73998021 on Build GRCh38
rs753870747 Chr.10: 73998213-73998213 on Build GRCh38
rs559106123 Chr.10: 73998111-73998111 on Build GRCh38
rs1017993550 Chr.10: 73998078-73998078 on Build GRCh38
rs947667981 Chr.10: 73997938-73997938 on Build GRCh38
rs956234820 Chr.10: 73997921-73997921 on Build GRCh38
rs564688063 Chr.10: 73998038-73998038 on Build GRCh38
rs896943378 Chr.10: 73998026-73998026 on Build GRCh38
rs750937217 Chr.10: 73998288-73998289 on Build GRCh38
rs533231503 Chr.10: 73998142-73998142 on Build GRCh38
rs794729189 Chr.10: 73998257-73998257 on Build GRCh38
rs551500830 Chr.10: 73997982-73997982 on Build GRCh38
rs929792202 Chr.10: 73998032-73998032 on Build GRCh38
rs968920523 Chr.10: 73998122-73998122 on Build GRCh38
rs777811020 Chr.10: 73998239-73998239 on Build GRCh38
rs543542145 Chr.10: 73998076-73998076 on Build GRCh38
rs754654170 Chr.10: 73998203-73998203 on Build GRCh38
rs763933845 Chr.10: 73998119-73998119 on Build GRCh38
rs775062250 Chr.10: 73998252-73998252 on Build GRCh38
rs551418878 Chr.10: 73998144-73998144 on Build GRCh38

View Full Product Details