Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 75401303-75401801 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF503
Entrez Gene ID
84858
Gene Name
zinc finger protein 503
Gene Aliases
NOLZ-1, NOLZ1, Nlz2
UniGene
Hs.195710
Gene Chromosome Location
Chr.10: 75279726-75401894 on Build GRCh38
Transcripts
NM_032772.5 NR_120651.1 NR_024421.1

Gene Symbol
ZNF503-AS2
Entrez Gene ID
100131213
Gene Name
ZNF503 antisense RNA 2
Gene Aliases
C10orf41, NCRNA00245
UniGene
Hs.534598
Gene Chromosome Location
Chr.10: 75401528-75408982 on Build GRCh38
Transcripts
NM_032772.5 NR_120651.1 NR_024421.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3807752 c.354T>G p.P118P Chr.10: 75401354-75401354 on Build GRCh38
6458476 c.355_357delCCG p.P124delP Chr.10: 75401355-75401357 on Build GRCh38
5958579 c.77_78ins12 p.G27_A28insGGGG Chr.10: 75401342-75401343 on Build GRCh38
6417024 c.419T>C p.M140T Chr.10: 75401419-75401419 on Build GRCh38
3807751 c.66A>C p.G22G Chr.10: 75401354-75401354 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs998080108 Chr.10: 75401479-75401479 on Build GRCh38
rs897236624 Chr.10: 75401330-75401330 on Build GRCh38
rs546695592 Chr.10: 75401768-75401768 on Build GRCh38
rs1035540527 Chr.10: 75401426-75401426 on Build GRCh38
rs923803746 Chr.10: 75401375-75401375 on Build GRCh38
rs367599528 Chr.10: 75401393-75401393 on Build GRCh38
rs956436529 Chr.10: 75401377-75401377 on Build GRCh38
rs772844174 Chr.10: 75401352-75401354 on Build GRCh38
rs938950499 Chr.10: 75401418-75401418 on Build GRCh38
rs762840921 Chr.10: 75401463-75401463 on Build GRCh38
rs780118403 Chr.10: 75401348-75401349 on Build GRCh38
rs552779373 Chr.10: 75401448-75401448 on Build GRCh38
rs750546401 Chr.10: 75401445-75401446 on Build GRCh38
rs960335038 Chr.10: 75401760-75401760 on Build GRCh38
rs111533383 Chr.10: 75401718-75401718 on Build GRCh38
rs560688833 Chr.10: 75401754-75401754 on Build GRCh38
rs1041379168 Chr.10: 75401656-75401656 on Build GRCh38
rs12779861 Chr.10: 75401579-75401579 on Build GRCh38
rs760830242 Chr.10: 75401539-75401539 on Build GRCh38
rs927649736 Chr.10: 75401553-75401553 on Build GRCh38
rs1003001504 Chr.10: 75401683-75401683 on Build GRCh38
rs901156171 Chr.10: 75401670-75401670 on Build GRCh38
rs998151625 Chr.10: 75401678-75401678 on Build GRCh38
rs901082706 Chr.10: 75401467-75401467 on Build GRCh38
rs972067667 Chr.10: 75401600-75401600 on Build GRCh38
rs958572572 Chr.10: 75401558-75401558 on Build GRCh38
rs574085187 Chr.10: 75401428-75401428 on Build GRCh38
rs969582073 Chr.10: 75401544-75401544 on Build GRCh38
rs1007318667 Chr.10: 75401351-75401351 on Build GRCh38
rs780357728 Chr.10: 75401603-75401604 on Build GRCh38
rs11542744 Chr.10: 75401354-75401354 on Build GRCh38
rs779751573 Chr.10: 75401401-75401401 on Build GRCh38
rs989166758 Chr.10: 75401378-75401378 on Build GRCh38
rs12780004 Chr.10: 75401541-75401541 on Build GRCh38
rs980952787 Chr.10: 75401741-75401741 on Build GRCh38
rs1054041251 Chr.10: 75401630-75401630 on Build GRCh38
rs767764991 Chr.10: 75401372-75401374 on Build GRCh38
rs1030892401 Chr.10: 75401492-75401492 on Build GRCh38
rs918858464 Chr.10: 75401582-75401582 on Build GRCh38
rs878911254 Chr.10: 75401339-75401339 on Build GRCh38
rs544944962 Chr.10: 75401603-75401603 on Build GRCh38
rs537447500 Chr.10: 75401394-75401394 on Build GRCh38
rs1016174408 Chr.10: 75401569-75401569 on Build GRCh38
rs963237484 Chr.10: 75401725-75401725 on Build GRCh38
rs746644250 Chr.10: 75401415-75401415 on Build GRCh38
rs569991258 Chr.10: 75401368-75401368 on Build GRCh38
rs1011267827 Chr.10: 75401516-75401518 on Build GRCh38
rs750763743 Chr.10: 75401389-75401389 on Build GRCh38
rs1007851962 Chr.10: 75401654-75401676 on Build GRCh38
rs897352608 Chr.10: 75401438-75401438 on Build GRCh38
rs1006855204 Chr.10: 75401452-75401452 on Build GRCh38
rs1022583407 Chr.10: 75401737-75401737 on Build GRCh38
rs1049042705 Chr.10: 75401614-75401614 on Build GRCh38
rs1011298922 Chr.10: 75401382-75401382 on Build GRCh38
rs775727472 Chr.10: 75401500-75401500 on Build GRCh38
rs866011897 Chr.10: 75401668-75401668 on Build GRCh38
rs548026488 Chr.10: 75401329-75401329 on Build GRCh38
rs759155569 Chr.10: 75401757-75401757 on Build GRCh38
rs762649774 Chr.10: 75401342-75401343 on Build GRCh38
rs113760752 Chr.10: 75401335-75401335 on Build GRCh38
rs764097121 Chr.10: 75401331-75401331 on Build GRCh38
rs1034786627 Chr.10: 75401755-75401755 on Build GRCh38
rs892801309 Chr.10: 75401514-75401514 on Build GRCh38
rs755325857 Chr.10: 75401733-75401733 on Build GRCh38
rs761931033 Chr.10: 75401606-75401606 on Build GRCh38
rs889310130 Chr.10: 75401507-75401507 on Build GRCh38
rs902901102 Chr.10: 75401525-75401525 on Build GRCh38
rs544101706 Chr.10: 75401671-75401671 on Build GRCh38
rs563598299 Chr.10: 75401654-75401654 on Build GRCh38
rs771606452 Chr.10: 75401349-75401357 on Build GRCh38
rs533859340 Chr.10: 75401343-75401348 on Build GRCh38
rs577402851 Chr.10: 75401466-75401466 on Build GRCh38
rs963330357 Chr.10: 75401573-75401573 on Build GRCh38
rs930148692 Chr.10: 75401446-75401446 on Build GRCh38
rs892384143 Chr.10: 75401711-75401711 on Build GRCh38
rs976544550 Chr.10: 75401374-75401374 on Build GRCh38
rs528211436 Chr.10: 75401765-75401765 on Build GRCh38
rs938897456 Chr.10: 75401566-75401566 on Build GRCh38
rs1039687649 Chr.10: 75401664-75401664 on Build GRCh38
rs1056916068 Chr.10: 75401686-75401686 on Build GRCh38
rs760375543 Chr.10: 75401354-75401355 on Build GRCh38
rs894296640 Chr.10: 75401691-75401691 on Build GRCh38
rs764195520 Chr.10: 75401548-75401548 on Build GRCh38
rs541089755 Chr.10: 75401447-75401447 on Build GRCh38
rs914988528 Chr.10: 75401384-75401384 on Build GRCh38
rs12780400 Chr.10: 75401709-75401709 on Build GRCh38
rs767771319 Chr.10: 75401327-75401327 on Build GRCh38
rs956509247 Chr.10: 75401520-75401520 on Build GRCh38
rs764457806 Chr.10: 75401355-75401360 on Build GRCh38
rs374168185 Chr.10: 75401354-75401359 on Build GRCh38
rs560022052 Chr.10: 75401775-75401775 on Build GRCh38
rs868261966 Chr.10: 75401436-75401436 on Build GRCh38
rs1026695948 Chr.10: 75401744-75401744 on Build GRCh38
rs778366983 Chr.10: 75401557-75401557 on Build GRCh38
rs773344807 Chr.10: 75401450-75401450 on Build GRCh38
rs1054050337 Chr.10: 75401513-75401513 on Build GRCh38
rs12780018 Chr.10: 75401570-75401570 on Build GRCh38
rs555427152 Chr.10: 75401403-75401403 on Build GRCh38
rs888538242 Chr.10: 75401629-75401629 on Build GRCh38
rs555023176 Chr.10: 75401756-75401756 on Build GRCh38
rs943580876 Chr.10: 75401511-75401511 on Build GRCh38
rs756340568 Chr.10: 75401546-75401565 on Build GRCh38
rs775648475 Chr.10: 75401355-75401357 on Build GRCh38
rs909182760 Chr.10: 75401446-75401446 on Build GRCh38
rs910679698 Chr.10: 75401627-75401627 on Build GRCh38
rs993056674 Chr.10: 75401567-75401567 on Build GRCh38
rs958847085 Chr.10: 75401431-75401431 on Build GRCh38
rs1057308331 Chr.10: 75401427-75401427 on Build GRCh38
rs867775264 Chr.10: 75401390-75401390 on Build GRCh38
rs969153591 Chr.10: 75401739-75401739 on Build GRCh38
rs367698599 Chr.10: 75401721-75401722 on Build GRCh38
rs1012727370 Chr.10: 75401693-75401693 on Build GRCh38
rs987577186 Chr.10: 75401617-75401617 on Build GRCh38
rs540164655 Chr.10: 75401360-75401360 on Build GRCh38
rs576950953 Chr.10: 75401366-75401366 on Build GRCh38
rs866977941 Chr.10: 75401440-75401440 on Build GRCh38
rs761979896 Chr.10: 75401368-75401369 on Build GRCh38
rs1027382680 Chr.10: 75401334-75401334 on Build GRCh38
rs989726631 Chr.10: 75401523-75401523 on Build GRCh38
rs575403442 Chr.10: 75401679-75401679 on Build GRCh38
rs752148679 Chr.10: 75401357-75401358 on Build GRCh38
rs1002643447 Chr.10: 75401530-75401530 on Build GRCh38
rs536270097 Chr.10: 75401732-75401732 on Build GRCh38

View Full Product Details