Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 79555719-79556218 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SFTPA2
Entrez Gene ID
729238
Gene Name
surfactant protein A2
Gene Aliases
COLEC5, PSAP, PSP-A, PSPA, SFTP1, SFTPA2B, SP-2A, SP-A, SPA2, SPAII
UniGene
Hs.523084
Gene Chromosome Location
Chr.10: 79555852-79560461 on Build GRCh38
Transcripts
NM_001098668.3 NM_001320813.1 NM_001320814.1 XM_005270128.3 XM_005270132.3 XM_011540124.1 XM_011540125.1 XM_017016608.1

SNP Detail

SNP ID Chromosome Location
rs1050576871 Chr.10: 79555967-79555967 on Build GRCh38
rs1059153 Chr.10: 79556110-79556110 on Build GRCh38
rs1059204 Chr.10: 79555974-79555974 on Build GRCh38
rs968655855 Chr.10: 79555845-79555845 on Build GRCh38
rs4794 Chr.10: 79556116-79556116 on Build GRCh38
rs548135801 Chr.10: 79556071-79556071 on Build GRCh38
rs989035379 Chr.10: 79555884-79555884 on Build GRCh38
rs955921193 Chr.10: 79555879-79555879 on Build GRCh38
rs1010112591 Chr.10: 79556132-79556132 on Build GRCh38
rs1059155 Chr.10: 79556100-79556100 on Build GRCh38
rs1059193 Chr.10: 79555991-79555991 on Build GRCh38
rs765030803 Chr.10: 79556065-79556065 on Build GRCh38
rs1049703999 Chr.10: 79555752-79555752 on Build GRCh38
rs1059207 Chr.10: 79555971-79555971 on Build GRCh38
rs940341116 Chr.10: 79555906-79555906 on Build GRCh38
rs974797553 Chr.10: 79556162-79556162 on Build GRCh38
rs1008956806 Chr.10: 79556045-79556045 on Build GRCh38
rs956936942 Chr.10: 79555811-79555811 on Build GRCh38
rs1059186 Chr.10: 79556019-79556019 on Build GRCh38
rs930905002 Chr.10: 79555922-79555922 on Build GRCh38
rs966128194 Chr.10: 79556159-79556159 on Build GRCh38
rs1015367910 Chr.10: 79555774-79555774 on Build GRCh38
rs1004388810 Chr.10: 79555771-79555771 on Build GRCh38
rs750066206 Chr.10: 79556087-79556087 on Build GRCh38
rs375397563 Chr.10: 79555741-79555741 on Build GRCh38
rs1059154 Chr.10: 79556101-79556101 on Build GRCh38
rs1059192 Chr.10: 79555997-79555997 on Build GRCh38
rs775783346 Chr.10: 79555899-79555899 on Build GRCh38
rs546458497 Chr.10: 79556033-79556033 on Build GRCh38
rs1000087773 Chr.10: 79555859-79555859 on Build GRCh38
rs974746643 Chr.10: 79555895-79555895 on Build GRCh38
rs17884933 Chr.10: 79555966-79555966 on Build GRCh38
rs189758520 Chr.10: 79556069-79556069 on Build GRCh38
rs921947177 Chr.10: 79556174-79556174 on Build GRCh38
rs1059190 Chr.10: 79556011-79556011 on Build GRCh38
rs1057026896 Chr.10: 79555995-79555995 on Build GRCh38
rs528969568 Chr.10: 79556175-79556175 on Build GRCh38
rs940898164 Chr.10: 79555968-79555968 on Build GRCh38
rs899417594 Chr.10: 79555956-79555956 on Build GRCh38
rs182060777 Chr.10: 79556125-79556125 on Build GRCh38
rs978691236 Chr.10: 79556143-79556143 on Build GRCh38
rs1059213 Chr.10: 79555916-79555916 on Build GRCh38
rs1032957275 Chr.10: 79556124-79556124 on Build GRCh38
rs532762756 Chr.10: 79556027-79556027 on Build GRCh38
rs774956394 Chr.10: 79555756-79555756 on Build GRCh38
rs561551858 Chr.10: 79556115-79556115 on Build GRCh38
rs912011633 Chr.10: 79555885-79555885 on Build GRCh38
rs905512052 Chr.10: 79556103-79556103 on Build GRCh38
rs186887288 Chr.10: 79555850-79555850 on Build GRCh38
rs1059164 Chr.10: 79556078-79556078 on Build GRCh38
rs577722028 Chr.10: 79555939-79555939 on Build GRCh38
rs574839970 Chr.10: 79555878-79555878 on Build GRCh38
rs921896293 Chr.10: 79555914-79555914 on Build GRCh38
rs1036025269 Chr.10: 79555942-79555942 on Build GRCh38
rs895571513 Chr.10: 79556060-79556060 on Build GRCh38
rs551668574 Chr.10: 79556061-79556061 on Build GRCh38
rs563192043 Chr.10: 79556066-79556066 on Build GRCh38
rs1033275717 Chr.10: 79555861-79555861 on Build GRCh38
rs544574632 Chr.10: 79555853-79555853 on Build GRCh38
rs759913253 Chr.10: 79555820-79555820 on Build GRCh38
rs1018752059 Chr.10: 79556148-79556148 on Build GRCh38
rs1022546448 Chr.10: 79555835-79555835 on Build GRCh38
rs540749007 Chr.10: 79555886-79555886 on Build GRCh38
rs559885358 Chr.10: 79555903-79555903 on Build GRCh38
rs1013914767 Chr.10: 79555827-79555827 on Build GRCh38
rs968582845 Chr.10: 79556133-79556133 on Build GRCh38
rs1059151 Chr.10: 79556149-79556149 on Build GRCh38
rs889302512 Chr.10: 79556016-79556016 on Build GRCh38
rs142992188 Chr.10: 79555858-79555858 on Build GRCh38
rs1059200 Chr.10: 79555980-79555980 on Build GRCh38

View Full Product Details